Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27501362-27501876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN7C
Entrez Gene ID
55327
Gene Name
lin-7 homolog C, crumbs cell polarity complex component
Gene Aliases
LIN-7-C, LIN-7C, MALS-3, MALS3, VELI3
UniGene
Hs.91393
Gene Chromosome Location
Chr.11: 27494418-27506779 on Build GRCh38
Transcripts
NM_018362.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926142 c.212C>T p.A71V Chr.11: 27501511-27501511 on Build GRCh38
6294153 c.179C>A p.T60N Chr.11: 27501544-27501544 on Build GRCh38
926143 c.130A>C p.S44R Chr.11: 27501828-27501828 on Build GRCh38
7277439 c.145G>A p.A49T Chr.11: 27501813-27501813 on Build GRCh38
4032216 c.216C>T p.N72N Chr.11: 27501507-27501507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773182363 Chr.11: 27501787-27501789 on Build GRCh38
rs186658795 Chr.11: 27501744-27501744 on Build GRCh38
rs986503674 Chr.11: 27501419-27501419 on Build GRCh38
rs753023492 Chr.11: 27501521-27501521 on Build GRCh38
rs769367980 Chr.11: 27501587-27501587 on Build GRCh38
rs200707615 Chr.11: 27501550-27501550 on Build GRCh38
rs11819903 Chr.11: 27501636-27501636 on Build GRCh38
rs770404740 Chr.11: 27501752-27501752 on Build GRCh38
rs531627200 Chr.11: 27501693-27501693 on Build GRCh38
rs550352232 Chr.11: 27501639-27501640 on Build GRCh38
rs772737920 Chr.11: 27501602-27501602 on Build GRCh38
rs903792260 Chr.11: 27501749-27501749 on Build GRCh38
rs554724542 Chr.11: 27501847-27501847 on Build GRCh38
rs917413658 Chr.11: 27501695-27501695 on Build GRCh38
rs567511930 Chr.11: 27501660-27501660 on Build GRCh38
rs765399060 Chr.11: 27501818-27501818 on Build GRCh38
rs760428610 Chr.11: 27501840-27501841 on Build GRCh38
rs780833666 Chr.11: 27501585-27501585 on Build GRCh38
rs772647960 Chr.11: 27501500-27501500 on Build GRCh38
rs748746985 Chr.11: 27501604-27501604 on Build GRCh38
rs370324945 Chr.11: 27501788-27501788 on Build GRCh38
rs10835188 Chr.11: 27501639-27501639 on Build GRCh38
rs774228244 Chr.11: 27501615-27501616 on Build GRCh38
rs766832039 Chr.11: 27501507-27501507 on Build GRCh38
rs768769166 Chr.11: 27501467-27501470 on Build GRCh38
rs200200710 Chr.11: 27501555-27501555 on Build GRCh38
rs759947909 Chr.11: 27501766-27501766 on Build GRCh38
rs955151552 Chr.11: 27501398-27501398 on Build GRCh38
rs869267881 Chr.11: 27501643-27501644 on Build GRCh38
rs908005260 Chr.11: 27501469-27501469 on Build GRCh38
rs746887115 Chr.11: 27501582-27501582 on Build GRCh38
rs538775685 Chr.11: 27501792-27501792 on Build GRCh38
rs190288799 Chr.11: 27501508-27501508 on Build GRCh38
rs908028442 Chr.11: 27501562-27501562 on Build GRCh38
rs753932786 Chr.11: 27501552-27501552 on Build GRCh38
rs781042537 Chr.11: 27501852-27501852 on Build GRCh38
rs181529416 Chr.11: 27501510-27501510 on Build GRCh38
rs769347453 Chr.11: 27501479-27501479 on Build GRCh38
rs547350739 Chr.11: 27501742-27501743 on Build GRCh38
rs763454591 Chr.11: 27501506-27501506 on Build GRCh38
rs374735648 Chr.11: 27501850-27501850 on Build GRCh38
rs796172717 Chr.11: 27501636-27501639 on Build GRCh38
rs756234972 Chr.11: 27501522-27501522 on Build GRCh38
rs754864719 Chr.11: 27501584-27501584 on Build GRCh38
rs754812273 Chr.11: 27501840-27501840 on Build GRCh38
rs747749368 Chr.11: 27501458-27501458 on Build GRCh38
rs747846656 Chr.11: 27501586-27501586 on Build GRCh38
rs193060268 Chr.11: 27501614-27501614 on Build GRCh38
rs1056949718 Chr.11: 27501674-27501674 on Build GRCh38
rs774991110 Chr.11: 27501759-27501759 on Build GRCh38
rs151336878 Chr.11: 27501617-27501618 on Build GRCh38
rs757455561 Chr.11: 27501815-27501815 on Build GRCh38
rs549029019 Chr.11: 27501648-27501648 on Build GRCh38
rs761012770 Chr.11: 27501793-27501793 on Build GRCh38
rs780192896 Chr.11: 27501566-27501566 on Build GRCh38
rs756418497 Chr.11: 27501422-27501422 on Build GRCh38
rs549851108 Chr.11: 27501709-27501709 on Build GRCh38
rs71050915 Chr.11: 27501635-27501636 on Build GRCh38
rs964053312 Chr.11: 27501436-27501436 on Build GRCh38
rs67138756 Chr.11: 27501618-27501619 on Build GRCh38
rs113112151 Chr.11: 27501730-27501730 on Build GRCh38
rs764364774 Chr.11: 27501804-27501804 on Build GRCh38
rs374984683 Chr.11: 27501464-27501464 on Build GRCh38
rs751568986 Chr.11: 27501830-27501830 on Build GRCh38

View Full Product Details