Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65883963-65884455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBP
Entrez Gene ID
9158
Gene Name
FGF1 intracellular binding protein
Gene Aliases
FGFIBP, FIBP-1
UniGene
Hs.7768
Gene Chromosome Location
Chr.11: 65883740-65888539 on Build GRCh38
Transcripts
NM_004214.4 NM_198897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252440 c.1045C>T p.R349C Chr.11: 65884024-65884024 on Build GRCh38
7368281 c.1012C>T p.L338F Chr.11: 65884405-65884405 on Build GRCh38
6207819 c.963C>T p.L321L Chr.11: 65884454-65884454 on Build GRCh38
1356096 c.966C>T p.S322S Chr.11: 65884451-65884451 on Build GRCh38
6284772 c.1032G>T p.Q344H Chr.11: 65884037-65884037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758150067 Chr.11: 65884061-65884061 on Build GRCh38
rs958932085 Chr.11: 65884166-65884166 on Build GRCh38
rs754335623 Chr.11: 65884317-65884317 on Build GRCh38
rs776028169 Chr.11: 65884354-65884354 on Build GRCh38
rs565550626 Chr.11: 65884054-65884054 on Build GRCh38
rs892393123 Chr.11: 65884298-65884298 on Build GRCh38
rs572368414 Chr.11: 65884337-65884337 on Build GRCh38
rs975349638 Chr.11: 65884124-65884124 on Build GRCh38
rs113243481 Chr.11: 65884045-65884045 on Build GRCh38
rs752879715 Chr.11: 65884332-65884332 on Build GRCh38
rs765308549 Chr.11: 65884329-65884329 on Build GRCh38
rs912295107 Chr.11: 65884211-65884211 on Build GRCh38
rs899180126 Chr.11: 65883994-65883994 on Build GRCh38
rs977078492 Chr.11: 65884179-65884179 on Build GRCh38
rs959957132 Chr.11: 65884430-65884430 on Build GRCh38
rs576180623 Chr.11: 65884293-65884293 on Build GRCh38
rs150979219 Chr.11: 65884328-65884328 on Build GRCh38
rs753754162 Chr.11: 65883995-65883995 on Build GRCh38
rs200771731 Chr.11: 65884392-65884392 on Build GRCh38
rs992557170 Chr.11: 65884432-65884432 on Build GRCh38
rs1034540674 Chr.11: 65884088-65884088 on Build GRCh38
rs746605016 Chr.11: 65884076-65884076 on Build GRCh38
rs372881353 Chr.11: 65884384-65884384 on Build GRCh38
rs200002664 Chr.11: 65884003-65884003 on Build GRCh38
rs779728742 Chr.11: 65884423-65884423 on Build GRCh38
rs943811352 Chr.11: 65884221-65884221 on Build GRCh38
rs929596752 Chr.11: 65884175-65884175 on Build GRCh38
rs1054074263 Chr.11: 65884296-65884296 on Build GRCh38
rs368941233 Chr.11: 65884027-65884027 on Build GRCh38
rs36090851 Chr.11: 65884191-65884191 on Build GRCh38
rs200017030 Chr.11: 65884004-65884004 on Build GRCh38
rs658938 Chr.11: 65884359-65884359 on Build GRCh38
rs769087439 Chr.11: 65884361-65884361 on Build GRCh38
rs34402671 Chr.11: 65884206-65884206 on Build GRCh38
rs750513587 Chr.11: 65884042-65884042 on Build GRCh38
rs763382803 Chr.11: 65884011-65884011 on Build GRCh38
rs556385725 Chr.11: 65884290-65884290 on Build GRCh38
rs140853801 Chr.11: 65883990-65883990 on Build GRCh38
rs202016111 Chr.11: 65884090-65884090 on Build GRCh38
rs752598076 Chr.11: 65884095-65884095 on Build GRCh38
rs377110830 Chr.11: 65884418-65884418 on Build GRCh38
rs767769226 Chr.11: 65884374-65884374 on Build GRCh38
rs974274568 Chr.11: 65884412-65884412 on Build GRCh38
rs773632783 Chr.11: 65884144-65884144 on Build GRCh38
rs372356056 Chr.11: 65884005-65884005 on Build GRCh38
rs1139703 Chr.11: 65884031-65884031 on Build GRCh38
rs777564474 Chr.11: 65884068-65884068 on Build GRCh38
rs376028477 Chr.11: 65883984-65883984 on Build GRCh38
rs36080962 Chr.11: 65883993-65883993 on Build GRCh38
rs534578285 Chr.11: 65884080-65884080 on Build GRCh38
rs890444172 Chr.11: 65884342-65884342 on Build GRCh38
rs968623000 Chr.11: 65884405-65884405 on Build GRCh38
rs767871039 Chr.11: 65884094-65884094 on Build GRCh38
rs780872889 Chr.11: 65884406-65884406 on Build GRCh38
rs769233818 Chr.11: 65884348-65884348 on Build GRCh38
rs936528998 Chr.11: 65884255-65884255 on Build GRCh38
rs2231893 Chr.11: 65884018-65884018 on Build GRCh38
rs763807212 Chr.11: 65884394-65884394 on Build GRCh38
rs769283257 Chr.11: 65884429-65884429 on Build GRCh38
rs543932342 Chr.11: 65884402-65884402 on Build GRCh38
rs954777696 Chr.11: 65884023-65884023 on Build GRCh38
rs750662527 Chr.11: 65884377-65884377 on Build GRCh38
rs375017310 Chr.11: 65884436-65884436 on Build GRCh38
rs142371864 Chr.11: 65884327-65884327 on Build GRCh38
rs746780051 Chr.11: 65884363-65884365 on Build GRCh38
rs947052249 Chr.11: 65884177-65884177 on Build GRCh38
rs116618583 Chr.11: 65884403-65884403 on Build GRCh38
rs561408158 Chr.11: 65883986-65883986 on Build GRCh38
rs146547919 Chr.11: 65884046-65884046 on Build GRCh38
rs376780734 Chr.11: 65884084-65884084 on Build GRCh38
rs774861566 Chr.11: 65884367-65884367 on Build GRCh38
rs780770250 Chr.11: 65884089-65884089 on Build GRCh38
rs1037931969 Chr.11: 65884301-65884301 on Build GRCh38
rs920191684 Chr.11: 65884174-65884174 on Build GRCh38
rs760775903 Chr.11: 65884381-65884381 on Build GRCh38
rs745540957 Chr.11: 65884411-65884411 on Build GRCh38
rs1000765021 Chr.11: 65884006-65884006 on Build GRCh38
rs749749422 Chr.11: 65884427-65884427 on Build GRCh38
rs1019491183 Chr.11: 65884072-65884072 on Build GRCh38
rs751786085 Chr.11: 65884024-65884024 on Build GRCh38
rs766917849 Chr.11: 65884228-65884228 on Build GRCh38
rs551274979 Chr.11: 65884393-65884393 on Build GRCh38
rs561112301 Chr.11: 65884368-65884368 on Build GRCh38
rs758013431 Chr.11: 65884197-65884197 on Build GRCh38
rs1027641192 Chr.11: 65884428-65884428 on Build GRCh38
rs899645192 Chr.11: 65884250-65884250 on Build GRCh38
rs746692808 Chr.11: 65883997-65883997 on Build GRCh38
rs370619260 Chr.11: 65884433-65884433 on Build GRCh38
rs369940484 Chr.11: 65884037-65884037 on Build GRCh38
rs113024632 Chr.11: 65884391-65884391 on Build GRCh38
rs2231892 Chr.11: 65884053-65884053 on Build GRCh38
rs938482343 Chr.11: 65884204-65884204 on Build GRCh38

View Full Product Details