Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32750034-32750582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YARS2
Entrez Gene ID
51067
Gene Name
tyrosyl-tRNA synthetase 2
Gene Aliases
CGI-04, MLASA2, MT-TYRRS, TYRRS
UniGene
Hs.505231
Gene Chromosome Location
Chr.12: 32725247-32755961 on Build GRCh38
Transcripts
NM_001040436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4431594 c.1156G>C p.D386H Chr.12: 32750055-32750055 on Build GRCh38
4950044 c.1147G>T p.V383F Chr.12: 32750064-32750064 on Build GRCh38
7203557 c.1122T>C p.Y374Y Chr.12: 32750089-32750089 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768932120 Chr.12: 32750264-32750264 on Build GRCh38
rs929177034 Chr.12: 32750314-32750314 on Build GRCh38
rs958565734 Chr.12: 32750211-32750211 on Build GRCh38
rs999333071 Chr.12: 32750369-32750369 on Build GRCh38
rs573053300 Chr.12: 32750159-32750159 on Build GRCh38
rs1008663977 Chr.12: 32750347-32750347 on Build GRCh38
rs539657695 Chr.12: 32750512-32750512 on Build GRCh38
rs577793752 Chr.12: 32750489-32750489 on Build GRCh38
rs756735477 Chr.12: 32750184-32750184 on Build GRCh38
rs138643327 Chr.12: 32750294-32750294 on Build GRCh38
rs10844339 Chr.12: 32750214-32750214 on Build GRCh38
rs541836600 Chr.12: 32750417-32750417 on Build GRCh38
rs970124000 Chr.12: 32750415-32750415 on Build GRCh38
rs370036689 Chr.12: 32750118-32750118 on Build GRCh38
rs983344668 Chr.12: 32750494-32750494 on Build GRCh38
rs959424436 Chr.12: 32750488-32750488 on Build GRCh38
rs753357287 Chr.12: 32750142-32750142 on Build GRCh38
rs368662425 Chr.12: 32750490-32750490 on Build GRCh38
rs577567134 Chr.12: 32750436-32750437 on Build GRCh38
rs930909357 Chr.12: 32750436-32750436 on Build GRCh38
rs983560337 Chr.12: 32750451-32750451 on Build GRCh38
rs770976759 Chr.12: 32750063-32750063 on Build GRCh38
rs78342381 Chr.12: 32750219-32750219 on Build GRCh38
rs956164426 Chr.12: 32750246-32750246 on Build GRCh38
rs1041163290 Chr.12: 32750172-32750172 on Build GRCh38
rs765397279 Chr.12: 32750201-32750201 on Build GRCh38
rs760307040 Chr.12: 32750113-32750113 on Build GRCh38
rs1047456448 Chr.12: 32750321-32750321 on Build GRCh38
rs528373649 Chr.12: 32750480-32750480 on Build GRCh38
rs996085180 Chr.12: 32750360-32750360 on Build GRCh38
rs1040402424 Chr.12: 32750350-32750350 on Build GRCh38
rs61926204 Chr.12: 32750491-32750491 on Build GRCh38
rs1022102710 Chr.12: 32750207-32750207 on Build GRCh38
rs769940270 Chr.12: 32750074-32750074 on Build GRCh38
rs766006803 Chr.12: 32750132-32750132 on Build GRCh38
rs1044505297 Chr.12: 32750094-32750094 on Build GRCh38
rs549681357 Chr.12: 32750411-32750411 on Build GRCh38
rs758438897 Chr.12: 32750390-32750390 on Build GRCh38
rs926390927 Chr.12: 32750291-32750291 on Build GRCh38
rs886086070 Chr.12: 32750133-32750133 on Build GRCh38
rs899979154 Chr.12: 32750189-32750189 on Build GRCh38
rs143432850 Chr.12: 32750318-32750318 on Build GRCh38
rs758457820 Chr.12: 32750247-32750247 on Build GRCh38
rs886158865 Chr.12: 32750337-32750337 on Build GRCh38
rs1018625610 Chr.12: 32750160-32750160 on Build GRCh38
rs774032188 Chr.12: 32750075-32750075 on Build GRCh38
rs1043427614 Chr.12: 32750496-32750496 on Build GRCh38
rs778866481 Chr.12: 32750199-32750199 on Build GRCh38
rs920523546 Chr.12: 32750306-32750306 on Build GRCh38
rs1047686241 Chr.12: 32750316-32750316 on Build GRCh38
rs999445982 Chr.12: 32750502-32750502 on Build GRCh38
rs994538891 Chr.12: 32750108-32750108 on Build GRCh38
rs748210942 Chr.12: 32750365-32750365 on Build GRCh38
rs773167596 Chr.12: 32750098-32750098 on Build GRCh38
rs554895317 Chr.12: 32750232-32750232 on Build GRCh38
rs530118789 Chr.12: 32750363-32750363 on Build GRCh38
rs779980398 Chr.12: 32750253-32750253 on Build GRCh38
rs976782350 Chr.12: 32750501-32750501 on Build GRCh38
rs202083773 Chr.12: 32750065-32750065 on Build GRCh38
rs542148044 Chr.12: 32750167-32750167 on Build GRCh38
rs776736900 Chr.12: 32750142-32750143 on Build GRCh38
rs187384054 Chr.12: 32750505-32750505 on Build GRCh38
rs896354895 Chr.12: 32750536-32750536 on Build GRCh38
rs568707849 Chr.12: 32750422-32750422 on Build GRCh38
rs761612441 Chr.12: 32750119-32750120 on Build GRCh38
rs899943596 Chr.12: 32750333-32750333 on Build GRCh38
rs369280067 Chr.12: 32750154-32750154 on Build GRCh38
rs560783820 Chr.12: 32750359-32750359 on Build GRCh38
rs903750717 Chr.12: 32750497-32750497 on Build GRCh38
rs997446330 Chr.12: 32750206-32750206 on Build GRCh38
rs572533168 Chr.12: 32750527-32750527 on Build GRCh38
rs556166099 Chr.12: 32750481-32750481 on Build GRCh38
rs374140841 Chr.12: 32750375-32750375 on Build GRCh38
rs767418254 Chr.12: 32750080-32750080 on Build GRCh38
rs148832666 Chr.12: 32750304-32750304 on Build GRCh38
rs1047508966 Chr.12: 32750126-32750126 on Build GRCh38
rs1032580820 Chr.12: 32750210-32750211 on Build GRCh38
rs746124254 Chr.12: 32750067-32750067 on Build GRCh38
rs190866316 Chr.12: 32750212-32750212 on Build GRCh38
rs543876265 Chr.12: 32750270-32750270 on Build GRCh38
rs1041133995 Chr.12: 32750329-32750329 on Build GRCh38
rs773382919 Chr.12: 32750401-32750401 on Build GRCh38
rs893935291 Chr.12: 32750533-32750533 on Build GRCh38
rs768896199 Chr.12: 32750120-32750121 on Build GRCh38
rs774698279 Chr.12: 32750540-32750540 on Build GRCh38
rs931666827 Chr.12: 32750493-32750493 on Build GRCh38
rs371406996 Chr.12: 32750263-32750263 on Build GRCh38
rs138032809 Chr.12: 32750397-32750397 on Build GRCh38

View Full Product Details