Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150676883-150677471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOZ3
Entrez Gene ID
91977
Gene Name
myozenin 3
Gene Aliases
CS-3, CS3, FRP3
UniGene
Hs.91626
Gene Chromosome Location
Chr.5: 150660841-150679368 on Build GRCh38
Transcripts
NM_001122853.2 NM_133371.4

SNP Detail

SNP ID Chromosome Location
rs897889313 Chr.5: 150677023-150677023 on Build GRCh38
rs1003710549 Chr.5: 150677120-150677120 on Build GRCh38
rs766328216 Chr.5: 150677199-150677199 on Build GRCh38
rs73276196 Chr.5: 150677061-150677061 on Build GRCh38
rs999434710 Chr.5: 150677034-150677034 on Build GRCh38
rs558224474 Chr.5: 150677197-150677197 on Build GRCh38
rs76816714 Chr.5: 150677435-150677435 on Build GRCh38
rs747515819 Chr.5: 150676924-150676924 on Build GRCh38
rs997763947 Chr.5: 150677439-150677439 on Build GRCh38
rs935811298 Chr.5: 150677289-150677289 on Build GRCh38
rs866407495 Chr.5: 150677313-150677313 on Build GRCh38
rs866594968 Chr.5: 150677260-150677260 on Build GRCh38
rs538511702 Chr.5: 150676930-150676930 on Build GRCh38
rs975465606 Chr.5: 150677167-150677167 on Build GRCh38
rs777293250 Chr.5: 150677401-150677401 on Build GRCh38
rs75800382 Chr.5: 150677140-150677140 on Build GRCh38
rs1051572178 Chr.5: 150676956-150676956 on Build GRCh38
rs79703932 Chr.5: 150677044-150677044 on Build GRCh38
rs1018241788 Chr.5: 150677148-150677148 on Build GRCh38
rs1044775148 Chr.5: 150677335-150677335 on Build GRCh38
rs577466492 Chr.5: 150677233-150677233 on Build GRCh38
rs952212555 Chr.5: 150677113-150677113 on Build GRCh38
rs573539965 Chr.5: 150677038-150677038 on Build GRCh38
rs111415534 Chr.5: 150677284-150677284 on Build GRCh38
rs375816009 Chr.5: 150677103-150677103 on Build GRCh38
rs377663921 Chr.5: 150676920-150676920 on Build GRCh38
rs113580160 Chr.5: 150677380-150677380 on Build GRCh38
rs76245815 Chr.5: 150677072-150677072 on Build GRCh38
rs950016117 Chr.5: 150677350-150677350 on Build GRCh38
rs112408743 Chr.5: 150676906-150676906 on Build GRCh38
rs112084120 Chr.5: 150677398-150677398 on Build GRCh38
rs769332536 Chr.5: 150676925-150676925 on Build GRCh38
rs902439119 Chr.5: 150677015-150677015 on Build GRCh38
rs111890776 Chr.5: 150677040-150677040 on Build GRCh38
rs763898351 Chr.5: 150677304-150677304 on Build GRCh38
rs112197156 Chr.5: 150677414-150677414 on Build GRCh38
rs189200570 Chr.5: 150676974-150676974 on Build GRCh38
rs745310342 Chr.5: 150676910-150676910 on Build GRCh38
rs530298640 Chr.5: 150677311-150677311 on Build GRCh38
rs1024312547 Chr.5: 150677020-150677020 on Build GRCh38
rs750168793 Chr.5: 150677161-150677161 on Build GRCh38
rs55723763 Chr.5: 150677434-150677434 on Build GRCh38
rs909858779 Chr.5: 150677160-150677160 on Build GRCh38
rs556852241 Chr.5: 150677210-150677210 on Build GRCh38
rs544765010 Chr.5: 150677154-150677154 on Build GRCh38
rs146728320 Chr.5: 150677388-150677388 on Build GRCh38
rs563111321 Chr.5: 150677294-150677294 on Build GRCh38
rs897812626 Chr.5: 150677374-150677374 on Build GRCh38
rs760454800 Chr.5: 150677169-150677169 on Build GRCh38
rs1025777425 Chr.5: 150677371-150677371 on Build GRCh38
rs977147803 Chr.5: 150677261-150677261 on Build GRCh38
rs571469572 Chr.5: 150676928-150676928 on Build GRCh38
rs572892350 Chr.5: 150677263-150677263 on Build GRCh38
rs527744496 Chr.5: 150677354-150677354 on Build GRCh38
rs757943119 Chr.5: 150676911-150676911 on Build GRCh38
rs113787782 Chr.5: 150677047-150677047 on Build GRCh38
rs935211543 Chr.5: 150677264-150677264 on Build GRCh38
rs542417575 Chr.5: 150677322-150677322 on Build GRCh38
rs993499820 Chr.5: 150677379-150677379 on Build GRCh38
rs778605607 Chr.5: 150676907-150676907 on Build GRCh38
rs1025850949 Chr.5: 150677121-150677121 on Build GRCh38
rs777120044 Chr.5: 150676926-150676926 on Build GRCh38
rs906273826 Chr.5: 150677342-150677342 on Build GRCh38
rs777068489 Chr.5: 150676955-150676955 on Build GRCh38
rs192102282 Chr.5: 150677330-150677330 on Build GRCh38

View Full Product Details