Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150677449-150677793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOZ3
Entrez Gene ID
91977
Gene Name
myozenin 3
Gene Aliases
CS-3, CS3, FRP3
UniGene
Hs.91626
Gene Chromosome Location
Chr.5: 150660841-150679368 on Build GRCh38
Transcripts
NM_001122853.2 NM_133371.4

SNP Detail

SNP ID Chromosome Location
rs768920629 Chr.5: 150677714-150677714 on Build GRCh38
rs80342032 Chr.5: 150677678-150677678 on Build GRCh38
rs925657907 Chr.5: 150677551-150677551 on Build GRCh38
rs955424570 Chr.5: 150677512-150677512 on Build GRCh38
rs977118097 Chr.5: 150677544-150677544 on Build GRCh38
rs1045092013 Chr.5: 150677639-150677639 on Build GRCh38
rs906200224 Chr.5: 150677662-150677662 on Build GRCh38
rs143023536 Chr.5: 150677568-150677569 on Build GRCh38
rs770314761 Chr.5: 150677558-150677558 on Build GRCh38
rs1045648555 Chr.5: 150677642-150677642 on Build GRCh38
rs117407806 Chr.5: 150677583-150677583 on Build GRCh38
rs1029700696 Chr.5: 150677510-150677510 on Build GRCh38
rs967031182 Chr.5: 150677481-150677481 on Build GRCh38
rs929275503 Chr.5: 150677688-150677688 on Build GRCh38
rs1017144250 Chr.5: 150677739-150677739 on Build GRCh38
rs78813230 Chr.5: 150677557-150677557 on Build GRCh38
rs1007658997 Chr.5: 150677708-150677708 on Build GRCh38
rs746414508 Chr.5: 150677498-150677498 on Build GRCh38
rs998129867 Chr.5: 150677763-150677763 on Build GRCh38
rs781648089 Chr.5: 150677495-150677495 on Build GRCh38
rs59423491 Chr.5: 150677748-150677748 on Build GRCh38
rs576748956 Chr.5: 150677707-150677707 on Build GRCh38
rs544951756 Chr.5: 150677735-150677735 on Build GRCh38
rs1021208037 Chr.5: 150677765-150677765 on Build GRCh38
rs575151861 Chr.5: 150677752-150677752 on Build GRCh38
rs561371286 Chr.5: 150677689-150677689 on Build GRCh38
rs548204031 Chr.5: 150677574-150677574 on Build GRCh38
rs184315576 Chr.5: 150677612-150677612 on Build GRCh38

View Full Product Details