Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42548591-42549087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS2
Entrez Gene ID
55142
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
C15orf25, CEP27, HsT17025
UniGene
Hs.14347
Gene Chromosome Location
Chr.15: 42548813-42569994 on Build GRCh38
Transcripts
NM_001130447.1 NM_001323629.1 NM_001323630.1 NM_001323631.1 NM_001323632.1 NM_018097.2 XM_017022396.1 XM_017022397.1 NM_153260.2

Gene Symbol
LRRC57
Entrez Gene ID
255252
Gene Name
leucine rich repeat containing 57
Gene Aliases
-
UniGene
Hs.234681
Gene Chromosome Location
Chr.15: 42528332-42548804 on Build GRCh38
Transcripts
NM_001130447.1 NM_001323629.1 NM_001323630.1 NM_001323631.1 NM_001323632.1 NM_018097.2 XM_017022396.1 XM_017022397.1 NM_153260.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4472699 c.17C>T p.P6L Chr.15: 42548889-42548889 on Build GRCh38
6411809 c.46G>A p.A16T Chr.15: 42548918-42548918 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199814168 Chr.15: 42548902-42548902 on Build GRCh38
rs898168506 Chr.15: 42548866-42548866 on Build GRCh38
rs757773034 Chr.15: 42548815-42548815 on Build GRCh38
rs895195197 Chr.15: 42548898-42548898 on Build GRCh38
rs770467092 Chr.15: 42548659-42548659 on Build GRCh38
rs889578283 Chr.15: 42548736-42548736 on Build GRCh38
rs534311844 Chr.15: 42548771-42548771 on Build GRCh38
rs911071708 Chr.15: 42549019-42549019 on Build GRCh38
rs369304796 Chr.15: 42548705-42548705 on Build GRCh38
rs898208032 Chr.15: 42548720-42548720 on Build GRCh38
rs1021209557 Chr.15: 42548809-42548809 on Build GRCh38
rs1032477002 Chr.15: 42548822-42548823 on Build GRCh38
rs1038064265 Chr.15: 42548742-42548742 on Build GRCh38
rs938068793 Chr.15: 42548749-42548749 on Build GRCh38
rs527664189 Chr.15: 42548669-42548669 on Build GRCh38
rs758573008 Chr.15: 42548861-42548861 on Build GRCh38
rs560295056 Chr.15: 42548872-42548872 on Build GRCh38
rs995396546 Chr.15: 42548633-42548633 on Build GRCh38
rs769888858 Chr.15: 42548884-42548884 on Build GRCh38
rs914044418 Chr.15: 42548670-42548670 on Build GRCh38
rs1049880041 Chr.15: 42548789-42548789 on Build GRCh38
rs545252366 Chr.15: 42548694-42548694 on Build GRCh38
rs756870870 Chr.15: 42548832-42548832 on Build GRCh38
rs761125331 Chr.15: 42548757-42548757 on Build GRCh38
rs1016634403 Chr.15: 42548717-42548717 on Build GRCh38
rs769559137 Chr.15: 42549042-42549042 on Build GRCh38
rs959732239 Chr.15: 42548846-42548846 on Build GRCh38
rs377359457 Chr.15: 42548987-42548987 on Build GRCh38
rs201932565 Chr.15: 42548999-42548999 on Build GRCh38
rs761336686 Chr.15: 42548976-42548976 on Build GRCh38
rs552006403 Chr.15: 42548772-42548772 on Build GRCh38
rs118132322 Chr.15: 42548731-42548731 on Build GRCh38
rs995112752 Chr.15: 42548752-42548752 on Build GRCh38
rs767298901 Chr.15: 42548725-42548725 on Build GRCh38
rs758703429 Chr.15: 42548998-42548998 on Build GRCh38
rs959514135 Chr.15: 42548764-42548764 on Build GRCh38
rs991235111 Chr.15: 42548760-42548760 on Build GRCh38
rs898258966 Chr.15: 42548780-42548781 on Build GRCh38
rs970748275 Chr.15: 42548858-42548858 on Build GRCh38
rs925355505 Chr.15: 42548747-42548747 on Build GRCh38
rs563140709 Chr.15: 42548698-42548698 on Build GRCh38
rs1008923465 Chr.15: 42548799-42548799 on Build GRCh38
rs890820743 Chr.15: 42548924-42548924 on Build GRCh38
rs566081936 Chr.15: 42548765-42548765 on Build GRCh38
rs191926586 Chr.15: 42548951-42548951 on Build GRCh38
rs986664773 Chr.15: 42548821-42548821 on Build GRCh38
rs942483833 Chr.15: 42548841-42548841 on Build GRCh38
rs116509460 Chr.15: 42548776-42548776 on Build GRCh38
rs543883377 Chr.15: 42549056-42549056 on Build GRCh38
rs987047440 Chr.15: 42548737-42548737 on Build GRCh38
rs369506094 Chr.15: 42549011-42549011 on Build GRCh38
rs950025862 Chr.15: 42548763-42548763 on Build GRCh38
rs929638710 Chr.15: 42549054-42549054 on Build GRCh38
rs553300137 Chr.15: 42548896-42548896 on Build GRCh38
rs369929649 Chr.15: 42548983-42548983 on Build GRCh38
rs780431061 Chr.15: 42548867-42548867 on Build GRCh38
rs886614004 Chr.15: 42548759-42548759 on Build GRCh38
rs201685144 Chr.15: 42549028-42549028 on Build GRCh38
rs748424848 Chr.15: 42548879-42548879 on Build GRCh38
rs537169023 Chr.15: 42548863-42548863 on Build GRCh38
rs573761871 Chr.15: 42548878-42548878 on Build GRCh38
rs1003708139 Chr.15: 42548761-42548761 on Build GRCh38
rs899389585 Chr.15: 42548750-42548750 on Build GRCh38
rs529496867 Chr.15: 42548738-42548738 on Build GRCh38
rs1025056856 Chr.15: 42549059-42549059 on Build GRCh38
rs747462059 Chr.15: 42548868-42548868 on Build GRCh38
rs753322849 Chr.15: 42548970-42548970 on Build GRCh38
rs1028992272 Chr.15: 42548619-42548619 on Build GRCh38
rs996920939 Chr.15: 42548724-42548724 on Build GRCh38
rs1035061633 Chr.15: 42548762-42548762 on Build GRCh38
rs925302851 Chr.15: 42548678-42548678 on Build GRCh38
rs773437782 Chr.15: 42548889-42548889 on Build GRCh38
rs549630239 Chr.15: 42548745-42548745 on Build GRCh38
rs1045985941 Chr.15: 42548700-42548700 on Build GRCh38
rs942411152 Chr.15: 42549030-42549030 on Build GRCh38
rs886916628 Chr.15: 42548712-42548712 on Build GRCh38
rs973852375 Chr.15: 42549032-42549032 on Build GRCh38
rs910945018 Chr.15: 42548831-42548831 on Build GRCh38
rs771692836 Chr.15: 42548735-42548735 on Build GRCh38
rs542482693 Chr.15: 42548890-42548890 on Build GRCh38
rs1026459320 Chr.15: 42548638-42548638 on Build GRCh38
rs774032416 Chr.15: 42548677-42548677 on Build GRCh38
rs923780414 Chr.15: 42548726-42548726 on Build GRCh38
rs188040203 Chr.15: 42548697-42548697 on Build GRCh38
rs541395607 Chr.15: 42548921-42548921 on Build GRCh38
rs780091262 Chr.15: 42548624-42548624 on Build GRCh38
rs923729877 Chr.15: 42548791-42548791 on Build GRCh38
rs750520496 Chr.15: 42548859-42548859 on Build GRCh38
rs749214987 Chr.15: 42549062-42549062 on Build GRCh38
rs760136124 Chr.15: 42548906-42548906 on Build GRCh38
rs200759756 Chr.15: 42548847-42548847 on Build GRCh38
rs899751442 Chr.15: 42548783-42548783 on Build GRCh38
rs946677175 Chr.15: 42548708-42548708 on Build GRCh38
rs16973349 Chr.15: 42548671-42548671 on Build GRCh38
rs987384095 Chr.15: 42548665-42548665 on Build GRCh38
rs986608857 Chr.15: 42548996-42548996 on Build GRCh38
rs780289055 Chr.15: 42548961-42548961 on Build GRCh38
rs996581696 Chr.15: 42548788-42548788 on Build GRCh38
rs1055754076 Chr.15: 42548695-42548695 on Build GRCh38
rs373315304 Chr.15: 42548985-42548985 on Build GRCh38
rs1022680916 Chr.15: 42548909-42548909 on Build GRCh38
rs559715353 Chr.15: 42548667-42548667 on Build GRCh38
rs978126683 Chr.15: 42548922-42548922 on Build GRCh38
rs927932198 Chr.15: 42548676-42548676 on Build GRCh38
rs968338983 Chr.15: 42548775-42548775 on Build GRCh38
rs763737735 Chr.15: 42548995-42548995 on Build GRCh38

View Full Product Details