Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42559114-42559586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS2
Entrez Gene ID
55142
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
C15orf25, CEP27, HsT17025
UniGene
Hs.14347
Gene Chromosome Location
Chr.15: 42548813-42569994 on Build GRCh38
Transcripts
NM_001130447.1 NM_001323629.1 NM_001323630.1 NM_001323631.1 NM_001323632.1 NM_018097.2 XM_017022396.1 XM_017022397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4150939 c.256+2T>A p.? Chr.15: 42559410-42559410 on Build GRCh38
6848980 c.217G>A p.E73K Chr.15: 42559369-42559369 on Build GRCh38
3501313 c.244C>T p.P82S Chr.15: 42559396-42559396 on Build GRCh38
6229366 c.238G>A p.V80I Chr.15: 42559390-42559390 on Build GRCh38
961754 c.236T>C p.V79A Chr.15: 42559388-42559388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760899448 Chr.15: 42559334-42559334 on Build GRCh38
rs777773858 Chr.15: 42559395-42559395 on Build GRCh38
rs371408509 Chr.15: 42559301-42559301 on Build GRCh38
rs914446414 Chr.15: 42559267-42559267 on Build GRCh38
rs750711760 Chr.15: 42559389-42559389 on Build GRCh38
rs192465821 Chr.15: 42559400-42559400 on Build GRCh38
rs771873522 Chr.15: 42559406-42559406 on Build GRCh38
rs774202835 Chr.15: 42559311-42559311 on Build GRCh38
rs1005907127 Chr.15: 42559182-42559182 on Build GRCh38
rs34678957 Chr.15: 42559378-42559378 on Build GRCh38
rs968969893 Chr.15: 42559387-42559387 on Build GRCh38
rs918762231 Chr.15: 42559205-42559205 on Build GRCh38
rs929999204 Chr.15: 42559226-42559226 on Build GRCh38
rs539145350 Chr.15: 42559324-42559324 on Build GRCh38
rs1034411593 Chr.15: 42559546-42559546 on Build GRCh38
rs576872448 Chr.15: 42559178-42559178 on Build GRCh38
rs761896441 Chr.15: 42559353-42559353 on Build GRCh38
rs758589168 Chr.15: 42559372-42559372 on Build GRCh38
rs755827503 Chr.15: 42559393-42559393 on Build GRCh38
rs1024404301 Chr.15: 42559462-42559462 on Build GRCh38
rs921806295 Chr.15: 42559550-42559550 on Build GRCh38
rs572189853 Chr.15: 42559544-42559544 on Build GRCh38
rs765341901 Chr.15: 42559434-42559434 on Build GRCh38
rs755606306 Chr.15: 42559526-42559526 on Build GRCh38
rs907814217 Chr.15: 42559252-42559252 on Build GRCh38
rs773254131 Chr.15: 42559437-42559437 on Build GRCh38
rs73406531 Chr.15: 42559505-42559505 on Build GRCh38
rs202188115 Chr.15: 42559313-42559313 on Build GRCh38
rs761753224 Chr.15: 42559281-42559281 on Build GRCh38
rs62019312 Chr.15: 42559199-42559199 on Build GRCh38
rs780902927 Chr.15: 42559376-42559376 on Build GRCh38
rs749222409 Chr.15: 42559397-42559397 on Build GRCh38
rs1019888960 Chr.15: 42559386-42559388 on Build GRCh38
rs545649165 Chr.15: 42559181-42559181 on Build GRCh38
rs762975508 Chr.15: 42559448-42559448 on Build GRCh38
rs139117761 Chr.15: 42559174-42559174 on Build GRCh38
rs149060624 Chr.15: 42559352-42559352 on Build GRCh38
rs766659458 Chr.15: 42559451-42559451 on Build GRCh38
rs906340162 Chr.15: 42559153-42559153 on Build GRCh38
rs981518626 Chr.15: 42559214-42559214 on Build GRCh38
rs563375466 Chr.15: 42559204-42559204 on Build GRCh38
rs762305646 Chr.15: 42559430-42559430 on Build GRCh38
rs982807052 Chr.15: 42559500-42559500 on Build GRCh38
rs971545219 Chr.15: 42559513-42559513 on Build GRCh38
rs775897433 Chr.15: 42559326-42559326 on Build GRCh38
rs973315325 Chr.15: 42559542-42559542 on Build GRCh38
rs372326307 Chr.15: 42559354-42559354 on Build GRCh38
rs367982973 Chr.15: 42559291-42559291 on Build GRCh38
rs35404961 Chr.15: 42559402-42559402 on Build GRCh38
rs1017578787 Chr.15: 42559196-42559196 on Build GRCh38
rs1015937746 Chr.15: 42559458-42559458 on Build GRCh38
rs777253784 Chr.15: 42559426-42559426 on Build GRCh38
rs530739988 Chr.15: 42559224-42559224 on Build GRCh38
rs369734718 Chr.15: 42559357-42559357 on Build GRCh38
rs1009875984 Chr.15: 42559363-42559363 on Build GRCh38
rs376613979 Chr.15: 42559355-42559355 on Build GRCh38
rs533505620 Chr.15: 42559533-42559533 on Build GRCh38
rs764691187 Chr.15: 42559339-42559339 on Build GRCh38
rs111483510 Chr.15: 42559279-42559279 on Build GRCh38
rs777526138 Chr.15: 42559290-42559290 on Build GRCh38
rs751669093 Chr.15: 42559455-42559455 on Build GRCh38
rs994429175 Chr.15: 42559490-42559490 on Build GRCh38
rs992922028 Chr.15: 42559208-42559208 on Build GRCh38
rs370748242 Chr.15: 42559280-42559280 on Build GRCh38
rs780328799 Chr.15: 42559501-42559501 on Build GRCh38
rs752563550 Chr.15: 42559384-42559384 on Build GRCh38
rs971597377 Chr.15: 42559206-42559206 on Build GRCh38
rs997218038 Chr.15: 42559175-42559175 on Build GRCh38
rs1041546427 Chr.15: 42559170-42559170 on Build GRCh38
rs368140825 Chr.15: 42559418-42559418 on Build GRCh38
rs774965243 Chr.15: 42559407-42559407 on Build GRCh38
rs1051525561 Chr.15: 42559310-42559310 on Build GRCh38
rs11070374 Chr.15: 42559266-42559266 on Build GRCh38
rs937321675 Chr.15: 42559231-42559231 on Build GRCh38
rs575219116 Chr.15: 42559515-42559515 on Build GRCh38
rs761103788 Chr.15: 42559415-42559415 on Build GRCh38
rs371863700 Chr.15: 42559466-42559466 on Build GRCh38

View Full Product Details