Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40922622-40923116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT23
Entrez Gene ID
25984
Gene Name
keratin 23
Gene Aliases
CK23, HAIK1, K23
UniGene
Hs.9029
Gene Chromosome Location
Chr.17: 40922696-40937643 on Build GRCh38
Transcripts
NM_001282433.1 NM_015515.4 XM_005257200.4 XM_011524595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
978985 c.1263C>T p.H421H Chr.17: 40922995-40922995 on Build GRCh38
5436383 c.1212G>A p.E404E Chr.17: 40923046-40923046 on Build GRCh38
978986 c.1243G>A p.V415M Chr.17: 40923015-40923015 on Build GRCh38
5462638 c.1175-2A>C p.? Chr.17: 40923085-40923085 on Build GRCh38
4845896 c.1268G>A p.*423* Chr.17: 40922990-40922990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995871680 Chr.17: 40922732-40922732 on Build GRCh38
rs547680301 Chr.17: 40922869-40922869 on Build GRCh38
rs755906599 Chr.17: 40922708-40922708 on Build GRCh38
rs910435737 Chr.17: 40922774-40922774 on Build GRCh38
rs747316547 Chr.17: 40922983-40922983 on Build GRCh38
rs928046307 Chr.17: 40922884-40922884 on Build GRCh38
rs749233409 Chr.17: 40922852-40922852 on Build GRCh38
rs755118437 Chr.17: 40923043-40923043 on Build GRCh38
rs1013719572 Chr.17: 40922851-40922851 on Build GRCh38
rs762754329 Chr.17: 40923082-40923082 on Build GRCh38
rs781675485 Chr.17: 40923083-40923083 on Build GRCh38
rs923124501 Chr.17: 40922725-40922725 on Build GRCh38
rs565177412 Chr.17: 40922808-40922808 on Build GRCh38
rs187778944 Chr.17: 40923037-40923037 on Build GRCh38
rs888688667 Chr.17: 40923015-40923015 on Build GRCh38
rs985942928 Chr.17: 40923040-40923040 on Build GRCh38
rs941697818 Chr.17: 40922791-40922791 on Build GRCh38
rs969705679 Chr.17: 40922915-40922915 on Build GRCh38
rs762669648 Chr.17: 40922946-40922946 on Build GRCh38
rs779907993 Chr.17: 40922717-40922717 on Build GRCh38
rs183044352 Chr.17: 40922955-40922955 on Build GRCh38
rs777084466 Chr.17: 40922991-40922991 on Build GRCh38
rs796246628 Chr.17: 40922858-40922859 on Build GRCh38
rs893751129 Chr.17: 40922669-40922669 on Build GRCh38
rs1050785353 Chr.17: 40922746-40922746 on Build GRCh38
rs368165611 Chr.17: 40922980-40922980 on Build GRCh38
rs759069821 Chr.17: 40922993-40922993 on Build GRCh38
rs750334193 Chr.17: 40923042-40923042 on Build GRCh38
rs775010306 Chr.17: 40922995-40922995 on Build GRCh38
rs748222056 Chr.17: 40923044-40923044 on Build GRCh38
rs17856805 Chr.17: 40923080-40923080 on Build GRCh38
rs759121679 Chr.17: 40922937-40922937 on Build GRCh38
rs751418799 Chr.17: 40923036-40923036 on Build GRCh38
rs768540862 Chr.17: 40922899-40922899 on Build GRCh38
rs570803610 Chr.17: 40922972-40922972 on Build GRCh38
rs563833439 Chr.17: 40922675-40922675 on Build GRCh38
rs1027364089 Chr.17: 40923002-40923002 on Build GRCh38
rs754009419 Chr.17: 40922964-40922964 on Build GRCh38
rs1037413125 Chr.17: 40922793-40922793 on Build GRCh38
rs772183466 Chr.17: 40922977-40922977 on Build GRCh38
rs900155461 Chr.17: 40922728-40922728 on Build GRCh38
rs988210534 Chr.17: 40922720-40922720 on Build GRCh38
rs763881830 Chr.17: 40922949-40922949 on Build GRCh38
rs992837734 Chr.17: 40922680-40922680 on Build GRCh38
rs751306985 Chr.17: 40922950-40922950 on Build GRCh38
rs772368127 Chr.17: 40923064-40923064 on Build GRCh38
rs2269861 Chr.17: 40922687-40922687 on Build GRCh38
rs532403666 Chr.17: 40922834-40922834 on Build GRCh38
rs781250693 Chr.17: 40922956-40922956 on Build GRCh38
rs773604297 Chr.17: 40923073-40923073 on Build GRCh38
rs747434633 Chr.17: 40923077-40923077 on Build GRCh38
rs746308413 Chr.17: 40923095-40923095 on Build GRCh38
rs755065519 Chr.17: 40922966-40922966 on Build GRCh38
rs979418242 Chr.17: 40922706-40922706 on Build GRCh38
rs1056782570 Chr.17: 40922814-40922814 on Build GRCh38
rs773242551 Chr.17: 40923073-40923076 on Build GRCh38
rs956672332 Chr.17: 40922970-40922970 on Build GRCh38
rs767591200 Chr.17: 40923039-40923039 on Build GRCh38
rs897490664 Chr.17: 40922794-40922794 on Build GRCh38
rs779296191 Chr.17: 40922971-40922971 on Build GRCh38
rs1015805378 Chr.17: 40922847-40922847 on Build GRCh38
rs565503399 Chr.17: 40922901-40922901 on Build GRCh38
rs530079611 Chr.17: 40922709-40922709 on Build GRCh38
rs868357650 Chr.17: 40922733-40922733 on Build GRCh38
rs561032594 Chr.17: 40922929-40922929 on Build GRCh38
rs371291648 Chr.17: 40923056-40923056 on Build GRCh38
rs956809188 Chr.17: 40922716-40922716 on Build GRCh38
rs763639171 Chr.17: 40923019-40923019 on Build GRCh38
rs750178302 Chr.17: 40922707-40922707 on Build GRCh38
rs915139411 Chr.17: 40922924-40922924 on Build GRCh38
rs937995878 Chr.17: 40922896-40922896 on Build GRCh38
rs544459987 Chr.17: 40922693-40922693 on Build GRCh38
rs368903518 Chr.17: 40922936-40922936 on Build GRCh38
rs769236996 Chr.17: 40922994-40922994 on Build GRCh38
rs982169588 Chr.17: 40922880-40922880 on Build GRCh38

View Full Product Details