Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41528078-41528618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT19
Entrez Gene ID
3880
Gene Name
keratin 19
Gene Aliases
CK19, K19, K1CS
UniGene
Hs.654568
Gene Chromosome Location
Chr.17: 41523617-41528389 on Build GRCh38
Transcripts
NM_002276.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5791711 c.76G>C p.G26R Chr.17: 41528172-41528172 on Build GRCh38
7150290 c.25T>C p.S9P Chr.17: 41528223-41528223 on Build GRCh38
4935884 c.8C>T p.S3F Chr.17: 41528240-41528240 on Build GRCh38
5771598 c.143C>T p.S48F Chr.17: 41528105-41528105 on Build GRCh38
7401527 c.165G>C p.S55S Chr.17: 41528083-41528083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556784801 Chr.17: 41528358-41528358 on Build GRCh38
rs1030064251 Chr.17: 41528525-41528525 on Build GRCh38
rs780473902 Chr.17: 41528155-41528155 on Build GRCh38
rs938768795 Chr.17: 41528355-41528355 on Build GRCh38
rs897074296 Chr.17: 41528266-41528266 on Build GRCh38
rs1049085222 Chr.17: 41528147-41528147 on Build GRCh38
rs770701407 Chr.17: 41528237-41528237 on Build GRCh38
rs779344248 Chr.17: 41528221-41528221 on Build GRCh38
rs11550878 Chr.17: 41528102-41528102 on Build GRCh38
rs780570359 Chr.17: 41528098-41528098 on Build GRCh38
rs760470135 Chr.17: 41528121-41528121 on Build GRCh38
rs757898492 Chr.17: 41528228-41528228 on Build GRCh38
rs11550883 Chr.17: 41528158-41528158 on Build GRCh38
rs773372080 Chr.17: 41528277-41528277 on Build GRCh38
rs761848259 Chr.17: 41528268-41528268 on Build GRCh38
rs766759618 Chr.17: 41528203-41528203 on Build GRCh38
rs748836644 Chr.17: 41528100-41528100 on Build GRCh38
rs1036617461 Chr.17: 41528342-41528342 on Build GRCh38
rs776604319 Chr.17: 41528132-41528132 on Build GRCh38
rs985777713 Chr.17: 41528537-41528538 on Build GRCh38
rs372117019 Chr.17: 41528292-41528292 on Build GRCh38
rs951994860 Chr.17: 41528276-41528276 on Build GRCh38
rs760953846 Chr.17: 41528278-41528278 on Build GRCh38
rs11550875 Chr.17: 41528170-41528170 on Build GRCh38
rs999455072 Chr.17: 41528206-41528206 on Build GRCh38
rs867929608 Chr.17: 41528536-41528536 on Build GRCh38
rs775532695 Chr.17: 41528267-41528267 on Build GRCh38
rs951942665 Chr.17: 41528495-41528495 on Build GRCh38
rs568942571 Chr.17: 41528225-41528225 on Build GRCh38
rs745607114 Chr.17: 41528263-41528263 on Build GRCh38
rs777002729 Chr.17: 41528283-41528283 on Build GRCh38
rs914762049 Chr.17: 41528595-41528595 on Build GRCh38
rs928778269 Chr.17: 41528314-41528314 on Build GRCh38
rs550878833 Chr.17: 41528191-41528191 on Build GRCh38
rs746663570 Chr.17: 41528234-41528234 on Build GRCh38
rs56051972 Chr.17: 41528346-41528346 on Build GRCh38
rs778401116 Chr.17: 41528163-41528163 on Build GRCh38
rs373982346 Chr.17: 41528254-41528254 on Build GRCh38
rs578205032 Chr.17: 41528405-41528405 on Build GRCh38
rs1057255725 Chr.17: 41528376-41528376 on Build GRCh38
rs769701409 Chr.17: 41528163-41528163 on Build GRCh38
rs981502785 Chr.17: 41528522-41528522 on Build GRCh38
rs907844710 Chr.17: 41528499-41528499 on Build GRCh38
rs902189095 Chr.17: 41528380-41528380 on Build GRCh38
rs766262038 Chr.17: 41528128-41528128 on Build GRCh38
rs1012953091 Chr.17: 41528396-41528396 on Build GRCh38
rs561307487 Chr.17: 41528177-41528177 on Build GRCh38
rs370965852 Chr.17: 41528136-41528136 on Build GRCh38
rs897422 Chr.17: 41528587-41528587 on Build GRCh38
rs1019108252 Chr.17: 41528397-41528397 on Build GRCh38
rs893425984 Chr.17: 41528202-41528202 on Build GRCh38
rs996400916 Chr.17: 41528437-41528437 on Build GRCh38
rs11550879 Chr.17: 41528160-41528160 on Build GRCh38
rs761002028 Chr.17: 41528200-41528200 on Build GRCh38
rs756469704 Chr.17: 41528151-41528151 on Build GRCh38
rs1011858026 Chr.17: 41528356-41528356 on Build GRCh38
rs566512747 Chr.17: 41528293-41528293 on Build GRCh38
rs772897568 Chr.17: 41528115-41528115 on Build GRCh38
rs1013172050 Chr.17: 41528169-41528169 on Build GRCh38
rs774046851 Chr.17: 41528110-41528110 on Build GRCh38
rs200777034 Chr.17: 41528199-41528199 on Build GRCh38
rs1055999637 Chr.17: 41528336-41528336 on Build GRCh38
rs767726398 Chr.17: 41528143-41528143 on Build GRCh38
rs747600254 Chr.17: 41528164-41528164 on Build GRCh38
rs768130773 Chr.17: 41528105-41528105 on Build GRCh38
rs754136976 Chr.17: 41528213-41528213 on Build GRCh38
rs533114218 Chr.17: 41528264-41528264 on Build GRCh38
rs766455957 Chr.17: 41528279-41528279 on Build GRCh38
rs200465465 Chr.17: 41528150-41528150 on Build GRCh38
rs1032257608 Chr.17: 41528494-41528495 on Build GRCh38
rs761940713 Chr.17: 41528188-41528188 on Build GRCh38
rs751620624 Chr.17: 41528137-41528137 on Build GRCh38
rs772177891 Chr.17: 41528274-41528274 on Build GRCh38
rs548558411 Chr.17: 41528309-41528309 on Build GRCh38
rs373835703 Chr.17: 41528229-41528229 on Build GRCh38
rs545290661 Chr.17: 41528145-41528145 on Build GRCh38
rs774591684 Chr.17: 41528187-41528187 on Build GRCh38
rs367995923 Chr.17: 41528289-41528289 on Build GRCh38
rs909102891 Chr.17: 41528538-41528538 on Build GRCh38
rs769793854 Chr.17: 41528186-41528186 on Build GRCh38
rs965857888 Chr.17: 41528431-41528431 on Build GRCh38
rs377642975 Chr.17: 41528112-41528112 on Build GRCh38
rs1010567437 Chr.17: 41528210-41528210 on Build GRCh38
rs754912899 Chr.17: 41528496-41528496 on Build GRCh38
rs770740803 Chr.17: 41528173-41528173 on Build GRCh38
rs566968286 Chr.17: 41528330-41528330 on Build GRCh38
rs781145770 Chr.17: 41528249-41528249 on Build GRCh38
rs997665049 Chr.17: 41528504-41528506 on Build GRCh38
rs1043388210 Chr.17: 41528366-41528366 on Build GRCh38
rs897921295 Chr.17: 41528120-41528120 on Build GRCh38
rs533648489 Chr.17: 41528300-41528300 on Build GRCh38
rs1028852086 Chr.17: 41528439-41528439 on Build GRCh38
rs937501680 Chr.17: 41528328-41528328 on Build GRCh38
rs975074674 Chr.17: 41528101-41528101 on Build GRCh38
rs543003088 Chr.17: 41528167-41528167 on Build GRCh38
rs1030728020 Chr.17: 41528224-41528224 on Build GRCh38
rs769879883 Chr.17: 41528265-41528265 on Build GRCh38
rs999244497 Chr.17: 41528383-41528383 on Build GRCh38
rs755491366 Chr.17: 41528219-41528219 on Build GRCh38
rs376181298 Chr.17: 41528511-41528511 on Build GRCh38
rs143519434 Chr.17: 41528582-41528582 on Build GRCh38
rs199953512 Chr.17: 41528185-41528185 on Build GRCh38
rs200623371 Chr.17: 41528158-41528158 on Build GRCh38
rs1002846655 Chr.17: 41528122-41528122 on Build GRCh38

View Full Product Details