Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 58781131-58781631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JUN
Entrez Gene ID
3725
Gene Name
Jun proto-oncogene, AP-1 transcription factor subunit
Gene Aliases
AP-1, AP1, c-Jun
UniGene
Hs.696684
Gene Chromosome Location
Chr.1: 58780791-58784113 on Build GRCh38
Transcripts
NM_002228.3

SNP Detail

SNP ID Chromosome Location
rs545706650 Chr.1: 58781500-58781500 on Build GRCh38
rs890316290 Chr.1: 58781412-58781412 on Build GRCh38
rs72911506 Chr.1: 58781167-58781167 on Build GRCh38
rs4647016 Chr.1: 58781546-58781546 on Build GRCh38
rs911562589 Chr.1: 58781591-58781591 on Build GRCh38
rs914940270 Chr.1: 58781576-58781576 on Build GRCh38
rs1041558908 Chr.1: 58781437-58781437 on Build GRCh38
rs926302914 Chr.1: 58781219-58781219 on Build GRCh38
rs543321580 Chr.1: 58781246-58781246 on Build GRCh38
rs536918038 Chr.1: 58781158-58781158 on Build GRCh38
rs1042231633 Chr.1: 58781373-58781376 on Build GRCh38
rs970826721 Chr.1: 58781340-58781340 on Build GRCh38
rs2654 Chr.1: 58781575-58781575 on Build GRCh38
rs538298755 Chr.1: 58781391-58781391 on Build GRCh38
rs577480140 Chr.1: 58781176-58781176 on Build GRCh38
rs945289019 Chr.1: 58781335-58781335 on Build GRCh38
rs544988738 Chr.1: 58781595-58781595 on Build GRCh38
rs996134137 Chr.1: 58781263-58781263 on Build GRCh38
rs754061451 Chr.1: 58781544-58781544 on Build GRCh38
rs564885432 Chr.1: 58781602-58781602 on Build GRCh38
rs554499791 Chr.1: 58781519-58781519 on Build GRCh38
rs1053459481 Chr.1: 58781291-58781291 on Build GRCh38
rs980827080 Chr.1: 58781417-58781417 on Build GRCh38
rs555021913 Chr.1: 58781382-58781383 on Build GRCh38
rs1008396823 Chr.1: 58781193-58781193 on Build GRCh38
rs1027257030 Chr.1: 58781271-58781271 on Build GRCh38
rs756167555 Chr.1: 58781316-58781316 on Build GRCh38
rs551399805 Chr.1: 58781164-58781164 on Build GRCh38
rs1008309454 Chr.1: 58781420-58781420 on Build GRCh38
rs915138546 Chr.1: 58781315-58781315 on Build GRCh38
rs866690671 Chr.1: 58781599-58781599 on Build GRCh38
rs952934135 Chr.1: 58781467-58781470 on Build GRCh38
rs997209887 Chr.1: 58781466-58781467 on Build GRCh38
rs576321378 Chr.1: 58781567-58781568 on Build GRCh38
rs1040237232 Chr.1: 58781216-58781216 on Build GRCh38
rs899216514 Chr.1: 58781155-58781163 on Build GRCh38
rs989119122 Chr.1: 58781571-58781571 on Build GRCh38
rs9282771 Chr.1: 58781284-58781284 on Build GRCh38
rs1004199037 Chr.1: 58781293-58781293 on Build GRCh38
rs1015714337 Chr.1: 58781480-58781482 on Build GRCh38
rs556690372 Chr.1: 58781251-58781251 on Build GRCh38
rs542343587 Chr.1: 58781466-58781466 on Build GRCh38
rs926417405 Chr.1: 58781438-58781438 on Build GRCh38
rs1051466508 Chr.1: 58781557-58781557 on Build GRCh38
rs930767622 Chr.1: 58781478-58781491 on Build GRCh38
rs900006123 Chr.1: 58781237-58781237 on Build GRCh38
rs957913542 Chr.1: 58781493-58781493 on Build GRCh38
rs2655 Chr.1: 58781531-58781531 on Build GRCh38

View Full Product Details