Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59815065-59815555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOOK1
Entrez Gene ID
51361
Gene Name
hook microtubule-tethering protein 1
Gene Aliases
HK1
UniGene
Hs.378836
Gene Chromosome Location
Chr.1: 59814861-59876378 on Build GRCh38
Transcripts
NM_015888.4 XM_006710676.1 XM_011541562.2 XM_011541563.1 XM_017001424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5562840 c.18G>A p.P6P Chr.1: 59815135-59815135 on Build GRCh38
6431583 c.2T>C p.M1T Chr.1: 59815119-59815119 on Build GRCh38
5514765 c.57G>T p.M19I Chr.1: 59815174-59815174 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965396085 Chr.1: 59815258-59815258 on Build GRCh38
rs770620727 Chr.1: 59815113-59815113 on Build GRCh38
rs559957345 Chr.1: 59815287-59815287 on Build GRCh38
rs77025030 Chr.1: 59815373-59815373 on Build GRCh38
rs984970634 Chr.1: 59815349-59815349 on Build GRCh38
rs902663908 Chr.1: 59815391-59815391 on Build GRCh38
rs533341434 Chr.1: 59815459-59815459 on Build GRCh38
rs939111062 Chr.1: 59815154-59815154 on Build GRCh38
rs34045351 Chr.1: 59815171-59815171 on Build GRCh38
rs1018023281 Chr.1: 59815276-59815276 on Build GRCh38
rs966900548 Chr.1: 59815201-59815201 on Build GRCh38
rs983990686 Chr.1: 59815478-59815478 on Build GRCh38
rs191651082 Chr.1: 59815396-59815396 on Build GRCh38
rs530946003 Chr.1: 59815428-59815428 on Build GRCh38
rs892297597 Chr.1: 59815213-59815213 on Build GRCh38
rs573300037 Chr.1: 59815311-59815311 on Build GRCh38
rs776273079 Chr.1: 59815134-59815134 on Build GRCh38
rs1031811279 Chr.1: 59815397-59815397 on Build GRCh38
rs867001721 Chr.1: 59815140-59815140 on Build GRCh38
rs965167423 Chr.1: 59815406-59815406 on Build GRCh38
rs1034318735 Chr.1: 59815153-59815153 on Build GRCh38
rs932588634 Chr.1: 59815265-59815265 on Build GRCh38
rs541403194 Chr.1: 59815284-59815284 on Build GRCh38
rs527701415 Chr.1: 59815517-59815520 on Build GRCh38
rs1043967740 Chr.1: 59815534-59815534 on Build GRCh38
rs950849971 Chr.1: 59815337-59815337 on Build GRCh38
rs961165804 Chr.1: 59815501-59815501 on Build GRCh38
rs1027986746 Chr.1: 59815444-59815444 on Build GRCh38
rs1043634276 Chr.1: 59815242-59815242 on Build GRCh38
rs909827231 Chr.1: 59815500-59815500 on Build GRCh38
rs921149641 Chr.1: 59815261-59815261 on Build GRCh38
rs976787882 Chr.1: 59815411-59815411 on Build GRCh38
rs1037111754 Chr.1: 59815141-59815141 on Build GRCh38
rs566803073 Chr.1: 59815531-59815531 on Build GRCh38
rs939447088 Chr.1: 59815535-59815535 on Build GRCh38
rs764131167 Chr.1: 59815338-59815338 on Build GRCh38
rs980428655 Chr.1: 59815405-59815405 on Build GRCh38
rs1011037152 Chr.1: 59815165-59815165 on Build GRCh38
rs895408046 Chr.1: 59815161-59815161 on Build GRCh38
rs906514763 Chr.1: 59815144-59815144 on Build GRCh38
rs532132464 Chr.1: 59815240-59815240 on Build GRCh38
rs868349658 Chr.1: 59815132-59815134 on Build GRCh38
rs977154265 Chr.1: 59815245-59815245 on Build GRCh38
rs939314270 Chr.1: 59815317-59815317 on Build GRCh38
rs759583116 Chr.1: 59815137-59815137 on Build GRCh38
rs551692113 Chr.1: 59815521-59815521 on Build GRCh38
rs1007533857 Chr.1: 59815247-59815247 on Build GRCh38
rs867721033 Chr.1: 59815110-59815110 on Build GRCh38
rs991851600 Chr.1: 59815471-59815471 on Build GRCh38
rs556830992 Chr.1: 59815210-59815210 on Build GRCh38
rs895175293 Chr.1: 59815361-59815361 on Build GRCh38
rs945235341 Chr.1: 59815237-59815237 on Build GRCh38
rs939058865 Chr.1: 59815465-59815465 on Build GRCh38
rs984043036 Chr.1: 59815282-59815282 on Build GRCh38
rs562229374 Chr.1: 59815362-59815362 on Build GRCh38
rs1003563813 Chr.1: 59815147-59815147 on Build GRCh38
rs548347772 Chr.1: 59815314-59815314 on Build GRCh38
rs546157824 Chr.1: 59815289-59815289 on Build GRCh38
rs1055136060 Chr.1: 59815334-59815334 on Build GRCh38
rs932450724 Chr.1: 59815104-59815104 on Build GRCh38
rs147210328 Chr.1: 59815454-59815454 on Build GRCh38
rs540233263 Chr.1: 59815321-59815321 on Build GRCh38
rs1031445511 Chr.1: 59815252-59815252 on Build GRCh38
rs41287714 Chr.1: 59815290-59815290 on Build GRCh38
rs769592128 Chr.1: 59815155-59815155 on Build GRCh38
rs976461186 Chr.1: 59815259-59815259 on Build GRCh38

View Full Product Details