Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 61680968-61681476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM2D1
Entrez Gene ID
83941
Gene Name
TM2 domain containing 1
Gene Aliases
BBP
UniGene
Hs.656790
Gene Chromosome Location
Chr.1: 61681047-61725423 on Build GRCh38
Transcripts
NM_032027.2 NR_135160.1 NR_135161.1 NR_135162.1

SNP Detail

SNP ID Chromosome Location
rs1043548934 Chr.1: 61681372-61681372 on Build GRCh38
rs998208086 Chr.1: 61681419-61681419 on Build GRCh38
rs999311129 Chr.1: 61681022-61681022 on Build GRCh38
rs59562457 Chr.1: 61681116-61681117 on Build GRCh38
rs967348062 Chr.1: 61681013-61681015 on Build GRCh38
rs796592827 Chr.1: 61681117-61681118 on Build GRCh38
rs537110429 Chr.1: 61681294-61681294 on Build GRCh38
rs910041319 Chr.1: 61681114-61681114 on Build GRCh38
rs998968129 Chr.1: 61681381-61681381 on Build GRCh38
rs76794916 Chr.1: 61681289-61681289 on Build GRCh38
rs11539585 Chr.1: 61681347-61681347 on Build GRCh38
rs543373329 Chr.1: 61681423-61681423 on Build GRCh38
rs41289420 Chr.1: 61681106-61681106 on Build GRCh38
rs371977020 Chr.1: 61681162-61681162 on Build GRCh38
rs1051973535 Chr.1: 61681132-61681133 on Build GRCh38
rs113642575 Chr.1: 61681112-61681112 on Build GRCh38
rs779615738 Chr.1: 61681384-61681384 on Build GRCh38
rs1031674591 Chr.1: 61681033-61681033 on Build GRCh38
rs939792072 Chr.1: 61681174-61681174 on Build GRCh38
rs912035270 Chr.1: 61681140-61681140 on Build GRCh38
rs904640140 Chr.1: 61681018-61681018 on Build GRCh38
rs532713785 Chr.1: 61681074-61681074 on Build GRCh38
rs903031523 Chr.1: 61681436-61681436 on Build GRCh38
rs965734952 Chr.1: 61681037-61681037 on Build GRCh38
rs1032143108 Chr.1: 61681398-61681398 on Build GRCh38
rs1028607700 Chr.1: 61681077-61681077 on Build GRCh38
rs578077460 Chr.1: 61681161-61681161 on Build GRCh38
rs925867551 Chr.1: 61681052-61681052 on Build GRCh38
rs1022083539 Chr.1: 61680997-61680997 on Build GRCh38
rs545171070 Chr.1: 61681373-61681373 on Build GRCh38
rs560719998 Chr.1: 61681437-61681437 on Build GRCh38
rs984336889 Chr.1: 61681444-61681444 on Build GRCh38
rs572188821 Chr.1: 61681428-61681428 on Build GRCh38
rs972873706 Chr.1: 61681259-61681259 on Build GRCh38
rs181393594 Chr.1: 61681290-61681290 on Build GRCh38
rs927666801 Chr.1: 61681279-61681279 on Build GRCh38
rs538435816 Chr.1: 61681413-61681413 on Build GRCh38

View Full Product Details