Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137578551-137579085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH7
Entrez Gene ID
92715
Gene Name
diphthamide biosynthesis 7
Gene Aliases
C9orf112, RRT2, WDR85
UniGene
Hs.292570
Gene Chromosome Location
Chr.9: 137554899-137578935 on Build GRCh38
Transcripts
NM_138778.3 XM_005266123.2 XM_005266126.3 XM_005266128.2 XM_005266129.2 XM_005266130.2 XM_011519188.1 XM_011519189.1 XM_011519190.2 XM_011519191.2 XM_011519192.2 XM_011519193.2 XM_011519194.1 XM_011519199.2 XM_011519200.1 XM_017015289.1 XM_017015290.1 XM_017015292.1 XM_017015293.1 XM_017015294.1 XM_017015295.1 XM_017015297.1 XM_017015298.1 XM_017015300.1 XM_017015301.1 XM_017015302.1 XM_017015303.1 XM_017015304.1 XM_017015305.1 XM_017015306.1 XM_017015307.1 XM_017015308.1 XM_017015309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7307920 c.152A>G p.K51R Chr.9: 137578626-137578626 on Build GRCh38
7365361 c.118C>T p.R40W Chr.9: 137578660-137578660 on Build GRCh38
6517603 c.72G>T p.L24L Chr.9: 137578706-137578706 on Build GRCh38
3699607 c.76G>T p.G26C Chr.9: 137578702-137578702 on Build GRCh38
4805599 c.53C>A p.S18* Chr.9: 137578725-137578725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369710922 Chr.9: 137578688-137578688 on Build GRCh38
rs753049264 Chr.9: 137578746-137578746 on Build GRCh38
rs760908119 Chr.9: 137578673-137578673 on Build GRCh38
rs544406780 Chr.9: 137578897-137578897 on Build GRCh38
rs756697564 Chr.9: 137578772-137578772 on Build GRCh38
rs1046579649 Chr.9: 137578954-137578954 on Build GRCh38
rs955339207 Chr.9: 137578839-137578839 on Build GRCh38
rs1024283432 Chr.9: 137578972-137578972 on Build GRCh38
rs772232435 Chr.9: 137578669-137578669 on Build GRCh38
rs977258113 Chr.9: 137578896-137578896 on Build GRCh38
rs546131967 Chr.9: 137579040-137579040 on Build GRCh38
rs977092942 Chr.9: 137579058-137579058 on Build GRCh38
rs114136335 Chr.9: 137578920-137578920 on Build GRCh38
rs985481765 Chr.9: 137578867-137578867 on Build GRCh38
rs962353615 Chr.9: 137578974-137578974 on Build GRCh38
rs1026181042 Chr.9: 137579046-137579046 on Build GRCh38
rs918426801 Chr.9: 137578601-137578601 on Build GRCh38
rs975329734 Chr.9: 137578820-137578820 on Build GRCh38
rs116587551 Chr.9: 137579061-137579061 on Build GRCh38
rs902253180 Chr.9: 137578961-137578961 on Build GRCh38
rs3763610 Chr.9: 137578936-137578936 on Build GRCh38
rs942592615 Chr.9: 137579036-137579036 on Build GRCh38
rs560245713 Chr.9: 137579028-137579028 on Build GRCh38
rs575540920 Chr.9: 137578783-137578783 on Build GRCh38
rs868549348 Chr.9: 137578741-137578741 on Build GRCh38
rs1020423445 Chr.9: 137578743-137578743 on Build GRCh38
rs780683373 Chr.9: 137578790-137578790 on Build GRCh38
rs900946278 Chr.9: 137578710-137578710 on Build GRCh38
rs771252462 Chr.9: 137578940-137578940 on Build GRCh38
rs368026956 Chr.9: 137578857-137578857 on Build GRCh38
rs745767329 Chr.9: 137578803-137578803 on Build GRCh38
rs200787106 Chr.9: 137578659-137578659 on Build GRCh38
rs941742421 Chr.9: 137578764-137578764 on Build GRCh38
rs1013733056 Chr.9: 137578619-137578619 on Build GRCh38
rs1034599526 Chr.9: 137578935-137578935 on Build GRCh38
rs759648645 Chr.9: 137578995-137578995 on Build GRCh38
rs751054772 Chr.9: 137578763-137578763 on Build GRCh38
rs929130050 Chr.9: 137578751-137578751 on Build GRCh38
rs958273218 Chr.9: 137578864-137578864 on Build GRCh38
rs909159807 Chr.9: 137578916-137578916 on Build GRCh38
rs1000401452 Chr.9: 137578927-137578927 on Build GRCh38
rs1041188118 Chr.9: 137578699-137578699 on Build GRCh38
rs566769908 Chr.9: 137578621-137578621 on Build GRCh38
rs1028188311 Chr.9: 137578993-137578993 on Build GRCh38
rs931204671 Chr.9: 137578658-137578658 on Build GRCh38
rs966875733 Chr.9: 137578634-137578634 on Build GRCh38
rs963717994 Chr.9: 137578632-137578632 on Build GRCh38
rs1030009419 Chr.9: 137578829-137578829 on Build GRCh38
rs918388085 Chr.9: 137578855-137578855 on Build GRCh38
rs867009348 Chr.9: 137578756-137578756 on Build GRCh38
rs972574571 Chr.9: 137578831-137578831 on Build GRCh38
rs1036894340 Chr.9: 137578583-137578583 on Build GRCh38
rs983304317 Chr.9: 137578915-137578915 on Build GRCh38
rs760149598 Chr.9: 137578738-137578738 on Build GRCh38
rs777032503 Chr.9: 137578984-137578984 on Build GRCh38
rs902684943 Chr.9: 137578849-137578849 on Build GRCh38
rs944085164 Chr.9: 137578705-137578705 on Build GRCh38
rs931155953 Chr.9: 137578856-137578856 on Build GRCh38
rs959114578 Chr.9: 137578605-137578605 on Build GRCh38
rs911176644 Chr.9: 137578872-137578872 on Build GRCh38
rs758033375 Chr.9: 137578759-137578759 on Build GRCh38
rs1014931613 Chr.9: 137578959-137578959 on Build GRCh38
rs745995757 Chr.9: 137579023-137579023 on Build GRCh38
rs975968697 Chr.9: 137578711-137578711 on Build GRCh38
rs1033426192 Chr.9: 137578597-137578597 on Build GRCh38
rs1007160497 Chr.9: 137579002-137579002 on Build GRCh38
rs945793150 Chr.9: 137578904-137578904 on Build GRCh38
rs1013303273 Chr.9: 137578805-137578805 on Build GRCh38
rs1024703896 Chr.9: 137578822-137578822 on Build GRCh38
rs773315177 Chr.9: 137578670-137578670 on Build GRCh38
rs918638392 Chr.9: 137578900-137578900 on Build GRCh38
rs543808150 Chr.9: 137578902-137578902 on Build GRCh38
rs1018274219 Chr.9: 137578884-137578884 on Build GRCh38
rs917306612 Chr.9: 137578807-137578807 on Build GRCh38
rs1000141894 Chr.9: 137578977-137578977 on Build GRCh38
rs1040745840 Chr.9: 137578890-137578890 on Build GRCh38
rs958689372 Chr.9: 137578801-137578801 on Build GRCh38
rs999666199 Chr.9: 137578850-137578850 on Build GRCh38
rs968895730 Chr.9: 137578757-137578757 on Build GRCh38
rs766982098 Chr.9: 137578682-137578682 on Build GRCh38
rs952543983 Chr.9: 137579004-137579004 on Build GRCh38
rs1041863000 Chr.9: 137578911-137578911 on Build GRCh38
rs942353246 Chr.9: 137578874-137578874 on Build GRCh38
rs894090745 Chr.9: 137578986-137578986 on Build GRCh38
rs532361559 Chr.9: 137579030-137579030 on Build GRCh38
rs992850432 Chr.9: 137578806-137578806 on Build GRCh38
rs879595577 Chr.9: 137579066-137579066 on Build GRCh38
rs777354928 Chr.9: 137578762-137578762 on Build GRCh38
rs765584512 Chr.9: 137578742-137578742 on Build GRCh38
rs1006544670 Chr.9: 137578692-137578692 on Build GRCh38
rs962121903 Chr.9: 137578826-137578826 on Build GRCh38

View Full Product Details