Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 2903862-2904401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSD
Entrez Gene ID
414
Gene Name
arylsulfatase D
Gene Aliases
ASD
UniGene
Hs.528631
Gene Chromosome Location
Chr.X: 2903970-2929375 on Build GRCh38
Transcripts
NM_001669.3 XM_005274514.2 XM_005274515.2

SNP Detail

SNP ID Chromosome Location
rs181104269 Chr.X: 2904294-2904294 on Build GRCh38
rs756744949 Chr.X: 2904007-2904007 on Build GRCh38
rs376437972 Chr.X: 2903937-2903938 on Build GRCh38
rs990872148 Chr.X: 2904246-2904246 on Build GRCh38
rs766890234 Chr.X: 2904117-2904117 on Build GRCh38
rs4141210 Chr.X: 2903937-2903937 on Build GRCh38
rs1010461297 Chr.X: 2903912-2903912 on Build GRCh38
rs150774859 Chr.X: 2903980-2903980 on Build GRCh38
rs9334 Chr.X: 2904212-2904212 on Build GRCh38
rs4141211 Chr.X: 2903938-2903938 on Build GRCh38
rs779804791 Chr.X: 2904326-2904326 on Build GRCh38
rs918512097 Chr.X: 2904300-2904300 on Build GRCh38
rs201692230 Chr.X: 2903951-2903954 on Build GRCh38
rs925650887 Chr.X: 2904258-2904258 on Build GRCh38
rs866754416 Chr.X: 2904086-2904086 on Build GRCh38
rs757195960 Chr.X: 2903967-2903967 on Build GRCh38
rs1035174517 Chr.X: 2904093-2904093 on Build GRCh38
rs893381294 Chr.X: 2903901-2903901 on Build GRCh38
rs933811016 Chr.X: 2904342-2904342 on Build GRCh38
rs754548011 Chr.X: 2904365-2904365 on Build GRCh38
rs1016582348 Chr.X: 2904267-2904267 on Build GRCh38
rs1003048932 Chr.X: 2904092-2904092 on Build GRCh38
rs959191294 Chr.X: 2904195-2904195 on Build GRCh38
rs972586844 Chr.X: 2904295-2904295 on Build GRCh38
rs1020565137 Chr.X: 2903979-2903979 on Build GRCh38

View Full Product Details