Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 3320315-3320815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXRA5
Entrez Gene ID
25878
Gene Name
matrix remodeling associated 5
Gene Aliases
-
UniGene
Hs.369422
Gene Chromosome Location
Chr.X: 3308565-3347098 on Build GRCh38
Transcripts
NM_015419.3 XM_005274485.2

SNP Detail

SNP ID Chromosome Location
rs751478479 Chr.X: 3320673-3320673 on Build GRCh38
rs766261229 Chr.X: 3320669-3320669 on Build GRCh38
rs915596717 Chr.X: 3320631-3320631 on Build GRCh38
rs982684282 Chr.X: 3320382-3320382 on Build GRCh38
rs763560468 Chr.X: 3320613-3320613 on Build GRCh38
rs779975747 Chr.X: 3320446-3320446 on Build GRCh38
rs763762016 Chr.X: 3320408-3320408 on Build GRCh38
rs760162200 Chr.X: 3320349-3320349 on Build GRCh38
rs764222142 Chr.X: 3320780-3320780 on Build GRCh38
rs772649054 Chr.X: 3320592-3320592 on Build GRCh38
rs761437725 Chr.X: 3320618-3320618 on Build GRCh38
rs776314231 Chr.X: 3320616-3320616 on Build GRCh38
rs34070401 Chr.X: 3320389-3320390 on Build GRCh38
rs769799144 Chr.X: 3320480-3320480 on Build GRCh38
rs138419461 Chr.X: 3320747-3320747 on Build GRCh38
rs753454467 Chr.X: 3320409-3320409 on Build GRCh38
rs776042560 Chr.X: 3320745-3320745 on Build GRCh38
rs145151600 Chr.X: 3320594-3320594 on Build GRCh38
rs778007682 Chr.X: 3320504-3320504 on Build GRCh38
rs767759204 Chr.X: 3320437-3320445 on Build GRCh38
rs201840964 Chr.X: 3320475-3320475 on Build GRCh38
rs777758658 Chr.X: 3320689-3320689 on Build GRCh38
rs867386062 Chr.X: 3320698-3320698 on Build GRCh38
rs867173760 Chr.X: 3320661-3320661 on Build GRCh38
rs966926453 Chr.X: 3320376-3320376 on Build GRCh38
rs758385816 Chr.X: 3320432-3320432 on Build GRCh38
rs146888977 Chr.X: 3320606-3320606 on Build GRCh38
rs777185808 Chr.X: 3320368-3320368 on Build GRCh38
rs756200054 Chr.X: 3320679-3320679 on Build GRCh38
rs868582760 Chr.X: 3320548-3320548 on Build GRCh38
rs150094241 Chr.X: 3320746-3320746 on Build GRCh38
rs751486197 Chr.X: 3320767-3320767 on Build GRCh38
rs758189271 Chr.X: 3320660-3320660 on Build GRCh38
rs771393570 Chr.X: 3320528-3320528 on Build GRCh38
rs772587396 Chr.X: 3320737-3320737 on Build GRCh38
rs757052581 Chr.X: 3320414-3320414 on Build GRCh38
rs76535805 Chr.X: 3320789-3320789 on Build GRCh38
rs750157854 Chr.X: 3320637-3320637 on Build GRCh38
rs1011174456 Chr.X: 3320359-3320359 on Build GRCh38
rs765000731 Chr.X: 3320625-3320625 on Build GRCh38
rs749524985 Chr.X: 3320505-3320505 on Build GRCh38
rs868250427 Chr.X: 3320437-3320437 on Build GRCh38
rs774924750 Chr.X: 3320564-3320564 on Build GRCh38
rs868313548 Chr.X: 3320709-3320709 on Build GRCh38
rs754940257 Chr.X: 3320778-3320778 on Build GRCh38
rs79552860 Chr.X: 3320435-3320435 on Build GRCh38
rs760117677 Chr.X: 3320391-3320391 on Build GRCh38
rs747125005 Chr.X: 3320427-3320427 on Build GRCh38
rs779291851 Chr.X: 3320725-3320725 on Build GRCh38
rs762235263 Chr.X: 3320374-3320374 on Build GRCh38
rs762691003 Chr.X: 3320758-3320758 on Build GRCh38
rs764799373 Chr.X: 3320755-3320755 on Build GRCh38
rs781768107 Chr.X: 3320722-3320722 on Build GRCh38
rs748188513 Chr.X: 3320457-3320457 on Build GRCh38
rs774980117 Chr.X: 3320340-3320340 on Build GRCh38
rs760182088 Chr.X: 3320599-3320599 on Build GRCh38
rs112302509 Chr.X: 3320417-3320417 on Build GRCh38
rs371923940 Chr.X: 3320731-3320731 on Build GRCh38
rs375262438 Chr.X: 3320756-3320756 on Build GRCh38
rs779238626 Chr.X: 3320794-3320794 on Build GRCh38
rs776059397 Chr.X: 3320748-3320748 on Build GRCh38
rs753805632 Chr.X: 3320786-3320786 on Build GRCh38
rs867743785 Chr.X: 3320464-3320464 on Build GRCh38
rs1974522 Chr.X: 3320692-3320692 on Build GRCh38
rs369503229 Chr.X: 3320766-3320766 on Build GRCh38
rs941672347 Chr.X: 3320708-3320708 on Build GRCh38
rs760108956 Chr.X: 3320571-3320571 on Build GRCh38
rs866073929 Chr.X: 3320418-3320418 on Build GRCh38
rs991160394 Chr.X: 3320617-3320617 on Build GRCh38

View Full Product Details