Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9653979-9654526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBL1X
Entrez Gene ID
6907
Gene Name
transducin (beta)-like 1X-linked
Gene Aliases
EBI, SMAP55, TBL1
UniGene
Hs.495656
Gene Chromosome Location
Chr.X: 9463295-9719740 on Build GRCh38
Transcripts
NM_001139466.1 NM_001139467.1 NM_001139468.1 NM_005647.3 XM_011545571.2

SNP Detail

SNP ID Chromosome Location
rs765556216 Chr.X: 9654388-9654388 on Build GRCh38
rs374600399 Chr.X: 9654282-9654282 on Build GRCh38
rs771607809 Chr.X: 9654246-9654246 on Build GRCh38
rs369334079 Chr.X: 9654410-9654410 on Build GRCh38
rs1053915074 Chr.X: 9654095-9654095 on Build GRCh38
rs367680489 Chr.X: 9654155-9654155 on Build GRCh38
rs764823206 Chr.X: 9654369-9654369 on Build GRCh38
rs767003874 Chr.X: 9654437-9654437 on Build GRCh38
rs902734522 Chr.X: 9654350-9654350 on Build GRCh38
rs1023756578 Chr.X: 9654386-9654386 on Build GRCh38
rs376248170 Chr.X: 9654408-9654408 on Build GRCh38
rs751546228 Chr.X: 9654204-9654205 on Build GRCh38
rs756892029 Chr.X: 9654184-9654184 on Build GRCh38
rs1039445025 Chr.X: 9654234-9654234 on Build GRCh38
rs756463135 Chr.X: 9654411-9654413 on Build GRCh38
rs763022120 Chr.X: 9654165-9654167 on Build GRCh38
rs777314594 Chr.X: 9654366-9654366 on Build GRCh38
rs376246266 Chr.X: 9654165-9654165 on Build GRCh38
rs781174429 Chr.X: 9654269-9654269 on Build GRCh38
rs142075238 Chr.X: 9654237-9654237 on Build GRCh38
rs771048383 Chr.X: 9654279-9654279 on Build GRCh38
rs397823623 Chr.X: 9654428-9654429 on Build GRCh38
rs769134406 Chr.X: 9654357-9654357 on Build GRCh38
rs761182554 Chr.X: 9654345-9654345 on Build GRCh38
rs113626940 Chr.X: 9654426-9654427 on Build GRCh38
rs34645456 Chr.X: 9654129-9654130 on Build GRCh38
rs190435148 Chr.X: 9654342-9654342 on Build GRCh38
rs893986168 Chr.X: 9654372-9654372 on Build GRCh38
rs922253594 Chr.X: 9654242-9654242 on Build GRCh38
rs943524131 Chr.X: 9654011-9654011 on Build GRCh38
rs918539560 Chr.X: 9654003-9654003 on Build GRCh38
rs775938372 Chr.X: 9654344-9654344 on Build GRCh38
rs781557516 Chr.X: 9654413-9654414 on Build GRCh38
rs200176756 Chr.X: 9654429-9654429 on Build GRCh38
rs146358253 Chr.X: 9654271-9654271 on Build GRCh38
rs1003582821 Chr.X: 9654389-9654389 on Build GRCh38
rs368960787 Chr.X: 9654330-9654330 on Build GRCh38
rs951285252 Chr.X: 9654053-9654053 on Build GRCh38
rs751243843 Chr.X: 9654248-9654248 on Build GRCh38
rs57370748 Chr.X: 9654497-9654497 on Build GRCh38
rs71801536 Chr.X: 9654411-9654412 on Build GRCh38
rs186053249 Chr.X: 9654196-9654196 on Build GRCh38
rs764268848 Chr.X: 9654166-9654167 on Build GRCh38
rs1000194609 Chr.X: 9654352-9654352 on Build GRCh38
rs373041302 Chr.X: 9654407-9654407 on Build GRCh38
rs1035626789 Chr.X: 9654411-9654416 on Build GRCh38
rs1053930762 Chr.X: 9654364-9654364 on Build GRCh38
rs780012380 Chr.X: 9654489-9654489 on Build GRCh38
rs183188932 Chr.X: 9654405-9654405 on Build GRCh38
rs34335400 Chr.X: 9654410-9654411 on Build GRCh38
rs951294601 Chr.X: 9654485-9654485 on Build GRCh38
rs761075485 Chr.X: 9654126-9654126 on Build GRCh38
rs750136161 Chr.X: 9654221-9654221 on Build GRCh38
rs143778785 Chr.X: 9654488-9654488 on Build GRCh38
rs201434884 Chr.X: 9654180-9654180 on Build GRCh38
rs1012316609 Chr.X: 9654382-9654382 on Build GRCh38
rs200551470 Chr.X: 9654249-9654249 on Build GRCh38
rs749999807 Chr.X: 9654482-9654482 on Build GRCh38

View Full Product Details