Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9691922-9692414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBL1X
Entrez Gene ID
6907
Gene Name
transducin (beta)-like 1X-linked
Gene Aliases
EBI, SMAP55, TBL1
UniGene
Hs.495656
Gene Chromosome Location
Chr.X: 9463295-9719740 on Build GRCh38
Transcripts
NM_001139466.1 NM_001139467.1 NM_001139468.1 NM_005647.3 XM_011545571.2

SNP Detail

SNP ID Chromosome Location
rs371683874 Chr.X: 9692085-9692085 on Build GRCh38
rs569905842 Chr.X: 9692346-9692346 on Build GRCh38
rs143231361 Chr.X: 9692215-9692215 on Build GRCh38
rs749555321 Chr.X: 9692252-9692252 on Build GRCh38
rs149554270 Chr.X: 9692146-9692146 on Build GRCh38
rs778526595 Chr.X: 9692076-9692076 on Build GRCh38
rs778770075 Chr.X: 9692195-9692195 on Build GRCh38
rs753253491 Chr.X: 9692150-9692150 on Build GRCh38
rs772536314 Chr.X: 9692280-9692280 on Build GRCh38
rs144071784 Chr.X: 9692327-9692327 on Build GRCh38
rs148306368 Chr.X: 9692245-9692245 on Build GRCh38
rs999242753 Chr.X: 9692377-9692377 on Build GRCh38
rs760304993 Chr.X: 9692157-9692157 on Build GRCh38
rs776190984 Chr.X: 9692287-9692287 on Build GRCh38
rs902275050 Chr.X: 9692363-9692363 on Build GRCh38
rs963202405 Chr.X: 9692263-9692263 on Build GRCh38
rs771000158 Chr.X: 9692271-9692271 on Build GRCh38
rs7062490 Chr.X: 9692110-9692110 on Build GRCh38
rs1044498065 Chr.X: 9691997-9691997 on Build GRCh38
rs903277655 Chr.X: 9692336-9692336 on Build GRCh38
rs7057012 Chr.X: 9692109-9692109 on Build GRCh38
rs138100745 Chr.X: 9692134-9692134 on Build GRCh38
rs758196673 Chr.X: 9692203-9692203 on Build GRCh38
rs769909829 Chr.X: 9692239-9692239 on Build GRCh38
rs753571565 Chr.X: 9692162-9692162 on Build GRCh38
rs368174968 Chr.X: 9692077-9692077 on Build GRCh38
rs1028752483 Chr.X: 9692313-9692313 on Build GRCh38
rs201963539 Chr.X: 9692216-9692216 on Build GRCh38
rs7062597 Chr.X: 9692116-9692116 on Build GRCh38
rs757127324 Chr.X: 9692066-9692066 on Build GRCh38
rs761217443 Chr.X: 9692291-9692291 on Build GRCh38
rs148172560 Chr.X: 9692295-9692295 on Build GRCh38
rs769924371 Chr.X: 9692017-9692017 on Build GRCh38
rs765889696 Chr.X: 9692221-9692221 on Build GRCh38
rs942939027 Chr.X: 9692273-9692273 on Build GRCh38
rs377690435 Chr.X: 9691982-9691983 on Build GRCh38
rs775941741 Chr.X: 9692082-9692082 on Build GRCh38
rs1043097155 Chr.X: 9692305-9692305 on Build GRCh38
rs386823863 Chr.X: 9692109-9692110 on Build GRCh38
rs759886484 Chr.X: 9692279-9692279 on Build GRCh38
rs767968686 Chr.X: 9692149-9692149 on Build GRCh38
rs763679092 Chr.X: 9692161-9692161 on Build GRCh38
rs764881543 Chr.X: 9692372-9692372 on Build GRCh38
rs765101713 Chr.X: 9692066-9692066 on Build GRCh38
rs780100232 Chr.X: 9692050-9692050 on Build GRCh38
rs750124548 Chr.X: 9692199-9692199 on Build GRCh38
rs756823483 Chr.X: 9692179-9692179 on Build GRCh38
rs941237332 Chr.X: 9692084-9692084 on Build GRCh38
rs774619557 Chr.X: 9692277-9692277 on Build GRCh38
rs781258406 Chr.X: 9692090-9692090 on Build GRCh38
rs774513471 Chr.X: 9692141-9692141 on Build GRCh38
rs1020440057 Chr.X: 9692343-9692343 on Build GRCh38
rs748282665 Chr.X: 9692093-9692093 on Build GRCh38
rs753336586 Chr.X: 9692299-9692299 on Build GRCh38

View Full Product Details