Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9873549-9874078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHROOM2
Entrez Gene ID
357
Gene Name
shroom family member 2
Gene Aliases
APXL, HSAPXL
UniGene
Hs.567236
Gene Chromosome Location
Chr.X: 9786406-9949443 on Build GRCh38
Transcripts
NM_001649.3 XM_005274500.4

SNP Detail

SNP ID Chromosome Location
rs758297863 Chr.X: 9873637-9873637 on Build GRCh38
rs372682515 Chr.X: 9873829-9873829 on Build GRCh38
rs149912209 Chr.X: 9873681-9873681 on Build GRCh38
rs755936634 Chr.X: 9873950-9873950 on Build GRCh38
rs761019672 Chr.X: 9873686-9873686 on Build GRCh38
rs201114383 Chr.X: 9873674-9873674 on Build GRCh38
rs779929341 Chr.X: 9873638-9873638 on Build GRCh38
rs149561570 Chr.X: 9873906-9873906 on Build GRCh38
rs146757558 Chr.X: 9873753-9873753 on Build GRCh38
rs981826009 Chr.X: 9873943-9873943 on Build GRCh38
rs202237359 Chr.X: 9873835-9873835 on Build GRCh38
rs990599228 Chr.X: 9873585-9873585 on Build GRCh38
rs758068903 Chr.X: 9873738-9873738 on Build GRCh38
rs778904040 Chr.X: 9873808-9873808 on Build GRCh38
rs918310510 Chr.X: 9873953-9873953 on Build GRCh38
rs569377408 Chr.X: 9873989-9873989 on Build GRCh38
rs868696257 Chr.X: 9873757-9873757 on Build GRCh38
rs113195924 Chr.X: 9873580-9873580 on Build GRCh38
rs1046291397 Chr.X: 9874035-9874035 on Build GRCh38
rs368631870 Chr.X: 9873813-9873813 on Build GRCh38
rs1039253904 Chr.X: 9873992-9873992 on Build GRCh38
rs1047996391 Chr.X: 9873712-9873712 on Build GRCh38
rs984180965 Chr.X: 9873971-9873971 on Build GRCh38
rs374833426 Chr.X: 9873672-9873672 on Build GRCh38
rs753351952 Chr.X: 9873605-9873605 on Build GRCh38
rs750045862 Chr.X: 9873623-9873623 on Build GRCh38
rs371447346 Chr.X: 9873617-9873617 on Build GRCh38
rs201633094 Chr.X: 9873675-9873675 on Build GRCh38
rs34465284 Chr.X: 9874008-9874009 on Build GRCh38
rs908800096 Chr.X: 9873825-9873825 on Build GRCh38
rs199860180 Chr.X: 9873652-9873652 on Build GRCh38
rs879221119 Chr.X: 9873687-9873687 on Build GRCh38
rs140479341 Chr.X: 9873795-9873795 on Build GRCh38
rs756760664 Chr.X: 9873616-9873616 on Build GRCh38
rs149096987 Chr.X: 9873694-9873694 on Build GRCh38
rs754772424 Chr.X: 9873771-9873771 on Build GRCh38
rs146442193 Chr.X: 9873937-9873937 on Build GRCh38
rs909427616 Chr.X: 9873723-9873723 on Build GRCh38
rs936358491 Chr.X: 9873930-9873930 on Build GRCh38
rs201837863 Chr.X: 9873708-9873708 on Build GRCh38
rs781044272 Chr.X: 9873779-9873779 on Build GRCh38
rs1032289318 Chr.X: 9873901-9873901 on Build GRCh38
rs755211227 Chr.X: 9874009-9874009 on Build GRCh38
rs909492101 Chr.X: 9873973-9873973 on Build GRCh38
rs145000598 Chr.X: 9873683-9873683 on Build GRCh38
rs866150686 Chr.X: 9873889-9873889 on Build GRCh38
rs367719751 Chr.X: 9873603-9873603 on Build GRCh38
rs999457116 Chr.X: 9873579-9873579 on Build GRCh38
rs757183152 Chr.X: 9873887-9873887 on Build GRCh38
rs1012072677 Chr.X: 9873908-9873908 on Build GRCh38
rs922102941 Chr.X: 9873993-9873993 on Build GRCh38
rs931705837 Chr.X: 9873572-9873573 on Build GRCh38
rs139542964 Chr.X: 9873678-9873678 on Build GRCh38
rs976903787 Chr.X: 9873877-9873877 on Build GRCh38
rs138116190 Chr.X: 9873834-9873834 on Build GRCh38
rs749414935 Chr.X: 9873987-9873987 on Build GRCh38
rs750103047 Chr.X: 9873711-9873711 on Build GRCh38
rs985898499 Chr.X: 9873905-9873905 on Build GRCh38
rs755574980 Chr.X: 9873789-9873789 on Build GRCh38
rs1023415028 Chr.X: 9873928-9873928 on Build GRCh38
rs780430548 Chr.X: 9873954-9873954 on Build GRCh38
rs73474560 Chr.X: 9873584-9873584 on Build GRCh38
rs751468736 Chr.X: 9873759-9873759 on Build GRCh38
rs951094743 Chr.X: 9873957-9873957 on Build GRCh38
rs771024837 Chr.X: 9873677-9873677 on Build GRCh38
rs142199040 Chr.X: 9873709-9873709 on Build GRCh38
rs777173762 Chr.X: 9873699-9873699 on Build GRCh38
rs200846360 Chr.X: 9873724-9873724 on Build GRCh38
rs764891545 Chr.X: 9873783-9873783 on Build GRCh38
rs772625017 Chr.X: 9873682-9873682 on Build GRCh38
rs371735679 Chr.X: 9873836-9873836 on Build GRCh38
rs762640194 Chr.X: 9873596-9873596 on Build GRCh38
rs777299231 Chr.X: 9873886-9873886 on Build GRCh38

View Full Product Details