Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 12886206-12886729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR7
Entrez Gene ID
51284
Gene Name
toll like receptor 7
Gene Aliases
TLR7-like
UniGene
Hs.659215
Gene Chromosome Location
Chr.X: 12867083-12890361 on Build GRCh38
Transcripts
NM_016562.3

SNP Detail

SNP ID Chromosome Location
rs755359069 Chr.X: 12886577-12886577 on Build GRCh38
rs201804650 Chr.X: 12886362-12886362 on Build GRCh38
rs34772592 Chr.X: 12886312-12886313 on Build GRCh38
rs867568508 Chr.X: 12886250-12886250 on Build GRCh38
rs141848808 Chr.X: 12886320-12886320 on Build GRCh38
rs140793578 Chr.X: 12886537-12886537 on Build GRCh38
rs778363770 Chr.X: 12886642-12886642 on Build GRCh38
rs147254880 Chr.X: 12886426-12886426 on Build GRCh38
rs1049745959 Chr.X: 12886585-12886585 on Build GRCh38
rs747359081 Chr.X: 12886234-12886234 on Build GRCh38
rs748809108 Chr.X: 12886660-12886660 on Build GRCh38
rs765435694 Chr.X: 12886705-12886705 on Build GRCh38
rs201564398 Chr.X: 12886321-12886321 on Build GRCh38
rs1012897715 Chr.X: 12886692-12886692 on Build GRCh38
rs201232148 Chr.X: 12886456-12886456 on Build GRCh38
rs200549906 Chr.X: 12886561-12886561 on Build GRCh38
rs368187775 Chr.X: 12886372-12886372 on Build GRCh38
rs773983742 Chr.X: 12886441-12886441 on Build GRCh38
rs781601963 Chr.X: 12886578-12886578 on Build GRCh38
rs5743780 Chr.X: 12886543-12886543 on Build GRCh38
rs767358918 Chr.X: 12886322-12886322 on Build GRCh38
rs778485716 Chr.X: 12886670-12886670 on Build GRCh38
rs750684005 Chr.X: 12886357-12886357 on Build GRCh38
rs867083961 Chr.X: 12886233-12886233 on Build GRCh38
rs769481664 Chr.X: 12886411-12886411 on Build GRCh38
rs868378372 Chr.X: 12886314-12886314 on Build GRCh38
rs770468749 Chr.X: 12886661-12886661 on Build GRCh38
rs200661477 Chr.X: 12886330-12886330 on Build GRCh38
rs922684733 Chr.X: 12886571-12886571 on Build GRCh38
rs201092041 Chr.X: 12886389-12886389 on Build GRCh38
rs760573915 Chr.X: 12886523-12886523 on Build GRCh38
rs769066395 Chr.X: 12886255-12886255 on Build GRCh38
rs867293797 Chr.X: 12886477-12886477 on Build GRCh38
rs201359066 Chr.X: 12886451-12886451 on Build GRCh38
rs144770372 Chr.X: 12886540-12886540 on Build GRCh38
rs202063253 Chr.X: 12886396-12886396 on Build GRCh38
rs978435766 Chr.X: 12886504-12886504 on Build GRCh38
rs757802922 Chr.X: 12886699-12886699 on Build GRCh38
rs758416427 Chr.X: 12886541-12886541 on Build GRCh38
rs200463301 Chr.X: 12886508-12886508 on Build GRCh38
rs368887343 Chr.X: 12886276-12886276 on Build GRCh38
rs747349276 Chr.X: 12886379-12886379 on Build GRCh38
rs745510673 Chr.X: 12886681-12886681 on Build GRCh38
rs769426552 Chr.X: 12886288-12886288 on Build GRCh38
rs748708984 Chr.X: 12886634-12886634 on Build GRCh38
rs539198596 Chr.X: 12886624-12886624 on Build GRCh38
rs201181342 Chr.X: 12886586-12886586 on Build GRCh38
rs770716877 Chr.X: 12886428-12886428 on Build GRCh38
rs756706437 Chr.X: 12886635-12886635 on Build GRCh38
rs199881624 Chr.X: 12886403-12886403 on Build GRCh38
rs199700914 Chr.X: 12886639-12886639 on Build GRCh38
rs202129610 Chr.X: 12886503-12886503 on Build GRCh38
rs748655746 Chr.X: 12886395-12886395 on Build GRCh38
rs1042239368 Chr.X: 12886474-12886474 on Build GRCh38
rs762676425 Chr.X: 12886315-12886315 on Build GRCh38
rs752972366 Chr.X: 12886420-12886420 on Build GRCh38
rs759793723 Chr.X: 12886344-12886344 on Build GRCh38
rs199567286 Chr.X: 12886273-12886273 on Build GRCh38
rs867918501 Chr.X: 12886313-12886313 on Build GRCh38
rs773060173 Chr.X: 12886291-12886291 on Build GRCh38
rs200031329 Chr.X: 12886324-12886324 on Build GRCh38
rs765278612 Chr.X: 12886356-12886356 on Build GRCh38
rs1003287550 Chr.X: 12886266-12886266 on Build GRCh38
rs767328024 Chr.X: 12886488-12886488 on Build GRCh38
rs200146658 Chr.X: 12886371-12886371 on Build GRCh38
rs781655586 Chr.X: 12886394-12886394 on Build GRCh38

View Full Product Details