Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130704145-130704653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC2
Entrez Gene ID
23404
Gene Name
exosome component 2
Gene Aliases
RRP4, Rrp4p, hRrp4p, p7
UniGene
Hs.654643
Gene Chromosome Location
Chr.9: 130693760-130704894 on Build GRCh38
Transcripts
NM_001282708.1 NM_001282709.1 NM_014285.6 NR_104230.1 XM_005272176.2 XM_006717023.2 XM_017014558.1

SNP Detail

SNP ID Chromosome Location
rs1046506147 Chr.9: 130704595-130704595 on Build GRCh38
rs540222300 Chr.9: 130704274-130704274 on Build GRCh38
rs984342557 Chr.9: 130704177-130704177 on Build GRCh38
rs559824550 Chr.9: 130704281-130704281 on Build GRCh38
rs966405415 Chr.9: 130704485-130704485 on Build GRCh38
rs11552574 Chr.9: 130704171-130704171 on Build GRCh38
rs533198686 Chr.9: 130704585-130704585 on Build GRCh38
rs929340455 Chr.9: 130704287-130704287 on Build GRCh38
rs935868688 Chr.9: 130704315-130704315 on Build GRCh38
rs1139028 Chr.9: 130704366-130704366 on Build GRCh38
rs910450402 Chr.9: 130704613-130704613 on Build GRCh38
rs977231087 Chr.9: 130704284-130704284 on Build GRCh38
rs752674730 Chr.9: 130704348-130704348 on Build GRCh38
rs991934751 Chr.9: 130704259-130704259 on Build GRCh38
rs759378754 Chr.9: 130704169-130704169 on Build GRCh38
rs924452624 Chr.9: 130704311-130704311 on Build GRCh38
rs1024747638 Chr.9: 130704512-130704512 on Build GRCh38
rs954856425 Chr.9: 130704504-130704504 on Build GRCh38
rs546954827 Chr.9: 130704623-130704623 on Build GRCh38
rs573467191 Chr.9: 130704328-130704328 on Build GRCh38
rs372332656 Chr.9: 130704618-130704618 on Build GRCh38
rs977511948 Chr.9: 130704524-130704524 on Build GRCh38
rs1017176760 Chr.9: 130704488-130704488 on Build GRCh38
rs971445338 Chr.9: 130704516-130704516 on Build GRCh38
rs867616785 Chr.9: 130704626-130704626 on Build GRCh38
rs144279301 Chr.9: 130704439-130704439 on Build GRCh38
rs1025731338 Chr.9: 130704463-130704463 on Build GRCh38
rs374343001 Chr.9: 130704454-130704454 on Build GRCh38
rs1139032 Chr.9: 130704446-130704446 on Build GRCh38
rs943035972 Chr.9: 130704429-130704429 on Build GRCh38
rs933523888 Chr.9: 130704179-130704179 on Build GRCh38
rs1017307420 Chr.9: 130704187-130704187 on Build GRCh38
rs950140031 Chr.9: 130704277-130704277 on Build GRCh38
rs906358562 Chr.9: 130704389-130704389 on Build GRCh38
rs887277579 Chr.9: 130704485-130704485 on Build GRCh38
rs147802511 Chr.9: 130704493-130704493 on Build GRCh38
rs140326492 Chr.9: 130704256-130704256 on Build GRCh38
rs1005713410 Chr.9: 130704486-130704486 on Build GRCh38
rs1139029 Chr.9: 130704388-130704388 on Build GRCh38
rs919251586 Chr.9: 130704552-130704552 on Build GRCh38
rs910826706 Chr.9: 130704255-130704255 on Build GRCh38
rs990417280 Chr.9: 130704586-130704586 on Build GRCh38
rs1046559548 Chr.9: 130704321-130704321 on Build GRCh38
rs1061967 Chr.9: 130704571-130704571 on Build GRCh38
rs1002068791 Chr.9: 130704410-130704410 on Build GRCh38
rs957190574 Chr.9: 130704584-130704584 on Build GRCh38
rs1038076375 Chr.9: 130704264-130704264 on Build GRCh38
rs1012902877 Chr.9: 130704505-130704505 on Build GRCh38
rs901578204 Chr.9: 130704443-130704443 on Build GRCh38
rs973682097 Chr.9: 130704534-130704534 on Build GRCh38
rs928059464 Chr.9: 130704611-130704611 on Build GRCh38
rs963928693 Chr.9: 130704533-130704534 on Build GRCh38
rs561920287 Chr.9: 130704462-130704462 on Build GRCh38
rs193107609 Chr.9: 130704258-130704258 on Build GRCh38
rs924401619 Chr.9: 130704534-130704534 on Build GRCh38
rs550622402 Chr.9: 130704501-130704501 on Build GRCh38
rs564235845 Chr.9: 130704548-130704548 on Build GRCh38

View Full Product Details