Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59794331-59794734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC6
Entrez Gene ID
8030
Gene Name
coiled-coil domain containing 6
Gene Aliases
D10S170, H4, PTC, TPC, TST1
UniGene
Hs.591360
Gene Chromosome Location
Chr.10: 59788747-59906656 on Build GRCh38
Transcripts
NM_005436.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7099025 c.1227C>G p.I409M Chr.10: 59794476-59794476 on Build GRCh38
4922142 c.1219C>A p.H407N Chr.10: 59794484-59794484 on Build GRCh38
1247536 c.1149T>G p.T383T Chr.10: 59794554-59794554 on Build GRCh38
4014910 c.1166G>A p.G389E Chr.10: 59794537-59794537 on Build GRCh38
5721526 c.1217C>T p.S406F Chr.10: 59794486-59794486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774749017 Chr.10: 59794490-59794490 on Build GRCh38
rs370071074 Chr.10: 59794516-59794516 on Build GRCh38
rs757020898 Chr.10: 59794457-59794457 on Build GRCh38
rs759165857 Chr.10: 59794522-59794522 on Build GRCh38
rs148666580 Chr.10: 59794705-59794705 on Build GRCh38
rs759880044 Chr.10: 59794675-59794675 on Build GRCh38
rs370532339 Chr.10: 59794447-59794447 on Build GRCh38
rs781095777 Chr.10: 59794582-59794582 on Build GRCh38
rs577220704 Chr.10: 59794420-59794420 on Build GRCh38
rs773052682 Chr.10: 59794615-59794615 on Build GRCh38
rs965250870 Chr.10: 59794680-59794680 on Build GRCh38
rs767992631 Chr.10: 59794525-59794525 on Build GRCh38
rs757438905 Chr.10: 59794563-59794563 on Build GRCh38
rs201036996 Chr.10: 59794438-59794438 on Build GRCh38
rs759097104 Chr.10: 59794453-59794453 on Build GRCh38
rs777968794 Chr.10: 59794477-59794477 on Build GRCh38
rs762827860 Chr.10: 59794629-59794629 on Build GRCh38
rs563630486 Chr.10: 59794656-59794656 on Build GRCh38
rs752992147 Chr.10: 59794531-59794531 on Build GRCh38
rs1045381539 Chr.10: 59794463-59794463 on Build GRCh38
rs114960441 Chr.10: 59794397-59794397 on Build GRCh38
rs574676584 Chr.10: 59794483-59794483 on Build GRCh38
rs767350114 Chr.10: 59794524-59794524 on Build GRCh38
rs948288544 Chr.10: 59794439-59794439 on Build GRCh38
rs761874280 Chr.10: 59794506-59794506 on Build GRCh38
rs765068975 Chr.10: 59794515-59794515 on Build GRCh38
rs746128939 Chr.10: 59794573-59794573 on Build GRCh38
rs768625365 Chr.10: 59794494-59794494 on Build GRCh38
rs545738565 Chr.10: 59794693-59794693 on Build GRCh38
rs908490417 Chr.10: 59794663-59794663 on Build GRCh38
rs751144475 Chr.10: 59794436-59794436 on Build GRCh38
rs374602904 Chr.10: 59794517-59794517 on Build GRCh38
rs756574614 Chr.10: 59794462-59794462 on Build GRCh38
rs770476359 Chr.10: 59794648-59794648 on Build GRCh38
rs779118114 Chr.10: 59794570-59794570 on Build GRCh38
rs374596318 Chr.10: 59794474-59794474 on Build GRCh38
rs368599035 Chr.10: 59794556-59794556 on Build GRCh38
rs376234238 Chr.10: 59794505-59794505 on Build GRCh38
rs984063972 Chr.10: 59794654-59794654 on Build GRCh38
rs201991808 Chr.10: 59794423-59794423 on Build GRCh38
rs781486348 Chr.10: 59794441-59794441 on Build GRCh38
rs754359397 Chr.10: 59794562-59794562 on Build GRCh38
rs1056918865 Chr.10: 59794689-59794689 on Build GRCh38
rs531120606 Chr.10: 59794676-59794676 on Build GRCh38
rs372072596 Chr.10: 59794575-59794575 on Build GRCh38
rs1025620601 Chr.10: 59794691-59794691 on Build GRCh38
rs747906759 Chr.10: 59794586-59794586 on Build GRCh38
rs141402057 Chr.10: 59794550-59794550 on Build GRCh38
rs769564269 Chr.10: 59794595-59794595 on Build GRCh38
rs185704771 Chr.10: 59794356-59794356 on Build GRCh38
rs766031886 Chr.10: 59794434-59794434 on Build GRCh38
rs918695659 Chr.10: 59794398-59794398 on Build GRCh38
rs965387280 Chr.10: 59794366-59794366 on Build GRCh38
rs1006674870 Chr.10: 59794659-59794659 on Build GRCh38
rs977181125 Chr.10: 59794690-59794690 on Build GRCh38
rs1012851039 Chr.10: 59794364-59794364 on Build GRCh38
rs749343006 Chr.10: 59794482-59794482 on Build GRCh38
rs758101437 Chr.10: 59794430-59794430 on Build GRCh38
rs756445063 Chr.10: 59794534-59794534 on Build GRCh38
rs904323558 Chr.10: 59794626-59794626 on Build GRCh38

View Full Product Details