Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 60026220-60026725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK3
Entrez Gene ID
288
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases
ANKYRIN-G, MRT37
UniGene
Hs.499725
Gene Chromosome Location
Chr.10: 60026298-60733526 on Build GRCh38
Transcripts
NM_001149.3 NM_001204403.1 NM_001204404.1 NM_001320874.1 NM_020987.4

SNP Detail

SNP ID Chromosome Location
rs1015185163 Chr.10: 60026477-60026478 on Build GRCh38
rs1046585 Chr.10: 60026622-60026622 on Build GRCh38
rs1053037383 Chr.10: 60026319-60026319 on Build GRCh38
rs1047832744 Chr.10: 60026694-60026694 on Build GRCh38
rs900286826 Chr.10: 60026641-60026641 on Build GRCh38
rs113696354 Chr.10: 60026553-60026553 on Build GRCh38
rs186700771 Chr.10: 60026328-60026328 on Build GRCh38
rs978912684 Chr.10: 60026530-60026530 on Build GRCh38
rs970610438 Chr.10: 60026481-60026481 on Build GRCh38
rs1040151549 Chr.10: 60026332-60026332 on Build GRCh38
rs567301078 Chr.10: 60026601-60026601 on Build GRCh38
rs1006873739 Chr.10: 60026428-60026428 on Build GRCh38
rs1007411448 Chr.10: 60026313-60026313 on Build GRCh38
rs764647934 Chr.10: 60026398-60026398 on Build GRCh38
rs913545133 Chr.10: 60026560-60026560 on Build GRCh38
rs952103937 Chr.10: 60026422-60026422 on Build GRCh38
rs935552815 Chr.10: 60026253-60026253 on Build GRCh38
rs1023232399 Chr.10: 60026465-60026465 on Build GRCh38
rs951949858 Chr.10: 60026393-60026393 on Build GRCh38
rs1027315712 Chr.10: 60026361-60026361 on Build GRCh38
rs548680429 Chr.10: 60026571-60026571 on Build GRCh38
rs113508665 Chr.10: 60026597-60026597 on Build GRCh38
rs528393614 Chr.10: 60026607-60026607 on Build GRCh38
rs946416448 Chr.10: 60026574-60026574 on Build GRCh38
rs929375439 Chr.10: 60026663-60026663 on Build GRCh38
rs760893450 Chr.10: 60026347-60026347 on Build GRCh38
rs992384006 Chr.10: 60026549-60026549 on Build GRCh38
rs1043676143 Chr.10: 60026276-60026276 on Build GRCh38
rs533516304 Chr.10: 60026353-60026353 on Build GRCh38
rs34100743 Chr.10: 60026278-60026279 on Build GRCh38
rs777123123 Chr.10: 60026534-60026534 on Build GRCh38
rs191408649 Chr.10: 60026556-60026556 on Build GRCh38
rs935605266 Chr.10: 60026562-60026562 on Build GRCh38
rs552345212 Chr.10: 60026348-60026348 on Build GRCh38
rs993575131 Chr.10: 60026356-60026356 on Build GRCh38
rs78682113 Chr.10: 60026605-60026605 on Build GRCh38
rs888917738 Chr.10: 60026308-60026308 on Build GRCh38
rs1004762297 Chr.10: 60026420-60026420 on Build GRCh38
rs567134013 Chr.10: 60026555-60026555 on Build GRCh38
rs1048760738 Chr.10: 60026667-60026668 on Build GRCh38
rs183617969 Chr.10: 60026631-60026631 on Build GRCh38
rs946605686 Chr.10: 60026256-60026256 on Build GRCh38
rs114693358 Chr.10: 60026640-60026640 on Build GRCh38
rs74777864 Chr.10: 60026633-60026633 on Build GRCh38
rs1040395299 Chr.10: 60026320-60026320 on Build GRCh38
rs925574817 Chr.10: 60026251-60026251 on Build GRCh38

View Full Product Details