Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 53944622-53945062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS1
Entrez Gene ID
8537
Gene Name
breast carcinoma amplified sequence 1
Gene Aliases
AIBC1, NABC1
UniGene
Hs.400556
Gene Chromosome Location
Chr.20: 53943538-54070765 on Build GRCh38
Transcripts
NM_001316361.2 NM_001323347.1 NM_003657.3 XM_005260587.2 XM_005260589.2 XM_005260590.2 XM_005260591.2 XM_005260593.2 XM_005260594.2 XM_005260595.2 XM_011529090.2 XM_011529091.2 XM_017028110.1 XM_017028111.1 XM_017028112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4001946 c.1774T>C p.S592P Chr.20: 53944930-53944930 on Build GRCh38
6093890 c.1690C>T p.R564W Chr.20: 53944987-53944987 on Build GRCh38
1130600 c.1747T>C p.S583P Chr.20: 53944930-53944930 on Build GRCh38
6093891 c.1717C>T p.R573W Chr.20: 53944987-53944987 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746457888 Chr.20: 53944702-53944702 on Build GRCh38
rs778740639 Chr.20: 53944821-53944821 on Build GRCh38
rs943883120 Chr.20: 53944971-53944971 on Build GRCh38
rs776341797 Chr.20: 53945019-53945019 on Build GRCh38
rs368207025 Chr.20: 53944936-53944936 on Build GRCh38
rs762588091 Chr.20: 53944973-53944973 on Build GRCh38
rs2072241 Chr.20: 53944833-53944833 on Build GRCh38
rs559865139 Chr.20: 53944955-53944955 on Build GRCh38
rs1010437354 Chr.20: 53944989-53944989 on Build GRCh38
rs1008588237 Chr.20: 53944878-53944878 on Build GRCh38
rs746952233 Chr.20: 53945015-53945015 on Build GRCh38
rs771840885 Chr.20: 53944679-53944679 on Build GRCh38
rs375738750 Chr.20: 53945028-53945028 on Build GRCh38
rs752786931 Chr.20: 53944980-53944980 on Build GRCh38
rs772037629 Chr.20: 53944939-53944939 on Build GRCh38
rs780538789 Chr.20: 53944891-53944891 on Build GRCh38
rs954427733 Chr.20: 53944846-53944846 on Build GRCh38
rs748324471 Chr.20: 53944910-53944910 on Build GRCh38
rs749290407 Chr.20: 53944654-53944654 on Build GRCh38
rs769291256 Chr.20: 53945027-53945027 on Build GRCh38
rs184043231 Chr.20: 53944809-53944809 on Build GRCh38
rs749945110 Chr.20: 53944959-53944959 on Build GRCh38
rs373855332 Chr.20: 53944946-53944946 on Build GRCh38
rs777281469 Chr.20: 53945038-53945039 on Build GRCh38
rs919949694 Chr.20: 53944905-53944905 on Build GRCh38
rs532384806 Chr.20: 53944850-53944850 on Build GRCh38
rs375524459 Chr.20: 53945001-53945001 on Build GRCh38
rs368523547 Chr.20: 53944997-53944997 on Build GRCh38
rs886703828 Chr.20: 53944773-53944773 on Build GRCh38
rs553444640 Chr.20: 53944810-53944810 on Build GRCh38
rs756730419 Chr.20: 53944876-53944876 on Build GRCh38
rs535958023 Chr.20: 53944842-53944842 on Build GRCh38
rs755331001 Chr.20: 53944986-53944986 on Build GRCh38
rs979047083 Chr.20: 53944924-53944924 on Build GRCh38
rs770441070 Chr.20: 53944753-53944753 on Build GRCh38
rs745553150 Chr.20: 53944669-53944669 on Build GRCh38
rs1033822546 Chr.20: 53944816-53944816 on Build GRCh38
rs760410242 Chr.20: 53944945-53944945 on Build GRCh38
rs764655855 Chr.20: 53944880-53944880 on Build GRCh38
rs370759863 Chr.20: 53944942-53944942 on Build GRCh38
rs761433334 Chr.20: 53944951-53944951 on Build GRCh38
rs770834027 Chr.20: 53944785-53944785 on Build GRCh38
rs552320640 Chr.20: 53944909-53944909 on Build GRCh38
rs777771344 Chr.20: 53944923-53944923 on Build GRCh38
rs916655868 Chr.20: 53944724-53944724 on Build GRCh38
rs766944888 Chr.20: 53944974-53944974 on Build GRCh38
rs766838819 Chr.20: 53944895-53944895 on Build GRCh38
rs764785129 Chr.20: 53944953-53944953 on Build GRCh38
rs775209235 Chr.20: 53944681-53944681 on Build GRCh38
rs777874134 Chr.20: 53944996-53944996 on Build GRCh38
rs891421881 Chr.20: 53944721-53944721 on Build GRCh38
rs867342468 Chr.20: 53945016-53945016 on Build GRCh38
rs756337172 Chr.20: 53944995-53944995 on Build GRCh38
rs768433227 Chr.20: 53945017-53945017 on Build GRCh38
rs1021751135 Chr.20: 53944772-53944772 on Build GRCh38
rs142044318 Chr.20: 53944844-53944844 on Build GRCh38
rs992207862 Chr.20: 53944834-53944834 on Build GRCh38
rs751937063 Chr.20: 53944899-53944899 on Build GRCh38
rs752947937 Chr.20: 53944992-53944992 on Build GRCh38
rs767657877 Chr.20: 53944960-53944960 on Build GRCh38
rs147771144 Chr.20: 53944987-53944987 on Build GRCh38
rs868713130 Chr.20: 53944925-53944925 on Build GRCh38
rs1055246 Chr.20: 53944930-53944930 on Build GRCh38
rs758881161 Chr.20: 53944887-53944887 on Build GRCh38
rs769910938 Chr.20: 53944922-53944922 on Build GRCh38
rs779908134 Chr.20: 53945014-53945014 on Build GRCh38
rs979711009 Chr.20: 53944804-53944804 on Build GRCh38
rs530547267 Chr.20: 53944754-53944754 on Build GRCh38
rs776277655 Chr.20: 53944947-53944947 on Build GRCh38
rs774758065 Chr.20: 53944881-53944881 on Build GRCh38
rs187785898 Chr.20: 53944840-53944840 on Build GRCh38
rs761496482 Chr.20: 53945020-53945020 on Build GRCh38

View Full Product Details