Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42102703-42103286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
-
UniGene
Hs.242520
Gene Chromosome Location
Chr.21: 42062959-42142996 on Build GRCh38
Transcripts
NM_001004416.2 NM_001199527.1 NM_001199528.2 NM_173568.3 XM_011529797.1 XM_017028506.1 XM_017028507.1 NR_027243.1

Gene Symbol
UMODL1-AS1
Entrez Gene ID
150147
Gene Name
UMODL1 antisense RNA 1
Gene Aliases
C21orf128
UniGene
Hs.558646
Gene Chromosome Location
Chr.21: 42102134-42108534 on Build GRCh38
Transcripts
NM_001004416.2 NM_001199527.1 NM_001199528.2 NM_173568.3 XM_011529797.1 XM_017028506.1 XM_017028507.1 NR_027243.1

SNP Detail

SNP ID Chromosome Location
rs879656958 Chr.21: 42103116-42103116 on Build GRCh38
rs535790123 Chr.21: 42102952-42102952 on Build GRCh38
rs190174928 Chr.21: 42102957-42102957 on Build GRCh38
rs945320894 Chr.21: 42103157-42103157 on Build GRCh38
rs113440127 Chr.21: 42102925-42102925 on Build GRCh38
rs933448696 Chr.21: 42102969-42102969 on Build GRCh38
rs760004919 Chr.21: 42102939-42102939 on Build GRCh38
rs550832244 Chr.21: 42102906-42102906 on Build GRCh38
rs764134826 Chr.21: 42103018-42103018 on Build GRCh38
rs924834696 Chr.21: 42102981-42102981 on Build GRCh38
rs906383438 Chr.21: 42103166-42103166 on Build GRCh38
rs888229155 Chr.21: 42103216-42103216 on Build GRCh38
rs753946571 Chr.21: 42103094-42103094 on Build GRCh38
rs914734579 Chr.21: 42102946-42102946 on Build GRCh38
rs760883886 Chr.21: 42103152-42103153 on Build GRCh38
rs888269235 Chr.21: 42103077-42103077 on Build GRCh38
rs551787740 Chr.21: 42102936-42102936 on Build GRCh38
rs1053821265 Chr.21: 42103080-42103080 on Build GRCh38
rs947636410 Chr.21: 42102773-42102773 on Build GRCh38
rs1056818844 Chr.21: 42102835-42102835 on Build GRCh38
rs1024811194 Chr.21: 42102898-42102898 on Build GRCh38
rs901049477 Chr.21: 42103182-42103182 on Build GRCh38
rs1021687600 Chr.21: 42103262-42103262 on Build GRCh38
rs539848162 Chr.21: 42103198-42103198 on Build GRCh38
rs774631866 Chr.21: 42102920-42102920 on Build GRCh38
rs570207067 Chr.21: 42102940-42102940 on Build GRCh38
rs35509362 Chr.21: 42102752-42102752 on Build GRCh38
rs1001921004 Chr.21: 42102914-42102914 on Build GRCh38
rs577269171 Chr.21: 42103195-42103195 on Build GRCh38
rs181934261 Chr.21: 42103187-42103187 on Build GRCh38
rs372413364 Chr.21: 42103208-42103208 on Build GRCh38
rs1030338385 Chr.21: 42102927-42102927 on Build GRCh38
rs539866658 Chr.21: 42102937-42102937 on Build GRCh38
rs915585614 Chr.21: 42102998-42102998 on Build GRCh38
rs765474200 Chr.21: 42103248-42103248 on Build GRCh38
rs938960837 Chr.21: 42102983-42102983 on Build GRCh38
rs544360229 Chr.21: 42103135-42103135 on Build GRCh38
rs543706047 Chr.21: 42103017-42103017 on Build GRCh38
rs770589553 Chr.21: 42102849-42102849 on Build GRCh38
rs761981051 Chr.21: 42103246-42103246 on Build GRCh38
rs914744636 Chr.21: 42102759-42102759 on Build GRCh38
rs147118215 Chr.21: 42102883-42102883 on Build GRCh38
rs986234756 Chr.21: 42103007-42103007 on Build GRCh38
rs868443780 Chr.21: 42103032-42103032 on Build GRCh38
rs952444737 Chr.21: 42102921-42102921 on Build GRCh38
rs116734140 Chr.21: 42102947-42102947 on Build GRCh38
rs1046715344 Chr.21: 42103160-42103160 on Build GRCh38
rs1041984953 Chr.21: 42102791-42102791 on Build GRCh38
rs1048208656 Chr.21: 42103049-42103049 on Build GRCh38
rs948451054 Chr.21: 42103023-42103024 on Build GRCh38
rs910663926 Chr.21: 42103046-42103046 on Build GRCh38
rs892412541 Chr.21: 42102869-42102869 on Build GRCh38
rs528717176 Chr.21: 42102731-42102731 on Build GRCh38
rs1057167763 Chr.21: 42102988-42102988 on Build GRCh38
rs575382637 Chr.21: 42103072-42103072 on Build GRCh38
rs761202960 Chr.21: 42102990-42102990 on Build GRCh38
rs938371694 Chr.21: 42102796-42102796 on Build GRCh38
rs141933154 Chr.21: 42102868-42102868 on Build GRCh38
rs1050468314 Chr.21: 42102795-42102795 on Build GRCh38
rs548461874 Chr.21: 42102848-42102848 on Build GRCh38
rs939809836 Chr.21: 42103132-42103132 on Build GRCh38
rs892460467 Chr.21: 42103137-42103137 on Build GRCh38
rs894188480 Chr.21: 42102840-42102840 on Build GRCh38
rs936727363 Chr.21: 42103050-42103050 on Build GRCh38

View Full Product Details