Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19951388-19951608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMT
Entrez Gene ID
1312
Gene Name
catechol-O-methyltransferase
Gene Aliases
HEL-S-98n
UniGene
Hs.370408
Gene Chromosome Location
Chr.22: 19941740-19969975 on Build GRCh38
Transcripts
NM_000754.3 NM_001135161.1 NM_001135162.1 XM_017028594.1 XM_017028595.1

SNP Detail

SNP ID Chromosome Location
rs942537433 Chr.22: 19951493-19951493 on Build GRCh38
rs903164787 Chr.22: 19951535-19951535 on Build GRCh38
rs912597401 Chr.22: 19951453-19951453 on Build GRCh38
rs146658977 Chr.22: 19951531-19951531 on Build GRCh38
rs144623972 Chr.22: 19951516-19951516 on Build GRCh38
rs1049387848 Chr.22: 19951528-19951528 on Build GRCh38
rs1041710428 Chr.22: 19951534-19951534 on Build GRCh38
rs191619546 Chr.22: 19951560-19951560 on Build GRCh38
rs140215652 Chr.22: 19951559-19951559 on Build GRCh38
rs933873038 Chr.22: 19951533-19951533 on Build GRCh38
rs188402271 Chr.22: 19951483-19951483 on Build GRCh38
rs1013824380 Chr.22: 19951552-19951552 on Build GRCh38
rs896191010 Chr.22: 19951541-19951541 on Build GRCh38
rs901097371 Chr.22: 19951523-19951523 on Build GRCh38
rs1033080742 Chr.22: 19951555-19951555 on Build GRCh38
rs972274850 Chr.22: 19951584-19951584 on Build GRCh38
rs1000255869 Chr.22: 19951539-19951539 on Build GRCh38
rs544882417 Chr.22: 19951469-19951469 on Build GRCh38
rs900331572 Chr.22: 19951508-19951508 on Build GRCh38
rs113706839 Chr.22: 19951580-19951580 on Build GRCh38
rs983865428 Chr.22: 19951470-19951470 on Build GRCh38
rs1023921815 Chr.22: 19951557-19951557 on Build GRCh38
rs739368 Chr.22: 19951573-19951573 on Build GRCh38
rs988670987 Chr.22: 19951583-19951583 on Build GRCh38
rs1040292129 Chr.22: 19951496-19951496 on Build GRCh38

View Full Product Details