Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113387070-113387659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALAD
Entrez Gene ID
210
Gene Name
aminolevulinate dehydratase
Gene Aliases
ALADH, PBGS
UniGene
Hs.1227
Gene Chromosome Location
Chr.9: 113386312-113401338 on Build GRCh38
Transcripts
NM_000031.5 NM_001003945.2 NM_001317745.1 XM_011518364.2

SNP Detail

SNP ID Chromosome Location
rs8177822 Chr.9: 113387429-113387429 on Build GRCh38
rs959638785 Chr.9: 113387118-113387118 on Build GRCh38
rs984280624 Chr.9: 113387595-113387595 on Build GRCh38
rs990809416 Chr.9: 113387191-113387191 on Build GRCh38
rs1044749757 Chr.9: 113387371-113387371 on Build GRCh38
rs919413434 Chr.9: 113387249-113387249 on Build GRCh38
rs927597840 Chr.9: 113387628-113387628 on Build GRCh38
rs532401659 Chr.9: 113387623-113387624 on Build GRCh38
rs878996496 Chr.9: 113387411-113387411 on Build GRCh38
rs1052723290 Chr.9: 113387457-113387457 on Build GRCh38
rs942162529 Chr.9: 113387520-113387520 on Build GRCh38
rs530554500 Chr.9: 113387196-113387196 on Build GRCh38
rs8177824 Chr.9: 113387205-113387205 on Build GRCh38
rs36118229 Chr.9: 113387302-113387303 on Build GRCh38
rs1021139853 Chr.9: 113387377-113387377 on Build GRCh38
rs8177823 Chr.9: 113387293-113387293 on Build GRCh38
rs1003418996 Chr.9: 113387109-113387109 on Build GRCh38
rs541967139 Chr.9: 113387547-113387547 on Build GRCh38
rs952213474 Chr.9: 113387587-113387587 on Build GRCh38
rs972669661 Chr.9: 113387244-113387244 on Build GRCh38
rs138705750 Chr.9: 113387487-113387487 on Build GRCh38
rs1022729855 Chr.9: 113387162-113387162 on Build GRCh38
rs530839570 Chr.9: 113387517-113387517 on Build GRCh38
rs553095457 Chr.9: 113387156-113387156 on Build GRCh38
rs543806069 Chr.9: 113387496-113387496 on Build GRCh38
rs375648415 Chr.9: 113387262-113387262 on Build GRCh38
rs959606457 Chr.9: 113387447-113387447 on Build GRCh38
rs756786389 Chr.9: 113387599-113387599 on Build GRCh38
rs1048292844 Chr.9: 113387203-113387203 on Build GRCh38
rs925505683 Chr.9: 113387101-113387101 on Build GRCh38
rs1026912532 Chr.9: 113387586-113387586 on Build GRCh38
rs528302155 Chr.9: 113387462-113387462 on Build GRCh38
rs939833341 Chr.9: 113387222-113387222 on Build GRCh38
rs368158957 Chr.9: 113387358-113387358 on Build GRCh38
rs561342837 Chr.9: 113387284-113387284 on Build GRCh38
rs980463952 Chr.9: 113387600-113387600 on Build GRCh38
rs191395992 Chr.9: 113387594-113387594 on Build GRCh38
rs117612544 Chr.9: 113387338-113387338 on Build GRCh38
rs550472267 Chr.9: 113387229-113387229 on Build GRCh38
rs919762678 Chr.9: 113387597-113387597 on Build GRCh38
rs972316736 Chr.9: 113387541-113387541 on Build GRCh38
rs149384214 Chr.9: 113387529-113387529 on Build GRCh38
rs1011414484 Chr.9: 113387467-113387467 on Build GRCh38
rs11556127 Chr.9: 113387463-113387463 on Build GRCh38
rs1049346317 Chr.9: 113387323-113387323 on Build GRCh38
rs932536766 Chr.9: 113387613-113387613 on Build GRCh38
rs949465970 Chr.9: 113387199-113387199 on Build GRCh38
rs544531295 Chr.9: 113387269-113387269 on Build GRCh38
rs559328718 Chr.9: 113387240-113387240 on Build GRCh38
rs1010974505 Chr.9: 113387161-113387161 on Build GRCh38
rs979656783 Chr.9: 113387100-113387100 on Build GRCh38
rs1007949060 Chr.9: 113387365-113387365 on Build GRCh38
rs902282749 Chr.9: 113387386-113387386 on Build GRCh38
rs187307200 Chr.9: 113387558-113387558 on Build GRCh38
rs1032544289 Chr.9: 113387433-113387433 on Build GRCh38
rs1024948827 Chr.9: 113387468-113387468 on Build GRCh38
rs542817534 Chr.9: 113387243-113387243 on Build GRCh38
rs957037928 Chr.9: 113387120-113387120 on Build GRCh38
rs573525746 Chr.9: 113387171-113387171 on Build GRCh38
rs370842277 Chr.9: 113387316-113387316 on Build GRCh38
rs890793250 Chr.9: 113387350-113387350 on Build GRCh38
rs560189518 Chr.9: 113387444-113387444 on Build GRCh38
rs765493168 Chr.9: 113387275-113387275 on Build GRCh38

View Full Product Details