Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41964552-41965060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC25
Entrez Gene ID
83538
Gene Name
tetratricopeptide repeat domain 25
Gene Aliases
-
UniGene
Hs.201134
Gene Chromosome Location
Chr.17: 41930605-41966517 on Build GRCh38
Transcripts
NM_031421.3 NR_110662.1 XM_011525323.2 XM_011525324.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4627530 c.1533T>C p.A511A Chr.17: 41964997-41964997 on Build GRCh38
6315909 c.1539G>T p.L513L Chr.17: 41965003-41965003 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs116764089 Chr.17: 41964953-41964953 on Build GRCh38
rs540044830 Chr.17: 41964849-41964849 on Build GRCh38
rs564085642 Chr.17: 41964892-41964892 on Build GRCh38
rs1029800882 Chr.17: 41964628-41964628 on Build GRCh38
rs116259554 Chr.17: 41964594-41964594 on Build GRCh38
rs987197990 Chr.17: 41964708-41964708 on Build GRCh38
rs782105223 Chr.17: 41965001-41965001 on Build GRCh38
rs891416237 Chr.17: 41964973-41964973 on Build GRCh38
rs1030328441 Chr.17: 41964968-41964968 on Build GRCh38
rs949927899 Chr.17: 41964847-41964847 on Build GRCh38
rs1010516773 Chr.17: 41964625-41964625 on Build GRCh38
rs113583076 Chr.17: 41964768-41964768 on Build GRCh38
rs8080979 Chr.17: 41964801-41964801 on Build GRCh38
rs548838316 Chr.17: 41964938-41964938 on Build GRCh38
rs891820225 Chr.17: 41964949-41964949 on Build GRCh38
rs115448773 Chr.17: 41964615-41964615 on Build GRCh38
rs576960347 Chr.17: 41964834-41964834 on Build GRCh38
rs973027891 Chr.17: 41964767-41964767 on Build GRCh38
rs955736879 Chr.17: 41964675-41964675 on Build GRCh38
rs1018471886 Chr.17: 41965008-41965008 on Build GRCh38
rs530435098 Chr.17: 41964904-41964904 on Build GRCh38
rs781992024 Chr.17: 41964972-41964972 on Build GRCh38
rs782366923 Chr.17: 41965034-41965034 on Build GRCh38
rs782723414 Chr.17: 41965028-41965028 on Build GRCh38
rs964462284 Chr.17: 41964750-41964750 on Build GRCh38
rs1017768608 Chr.17: 41964747-41964747 on Build GRCh38
rs8080978 Chr.17: 41964799-41964799 on Build GRCh38
rs1044883577 Chr.17: 41964581-41964581 on Build GRCh38
rs386797138 Chr.17: 41964799-41964802 on Build GRCh38
rs782258512 Chr.17: 41964946-41964946 on Build GRCh38
rs188184469 Chr.17: 41964869-41964869 on Build GRCh38
rs1007460964 Chr.17: 41965004-41965004 on Build GRCh38
rs576141603 Chr.17: 41964788-41964788 on Build GRCh38
rs936100149 Chr.17: 41964895-41964895 on Build GRCh38
rs115916336 Chr.17: 41964593-41964593 on Build GRCh38
rs536511976 Chr.17: 41964717-41964717 on Build GRCh38
rs374928197 Chr.17: 41964746-41964746 on Build GRCh38
rs199986410 Chr.17: 41964944-41964944 on Build GRCh38
rs557814841 Chr.17: 41964601-41964601 on Build GRCh38

View Full Product Details