Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31764176-31764682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSH5-SAPCD1
Entrez Gene ID
100532732
Gene Name
MSH5-SAPCD1 readthrough (NMD candidate)
Gene Aliases
MSH5-C6orf26
UniGene
Hs.647011
Gene Chromosome Location
Chr.6: 31739948-31764850 on Build GRCh38
Transcripts
NR_037846.1 NM_001039651.1 NR_126423.1 XM_005249427.2 XM_017011327.1 XM_017011328.1 XM_017011329.1

Gene Symbol
SAPCD1
Entrez Gene ID
401251
Gene Name
suppressor APC domain containing 1
Gene Aliases
C6orf26, NG23
UniGene
Hs.371225
Gene Chromosome Location
Chr.6: 31762996-31764850 on Build GRCh38
Transcripts
NR_037846.1 NM_001039651.1 NR_126423.1 XM_005249427.2 XM_017011327.1 XM_017011328.1 XM_017011329.1

Gene Symbol
SAPCD1-AS1
Entrez Gene ID
104413891
Gene Name
SAPCD1 antisense RNA 1
Gene Aliases
C6orf26-AS1
UniGene
Hs.596130
Gene Chromosome Location
Chr.6: 31764135-31765588 on Build GRCh38
Transcripts
NR_037846.1 NM_001039651.1 NR_126423.1 XM_005249427.2 XM_017011327.1 XM_017011328.1 XM_017011329.1

Gene Symbol
VWA7
Entrez Gene ID
80737
Gene Name
von Willebrand factor A domain containing 7
Gene Aliases
C6orf27, G7c, NG37
UniGene
Hs.558553
Gene Chromosome Location
Chr.6: 31764135-31777331 on Build GRCh38
Transcripts
NR_037846.1 NM_001039651.1 NR_126423.1 XM_005249427.2 XM_017011327.1 XM_017011328.1 XM_017011329.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7294833 c.442-4C>T p.? Chr.6: 31764432-31764432 on Build GRCh38
5904836 c.354C>A p.F118L Chr.6: 31764268-31764268 on Build GRCh38
6383070 c.417G>T p.R139S Chr.6: 31764331-31764331 on Build GRCh38
6352161 c.442-6C>T p.? Chr.6: 31764430-31764430 on Build GRCh38
3624745 c.392C>T p.T131I Chr.6: 31764306-31764306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778943820 Chr.6: 31764385-31764385 on Build GRCh38
rs769094704 Chr.6: 31764578-31764578 on Build GRCh38
rs781182738 Chr.6: 31764280-31764280 on Build GRCh38
rs879668106 Chr.6: 31764569-31764569 on Build GRCh38
rs759156782 Chr.6: 31764437-31764437 on Build GRCh38
rs904596253 Chr.6: 31764648-31764648 on Build GRCh38
rs750014882 Chr.6: 31764221-31764221 on Build GRCh38
rs771918178 Chr.6: 31764399-31764399 on Build GRCh38
rs756212935 Chr.6: 31764505-31764505 on Build GRCh38
rs536501982 Chr.6: 31764632-31764632 on Build GRCh38
rs200395234 Chr.6: 31764418-31764418 on Build GRCh38
rs968740255 Chr.6: 31764542-31764542 on Build GRCh38
rs750607022 Chr.6: 31764492-31764492 on Build GRCh38
rs377446358 Chr.6: 31764372-31764372 on Build GRCh38
rs767252281 Chr.6: 31764220-31764220 on Build GRCh38
rs752542251 Chr.6: 31764365-31764367 on Build GRCh38
rs774136483 Chr.6: 31764300-31764300 on Build GRCh38
rs758666480 Chr.6: 31764263-31764263 on Build GRCh38
rs566897913 Chr.6: 31764336-31764336 on Build GRCh38
rs377628564 Chr.6: 31764602-31764606 on Build GRCh38
rs753111031 Chr.6: 31764345-31764345 on Build GRCh38
rs373118235 Chr.6: 31764307-31764307 on Build GRCh38
rs147916387 Chr.6: 31764450-31764450 on Build GRCh38
rs767770200 Chr.6: 31764491-31764491 on Build GRCh38
rs145384116 Chr.6: 31764526-31764526 on Build GRCh38
rs776344179 Chr.6: 31764326-31764326 on Build GRCh38
rs200117609 Chr.6: 31764429-31764429 on Build GRCh38
rs752680166 Chr.6: 31764516-31764516 on Build GRCh38
rs758331169 Chr.6: 31764522-31764522 on Build GRCh38
rs144721648 Chr.6: 31764447-31764447 on Build GRCh38
rs753160424 Chr.6: 31764251-31764251 on Build GRCh38
rs763207569 Chr.6: 31764434-31764436 on Build GRCh38
rs779944449 Chr.6: 31764509-31764509 on Build GRCh38
rs946532364 Chr.6: 31764627-31764627 on Build GRCh38
rs190883131 Chr.6: 31764250-31764250 on Build GRCh38
rs370709123 Chr.6: 31764273-31764273 on Build GRCh38
rs757333591 Chr.6: 31764533-31764533 on Build GRCh38
rs774930464 Chr.6: 31764556-31764556 on Build GRCh38
rs765986738 Chr.6: 31764321-31764321 on Build GRCh38
rs758821446 Chr.6: 31764350-31764350 on Build GRCh38
rs757349724 Chr.6: 31764275-31764275 on Build GRCh38
rs954167053 Chr.6: 31764410-31764410 on Build GRCh38
rs531176306 Chr.6: 31764368-31764368 on Build GRCh38
rs746791926 Chr.6: 31764427-31764427 on Build GRCh38
rs28399992 Chr.6: 31764282-31764282 on Build GRCh38
rs748018541 Chr.6: 31764305-31764305 on Build GRCh38
rs28399994 Chr.6: 31764524-31764524 on Build GRCh38
rs148653160 Chr.6: 31764320-31764320 on Build GRCh38
rs751900152 Chr.6: 31764520-31764531 on Build GRCh38
rs762144336 Chr.6: 31764490-31764490 on Build GRCh38
rs776398502 Chr.6: 31764436-31764436 on Build GRCh38
rs745468681 Chr.6: 31764544-31764544 on Build GRCh38
rs772812144 Chr.6: 31764423-31764423 on Build GRCh38
rs965615950 Chr.6: 31764325-31764325 on Build GRCh38
rs764657529 Chr.6: 31764468-31764468 on Build GRCh38
rs764401382 Chr.6: 31764483-31764484 on Build GRCh38
rs761501090 Chr.6: 31764210-31764210 on Build GRCh38
rs28399993 Chr.6: 31764378-31764378 on Build GRCh38
rs755465971 Chr.6: 31764236-31764236 on Build GRCh38
rs772942924 Chr.6: 31764311-31764311 on Build GRCh38
rs781021264 Chr.6: 31764528-31764528 on Build GRCh38
rs911424632 Chr.6: 31764208-31764208 on Build GRCh38
rs887430305 Chr.6: 31764327-31764327 on Build GRCh38
rs759504678 Chr.6: 31764352-31764352 on Build GRCh38
rs774810919 Chr.6: 31764580-31764580 on Build GRCh38
rs370349817 Chr.6: 31764389-31764389 on Build GRCh38
rs976900488 Chr.6: 31764641-31764641 on Build GRCh38
rs541253218 Chr.6: 31764599-31764603 on Build GRCh38
rs927693012 Chr.6: 31764508-31764508 on Build GRCh38
rs745997439 Chr.6: 31764379-31764379 on Build GRCh38
rs762511877 Chr.6: 31764560-31764560 on Build GRCh38
rs764610831 Chr.6: 31764440-31764440 on Build GRCh38
rs769360021 Chr.6: 31764546-31764546 on Build GRCh38
rs751979076 Chr.6: 31764475-31764475 on Build GRCh38
rs754406304 Chr.6: 31764234-31764234 on Build GRCh38
rs371396054 Chr.6: 31764366-31764366 on Build GRCh38
rs765038358 Chr.6: 31764244-31764245 on Build GRCh38
rs9461719 Chr.6: 31764430-31764430 on Build GRCh38
rs768640027 Chr.6: 31764295-31764295 on Build GRCh38
rs547799530 Chr.6: 31764226-31764226 on Build GRCh38
rs570215398 Chr.6: 31764340-31764340 on Build GRCh38
rs1043593644 Chr.6: 31764631-31764631 on Build GRCh38
rs747991080 Chr.6: 31764391-31764391 on Build GRCh38
rs777749400 Chr.6: 31764523-31764523 on Build GRCh38
rs376320701 Chr.6: 31764382-31764382 on Build GRCh38

View Full Product Details