Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31897549-31898075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2
Entrez Gene ID
717
Gene Name
complement component 2
Gene Aliases
ARMD14, CO2
UniGene
Hs.408903
Gene Chromosome Location
Chr.6: 31897785-31945674 on Build GRCh38
Transcripts
NM_001282457.1 NM_006709.4 NM_025256.6 XM_006714976.1 XM_017010202.1

Gene Symbol
EHMT2
Entrez Gene ID
10919
Gene Name
euchromatic histone lysine methyltransferase 2
Gene Aliases
BAT8, C6orf30, G9A, GAT8, KMT1C, NG36
UniGene
Hs.709218
Gene Chromosome Location
Chr.6: 31879759-31897707 on Build GRCh38
Transcripts
NM_001282457.1 NM_006709.4 NM_025256.6 XM_006714976.1 XM_017010202.1

SNP Detail

SNP ID Chromosome Location
rs1048439058 Chr.6: 31897740-31897740 on Build GRCh38
rs771142488 Chr.6: 31897697-31897697 on Build GRCh38
rs540227717 Chr.6: 31897944-31897944 on Build GRCh38
rs1044231646 Chr.6: 31897774-31897774 on Build GRCh38
rs888540168 Chr.6: 31897741-31897741 on Build GRCh38
rs1034390239 Chr.6: 31897634-31897634 on Build GRCh38
rs1026703155 Chr.6: 31898049-31898049 on Build GRCh38
rs112685574 Chr.6: 31897584-31897584 on Build GRCh38
rs969978885 Chr.6: 31897645-31897645 on Build GRCh38
rs985612316 Chr.6: 31897686-31897686 on Build GRCh38
rs1002064888 Chr.6: 31897623-31897623 on Build GRCh38
rs1008841485 Chr.6: 31897953-31897953 on Build GRCh38
rs1046223333 Chr.6: 31897777-31897777 on Build GRCh38
rs911385018 Chr.6: 31897970-31897970 on Build GRCh38
rs1010331105 Chr.6: 31897919-31897919 on Build GRCh38
rs1036055805 Chr.6: 31897810-31897810 on Build GRCh38
rs929967975 Chr.6: 31897716-31897716 on Build GRCh38
rs1019776159 Chr.6: 31897955-31897955 on Build GRCh38
rs981379318 Chr.6: 31897646-31897646 on Build GRCh38
rs964184887 Chr.6: 31897980-31897980 on Build GRCh38
rs762043023 Chr.6: 31897892-31897892 on Build GRCh38
rs1035341160 Chr.6: 31897648-31897648 on Build GRCh38
rs528617300 Chr.6: 31897907-31897907 on Build GRCh38
rs1011541398 Chr.6: 31897831-31897831 on Build GRCh38
rs1001552850 Chr.6: 31897804-31897804 on Build GRCh38
rs893149827 Chr.6: 31897811-31897811 on Build GRCh38
rs911416432 Chr.6: 31897691-31897691 on Build GRCh38
rs1020957927 Chr.6: 31897860-31897860 on Build GRCh38
rs144725072 Chr.6: 31897575-31897575 on Build GRCh38
rs1021351167 Chr.6: 31897935-31897935 on Build GRCh38
rs372445727 Chr.6: 31897986-31897987 on Build GRCh38
rs947552406 Chr.6: 31897682-31897682 on Build GRCh38
rs1006032948 Chr.6: 31897751-31897751 on Build GRCh38
rs887577467 Chr.6: 31897618-31897618 on Build GRCh38
rs918595845 Chr.6: 31897703-31897703 on Build GRCh38
rs975174304 Chr.6: 31898007-31898007 on Build GRCh38
rs976934971 Chr.6: 31897982-31897982 on Build GRCh38
rs562147290 Chr.6: 31897975-31897975 on Build GRCh38
rs1006065468 Chr.6: 31897642-31897642 on Build GRCh38
rs564523323 Chr.6: 31897805-31897805 on Build GRCh38
rs914315381 Chr.6: 31897663-31897663 on Build GRCh38
rs988574842 Chr.6: 31897661-31897662 on Build GRCh38
rs907323892 Chr.6: 31897801-31897801 on Build GRCh38
rs965558572 Chr.6: 31897971-31897971 on Build GRCh38
rs944284007 Chr.6: 31897701-31897701 on Build GRCh38
rs955930547 Chr.6: 31897651-31897651 on Build GRCh38
rs1022857134 Chr.6: 31897642-31897643 on Build GRCh38
rs967652882 Chr.6: 31897882-31897882 on Build GRCh38
rs776850499 Chr.6: 31897816-31897816 on Build GRCh38
rs868600151 Chr.6: 31897654-31897654 on Build GRCh38

View Full Product Details