Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31962353-31962857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKIV2L
Entrez Gene ID
6499
Gene Name
Ski2 like RNA helicase
Gene Aliases
170A, DDX13, HLP, SKI2, SKI2W, SKIV2, SKIV2L1, THES2
UniGene
Hs.89864
Gene Chromosome Location
Chr.6: 31958804-31969852 on Build GRCh38
Transcripts
NM_006929.4 XM_011514815.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6779002 c.1219C>A p.L407M Chr.6: 31962721-31962721 on Build GRCh38
1242847 c.1288G>A p.D430N Chr.6: 31962790-31962790 on Build GRCh38
3774237 c.1200A>G p.T400T Chr.6: 31962574-31962574 on Build GRCh38
3624921 c.1263C>T p.I421I Chr.6: 31962765-31962765 on Build GRCh38
6359890 c.1192A>G p.M398V Chr.6: 31962566-31962566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746781902 Chr.6: 31962560-31962560 on Build GRCh38
rs775365309 Chr.6: 31962671-31962674 on Build GRCh38
rs767586610 Chr.6: 31962791-31962791 on Build GRCh38
rs377014444 Chr.6: 31962437-31962437 on Build GRCh38
rs749792251 Chr.6: 31962628-31962628 on Build GRCh38
rs429608 Chr.6: 31962685-31962685 on Build GRCh38
rs764335100 Chr.6: 31962519-31962519 on Build GRCh38
rs765802910 Chr.6: 31962424-31962424 on Build GRCh38
rs745488686 Chr.6: 31962687-31962687 on Build GRCh38
rs745433848 Chr.6: 31962584-31962584 on Build GRCh38
rs571683357 Chr.6: 31962771-31962771 on Build GRCh38
rs141521384 Chr.6: 31962789-31962789 on Build GRCh38
rs560639276 Chr.6: 31962485-31962485 on Build GRCh38
rs372532139 Chr.6: 31962404-31962404 on Build GRCh38
rs138507648 Chr.6: 31962745-31962745 on Build GRCh38
rs1049585914 Chr.6: 31962792-31962792 on Build GRCh38
rs754494988 Chr.6: 31962830-31962830 on Build GRCh38
rs769045713 Chr.6: 31962739-31962739 on Build GRCh38
rs778754784 Chr.6: 31962483-31962483 on Build GRCh38
rs781111669 Chr.6: 31962457-31962457 on Build GRCh38
rs758226042 Chr.6: 31962550-31962550 on Build GRCh38
rs776131482 Chr.6: 31962506-31962506 on Build GRCh38
rs763479284 Chr.6: 31962516-31962516 on Build GRCh38
rs560943100 Chr.6: 31962660-31962660 on Build GRCh38
rs527344190 Chr.6: 31962689-31962689 on Build GRCh38
rs776038879 Chr.6: 31962495-31962495 on Build GRCh38
rs150540870 Chr.6: 31962819-31962819 on Build GRCh38
rs757399245 Chr.6: 31962454-31962454 on Build GRCh38
rs565047981 Chr.6: 31962674-31962674 on Build GRCh38
rs776721859 Chr.6: 31962788-31962788 on Build GRCh38
rs757274140 Chr.6: 31962379-31962379 on Build GRCh38
rs762168219 Chr.6: 31962637-31962637 on Build GRCh38
rs184417779 Chr.6: 31962539-31962539 on Build GRCh38
rs748516211 Chr.6: 31962751-31962751 on Build GRCh38
rs751726656 Chr.6: 31962453-31962453 on Build GRCh38
rs750465566 Chr.6: 31962475-31962475 on Build GRCh38
rs527940427 Chr.6: 31962546-31962546 on Build GRCh38
rs143563985 Chr.6: 31962526-31962526 on Build GRCh38
rs760145588 Chr.6: 31962408-31962408 on Build GRCh38
rs541520263 Chr.6: 31962781-31962781 on Build GRCh38
rs200818962 Chr.6: 31962494-31962494 on Build GRCh38
rs745357048 Chr.6: 31962505-31962505 on Build GRCh38
rs192525991 Chr.6: 31962812-31962812 on Build GRCh38
rs776181710 Chr.6: 31962596-31962596 on Build GRCh38
rs750197196 Chr.6: 31962809-31962809 on Build GRCh38
rs766537887 Chr.6: 31962783-31962783 on Build GRCh38
rs764468835 Chr.6: 31962448-31962448 on Build GRCh38
rs777232949 Chr.6: 31962405-31962405 on Build GRCh38
rs536282176 Chr.6: 31962373-31962373 on Build GRCh38
rs748770264 Chr.6: 31962703-31962703 on Build GRCh38
rs559629113 Chr.6: 31962795-31962795 on Build GRCh38
rs769159730 Chr.6: 31962632-31962632 on Build GRCh38
rs371878469 Chr.6: 31962497-31962497 on Build GRCh38
rs757046244 Chr.6: 31962807-31962807 on Build GRCh38
rs886061307 Chr.6: 31962525-31962525 on Build GRCh38
rs770497113 Chr.6: 31962571-31962571 on Build GRCh38
rs140859335 Chr.6: 31962638-31962638 on Build GRCh38
rs592229 Chr.6: 31962664-31962664 on Build GRCh38
rs779736816 Chr.6: 31962699-31962699 on Build GRCh38
rs200616435 Chr.6: 31962595-31962595 on Build GRCh38
rs201001669 Chr.6: 31962382-31962382 on Build GRCh38
rs767401166 Chr.6: 31962808-31962808 on Build GRCh38
rs61761945 Chr.6: 31962547-31962547 on Build GRCh38
rs371905619 Chr.6: 31962763-31962763 on Build GRCh38
rs371576196 Chr.6: 31962663-31962663 on Build GRCh38
rs757062062 Chr.6: 31962675-31962675 on Build GRCh38
rs368604484 Chr.6: 31962401-31962401 on Build GRCh38
rs2734331 Chr.6: 31962574-31962574 on Build GRCh38
rs758288399 Chr.6: 31962673-31962673 on Build GRCh38
rs962471847 Chr.6: 31962387-31962387 on Build GRCh38
rs979974887 Chr.6: 31962611-31962611 on Build GRCh38
rs570298416 Chr.6: 31962429-31962429 on Build GRCh38
rs866962006 Chr.6: 31962565-31962565 on Build GRCh38
rs1056757953 Chr.6: 31962447-31962447 on Build GRCh38
rs11541399 Chr.6: 31962736-31962736 on Build GRCh38
rs763997095 Chr.6: 31962790-31962790 on Build GRCh38
rs767736835 Chr.6: 31962529-31962529 on Build GRCh38
rs779783967 Chr.6: 31962825-31962825 on Build GRCh38
rs771837264 Chr.6: 31962486-31962486 on Build GRCh38
rs769314491 Chr.6: 31962667-31962668 on Build GRCh38
rs761966719 Chr.6: 31962527-31962527 on Build GRCh38
rs773462911 Chr.6: 31962757-31962757 on Build GRCh38
rs754937769 Chr.6: 31962476-31962476 on Build GRCh38
rs369294396 Chr.6: 31962507-31962507 on Build GRCh38
rs142429431 Chr.6: 31962548-31962548 on Build GRCh38
rs143334743 Chr.6: 31962450-31962450 on Build GRCh38
rs776383284 Chr.6: 31962408-31962413 on Build GRCh38
rs962422508 Chr.6: 31962725-31962725 on Build GRCh38
rs752678664 Chr.6: 31962668-31962668 on Build GRCh38
rs746607453 Chr.6: 31962493-31962493 on Build GRCh38
rs542306778 Chr.6: 31962636-31962636 on Build GRCh38
rs768322046 Chr.6: 31962505-31962505 on Build GRCh38
rs137885846 Chr.6: 31962399-31962399 on Build GRCh38
rs759570592 Chr.6: 31962753-31962753 on Build GRCh38
rs777406095 Chr.6: 31962557-31962557 on Build GRCh38
rs781067999 Chr.6: 31962479-31962479 on Build GRCh38
rs945424075 Chr.6: 31962518-31962518 on Build GRCh38
rs775564479 Chr.6: 31962826-31962826 on Build GRCh38
rs899709021 Chr.6: 31962580-31962580 on Build GRCh38

View Full Product Details