Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35458215-35458642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCE
Entrez Gene ID
2178
Gene Name
Fanconi anemia complementation group E
Gene Aliases
FACE, FAE
UniGene
Hs.302003
Gene Chromosome Location
Chr.6: 35452339-35467106 on Build GRCh38
Transcripts
NM_021922.2 XM_005248885.2 XM_005248886.2 XM_005248887.2 XM_005248888.3 XM_011514343.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4458373 c.1082C>T p.T361I Chr.6: 35458409-35458409 on Build GRCh38
6277698 c.985G>T p.A329S Chr.6: 35458312-35458312 on Build GRCh38
3626004 c.1000C>T p.P334S Chr.6: 35458327-35458327 on Build GRCh38
6660464 c.1017C>T p.L339L Chr.6: 35458344-35458344 on Build GRCh38
1312127 c.981G>T p.L327L Chr.6: 35458308-35458308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776082827 Chr.6: 35458326-35458326 on Build GRCh38
rs764119752 Chr.6: 35458446-35458446 on Build GRCh38
rs45467798 Chr.6: 35458355-35458355 on Build GRCh38
rs766204656 Chr.6: 35458275-35458276 on Build GRCh38
rs759421142 Chr.6: 35458273-35458273 on Build GRCh38
rs756414021 Chr.6: 35458285-35458285 on Build GRCh38
rs377505032 Chr.6: 35458389-35458389 on Build GRCh38
rs117779586 Chr.6: 35458281-35458281 on Build GRCh38
rs918624212 Chr.6: 35458540-35458540 on Build GRCh38
rs370603843 Chr.6: 35458463-35458463 on Build GRCh38
rs922200232 Chr.6: 35458259-35458259 on Build GRCh38
rs74567654 Chr.6: 35458603-35458603 on Build GRCh38
rs765193137 Chr.6: 35458277-35458277 on Build GRCh38
rs920363780 Chr.6: 35458510-35458510 on Build GRCh38
rs894142377 Chr.6: 35458601-35458601 on Build GRCh38
rs758179752 Chr.6: 35458280-35458280 on Build GRCh38
rs972752758 Chr.6: 35458527-35458527 on Build GRCh38
rs878855070 Chr.6: 35458384-35458384 on Build GRCh38
rs910119002 Chr.6: 35458498-35458498 on Build GRCh38
rs755510065 Chr.6: 35458298-35458298 on Build GRCh38
rs542433390 Chr.6: 35458274-35458274 on Build GRCh38
rs754902316 Chr.6: 35458361-35458362 on Build GRCh38
rs773632229 Chr.6: 35458309-35458309 on Build GRCh38
rs776318917 Chr.6: 35458419-35458419 on Build GRCh38
rs577613731 Chr.6: 35458249-35458249 on Build GRCh38
rs749409291 Chr.6: 35458297-35458297 on Build GRCh38
rs753852202 Chr.6: 35458326-35458326 on Build GRCh38
rs587778338 Chr.6: 35458401-35458402 on Build GRCh38
rs587778339 Chr.6: 35458385-35458385 on Build GRCh38
rs755165473 Chr.6: 35458380-35458380 on Build GRCh38
rs751388323 Chr.6: 35458371-35458371 on Build GRCh38
rs1007410805 Chr.6: 35458361-35458361 on Build GRCh38
rs752510326 Chr.6: 35458278-35458278 on Build GRCh38
rs748531682 Chr.6: 35458306-35458306 on Build GRCh38
rs940874632 Chr.6: 35458497-35458497 on Build GRCh38
rs377036453 Chr.6: 35458554-35458554 on Build GRCh38
rs748538800 Chr.6: 35458395-35458395 on Build GRCh38
rs775076977 Chr.6: 35458438-35458438 on Build GRCh38
rs770543570 Chr.6: 35458418-35458418 on Build GRCh38
rs117881500 Chr.6: 35458470-35458470 on Build GRCh38
rs374964864 Chr.6: 35458263-35458263 on Build GRCh38
rs912147314 Chr.6: 35458482-35458482 on Build GRCh38
rs542717013 Chr.6: 35458463-35458464 on Build GRCh38
rs141268133 Chr.6: 35458422-35458422 on Build GRCh38
rs756360940 Chr.6: 35458372-35458372 on Build GRCh38
rs886061330 Chr.6: 35458334-35458334 on Build GRCh38
rs377393916 Chr.6: 35458469-35458469 on Build GRCh38
rs114021030 Chr.6: 35458264-35458264 on Build GRCh38
rs934011285 Chr.6: 35458543-35458543 on Build GRCh38
rs781661435 Chr.6: 35458455-35458455 on Build GRCh38
rs770592868 Chr.6: 35458325-35458325 on Build GRCh38
rs1036520716 Chr.6: 35458507-35458507 on Build GRCh38
rs772385794 Chr.6: 35458308-35458308 on Build GRCh38
rs369631267 Chr.6: 35458282-35458282 on Build GRCh38
rs774356801 Chr.6: 35458501-35458501 on Build GRCh38
rs3823434 Chr.6: 35458398-35458398 on Build GRCh38
rs567451657 Chr.6: 35458570-35458570 on Build GRCh38
rs898909148 Chr.6: 35458276-35458276 on Build GRCh38
rs769363785 Chr.6: 35458425-35458425 on Build GRCh38
rs550037834 Chr.6: 35458288-35458288 on Build GRCh38
rs1055924483 Chr.6: 35458582-35458582 on Build GRCh38
rs778799162 Chr.6: 35458462-35458463 on Build GRCh38
rs762747463 Chr.6: 35458440-35458440 on Build GRCh38
rs45524646 Chr.6: 35458345-35458345 on Build GRCh38
rs185230199 Chr.6: 35458396-35458396 on Build GRCh38
rs932269895 Chr.6: 35458261-35458261 on Build GRCh38
rs145549434 Chr.6: 35458328-35458328 on Build GRCh38
rs372935921 Chr.6: 35458310-35458310 on Build GRCh38
rs146571065 Chr.6: 35458413-35458413 on Build GRCh38
rs192832780 Chr.6: 35458271-35458271 on Build GRCh38
rs779336261 Chr.6: 35458304-35458304 on Build GRCh38
rs567313440 Chr.6: 35458354-35458354 on Build GRCh38
rs766747763 Chr.6: 35458375-35458375 on Build GRCh38
rs45468994 Chr.6: 35458318-35458318 on Build GRCh38

View Full Product Details