Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 146774122-146774635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP2
Entrez Gene ID
26047
Gene Name
contactin associated protein-like 2
Gene Aliases
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1
UniGene
Hs.655684
Gene Chromosome Location
Chr.7: 146116035-148420998 on Build GRCh38
Transcripts
NM_014141.5 XM_017011950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6448997 c.206G>C p.G69A Chr.7: 146774379-146774379 on Build GRCh38
347346 c.133C>A p.H45N Chr.7: 146774306-146774306 on Build GRCh38
3259288 c.200A>G p.K67R Chr.7: 146774373-146774373 on Build GRCh38
7147807 c.144C>A p.F48L Chr.7: 146774317-146774317 on Build GRCh38
3259285 c.153C>A p.S51S Chr.7: 146774326-146774326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748265332 Chr.7: 146774435-146774435 on Build GRCh38
rs753396304 Chr.7: 146774353-146774353 on Build GRCh38
rs201121160 Chr.7: 146774236-146774236 on Build GRCh38
rs777706230 Chr.7: 146774302-146774302 on Build GRCh38
rs889232099 Chr.7: 146774542-146774542 on Build GRCh38
rs746949588 Chr.7: 146774387-146774387 on Build GRCh38
rs747525088 Chr.7: 146774346-146774346 on Build GRCh38
rs1050382487 Chr.7: 146774526-146774526 on Build GRCh38
rs528382745 Chr.7: 146774181-146774181 on Build GRCh38
rs994893502 Chr.7: 146774189-146774189 on Build GRCh38
rs777493334 Chr.7: 146774386-146774386 on Build GRCh38
rs148853070 Chr.7: 146774167-146774167 on Build GRCh38
rs376929846 Chr.7: 146774354-146774354 on Build GRCh38
rs778487279 Chr.7: 146774362-146774362 on Build GRCh38
rs528675746 Chr.7: 146774535-146774535 on Build GRCh38
rs928114479 Chr.7: 146774401-146774401 on Build GRCh38
rs771874359 Chr.7: 146774298-146774298 on Build GRCh38
rs796052367 Chr.7: 146774252-146774255 on Build GRCh38
rs552092384 Chr.7: 146774554-146774554 on Build GRCh38
rs768031513 Chr.7: 146774238-146774238 on Build GRCh38
rs767790084 Chr.7: 146774337-146774337 on Build GRCh38
rs781480662 Chr.7: 146774400-146774400 on Build GRCh38
rs116479521 Chr.7: 146774215-146774215 on Build GRCh38
rs931278183 Chr.7: 146774197-146774197 on Build GRCh38
rs773612421 Chr.7: 146774600-146774600 on Build GRCh38
rs1036324288 Chr.7: 146774180-146774180 on Build GRCh38
rs1036544851 Chr.7: 146774503-146774503 on Build GRCh38
rs969515416 Chr.7: 146774290-146774290 on Build GRCh38
rs551931845 Chr.7: 146774316-146774316 on Build GRCh38
rs564218122 Chr.7: 146774320-146774320 on Build GRCh38
rs930669219 Chr.7: 146774521-146774521 on Build GRCh38
rs563461480 Chr.7: 146774432-146774432 on Build GRCh38
rs780155429 Chr.7: 146774431-146774431 on Build GRCh38
rs940919254 Chr.7: 146774174-146774174 on Build GRCh38
rs1041277544 Chr.7: 146774560-146774560 on Build GRCh38
rs137924687 Chr.7: 146774309-146774309 on Build GRCh38
rs866359911 Chr.7: 146774293-146774293 on Build GRCh38
rs763855119 Chr.7: 146774241-146774244 on Build GRCh38
rs549740701 Chr.7: 146774340-146774340 on Build GRCh38
rs576178823 Chr.7: 146774216-146774216 on Build GRCh38
rs770819870 Chr.7: 146774391-146774391 on Build GRCh38
rs113167298 Chr.7: 146774446-146774447 on Build GRCh38
rs1025153306 Chr.7: 146774590-146774590 on Build GRCh38
rs529311609 Chr.7: 146774494-146774494 on Build GRCh38
rs546872812 Chr.7: 146774191-146774191 on Build GRCh38
rs755518759 Chr.7: 146774373-146774373 on Build GRCh38
rs1050499224 Chr.7: 146774211-146774211 on Build GRCh38
rs754683222 Chr.7: 146774277-146774277 on Build GRCh38
rs112724037 Chr.7: 146774230-146774230 on Build GRCh38
rs1045111434 Chr.7: 146774179-146774179 on Build GRCh38
rs187544703 Chr.7: 146774518-146774518 on Build GRCh38
rs999347930 Chr.7: 146774582-146774582 on Build GRCh38
rs571826491 Chr.7: 146774588-146774588 on Build GRCh38
rs778648331 Chr.7: 146774287-146774287 on Build GRCh38
rs764536218 Chr.7: 146774229-146774229 on Build GRCh38
rs750876987 Chr.7: 146774339-146774339 on Build GRCh38
rs900980561 Chr.7: 146774581-146774581 on Build GRCh38
rs866697251 Chr.7: 146774378-146774378 on Build GRCh38
rs199901768 Chr.7: 146774251-146774251 on Build GRCh38
rs753486205 Chr.7: 146774276-146774276 on Build GRCh38
rs372332415 Chr.7: 146774223-146774223 on Build GRCh38
rs779436784 Chr.7: 146774275-146774275 on Build GRCh38
rs899663772 Chr.7: 146774220-146774220 on Build GRCh38
rs899693455 Chr.7: 146774456-146774456 on Build GRCh38
rs747896321 Chr.7: 146774288-146774288 on Build GRCh38
rs561826203 Chr.7: 146774495-146774495 on Build GRCh38
rs763331605 Chr.7: 146774224-146774224 on Build GRCh38
rs749280132 Chr.7: 146774448-146774449 on Build GRCh38
rs371991891 Chr.7: 146774280-146774280 on Build GRCh38
rs183378415 Chr.7: 146774325-146774325 on Build GRCh38
rs746922194 Chr.7: 146774306-146774306 on Build GRCh38
rs113752065 Chr.7: 146774447-146774448 on Build GRCh38
rs550564732 Chr.7: 146774598-146774598 on Build GRCh38
rs762223005 Chr.7: 146774331-146774331 on Build GRCh38
rs916375383 Chr.7: 146774313-146774313 on Build GRCh38
rs1047471077 Chr.7: 146774360-146774360 on Build GRCh38
rs549173430 Chr.7: 146774496-146774496 on Build GRCh38
rs895466967 Chr.7: 146774565-146774565 on Build GRCh38
rs889188780 Chr.7: 146774213-146774213 on Build GRCh38
rs191067437 Chr.7: 146774154-146774154 on Build GRCh38
rs758143699 Chr.7: 146774381-146774381 on Build GRCh38

View Full Product Details