Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150946791-150947280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH2
Entrez Gene ID
3757
Gene Name
potassium voltage-gated channel subfamily H member 2
Gene Aliases
ERG-1, ERG1, H-ERG, HERG, HERG1, Kv11.1, LQT2, SQT1
UniGene
Hs.647099
Gene Chromosome Location
Chr.7: 150944956-150978314 on Build GRCh38
Transcripts
NM_000238.3 NM_172057.2 XM_011516185.2 XM_017012195.1 XM_017012196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7076297 c.2289C>T p.L763L Chr.7: 150946898-150946898 on Build GRCh38
4488101 c.2255C>T p.S752F Chr.7: 150946932-150946932 on Build GRCh38
6690861 c.3298C>A p.L1100I Chr.7: 150946909-150946909 on Build GRCh38
6690876 c.3239C>T p.A1080V Chr.7: 150946968-150946968 on Build GRCh38
1318998 c.3289G>A p.V1097I Chr.7: 150946918-150946918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767372791 Chr.7: 150946971-150946971 on Build GRCh38
rs1044888463 Chr.7: 150946847-150946848 on Build GRCh38
rs568807354 Chr.7: 150947113-150947113 on Build GRCh38
rs794728405 Chr.7: 150946902-150946902 on Build GRCh38
rs758395817 Chr.7: 150947069-150947069 on Build GRCh38
rs754108936 Chr.7: 150946899-150946899 on Build GRCh38
rs779333020 Chr.7: 150946977-150946977 on Build GRCh38
rs996549480 Chr.7: 150947106-150947106 on Build GRCh38
rs776100082 Chr.7: 150946931-150946931 on Build GRCh38
rs140395995 Chr.7: 150946853-150946853 on Build GRCh38
rs941037238 Chr.7: 150947077-150947077 on Build GRCh38
rs751559990 Chr.7: 150946960-150946960 on Build GRCh38
rs781609392 Chr.7: 150947064-150947064 on Build GRCh38
rs151031345 Chr.7: 150947004-150947004 on Build GRCh38
rs1011996432 Chr.7: 150947159-150947159 on Build GRCh38
rs199473028 Chr.7: 150946983-150946983 on Build GRCh38
rs915101523 Chr.7: 150946839-150946839 on Build GRCh38
rs75688212 Chr.7: 150947098-150947098 on Build GRCh38
rs199473029 Chr.7: 150946974-150946974 on Build GRCh38
rs747667985 Chr.7: 150947080-150947080 on Build GRCh38
rs758947934 Chr.7: 150946878-150946878 on Build GRCh38
rs990623569 Chr.7: 150946836-150946836 on Build GRCh38
rs570964313 Chr.7: 150947171-150947171 on Build GRCh38
rs772040715 Chr.7: 150947083-150947083 on Build GRCh38
rs552586241 Chr.7: 150946831-150946831 on Build GRCh38
rs139722868 Chr.7: 150947020-150947020 on Build GRCh38
rs774112063 Chr.7: 150947021-150947021 on Build GRCh38
rs899985020 Chr.7: 150946860-150946860 on Build GRCh38
rs794728473 Chr.7: 150946955-150946956 on Build GRCh38
rs541259035 Chr.7: 150947044-150947044 on Build GRCh38
rs201337201 Chr.7: 150947072-150947072 on Build GRCh38
rs1057518089 Chr.7: 150947006-150947007 on Build GRCh38
rs999138416 Chr.7: 150947134-150947134 on Build GRCh38
rs1020610826 Chr.7: 150946948-150946948 on Build GRCh38
rs769228898 Chr.7: 150946938-150946938 on Build GRCh38
rs753375396 Chr.7: 150946955-150946955 on Build GRCh38
rs1009829215 Chr.7: 150946941-150946941 on Build GRCh38
rs868543412 Chr.7: 150946897-150946897 on Build GRCh38
rs753938571 Chr.7: 150946973-150946973 on Build GRCh38
rs553337493 Chr.7: 150947031-150947031 on Build GRCh38
rs773125785 Chr.7: 150947030-150947030 on Build GRCh38
rs199473545 Chr.7: 150946929-150946929 on Build GRCh38
rs970093127 Chr.7: 150947060-150947060 on Build GRCh38
rs555797304 Chr.7: 150946996-150946996 on Build GRCh38
rs780113151 Chr.7: 150946896-150946896 on Build GRCh38
rs762510312 Chr.7: 150946956-150946956 on Build GRCh38
rs761157161 Chr.7: 150946854-150946854 on Build GRCh38
rs772691915 Chr.7: 150946927-150946927 on Build GRCh38
rs374915150 Chr.7: 150947103-150947103 on Build GRCh38
rs1051237734 Chr.7: 150947112-150947112 on Build GRCh38
rs190636009 Chr.7: 150947245-150947245 on Build GRCh38
rs527971628 Chr.7: 150947136-150947136 on Build GRCh38
rs778119775 Chr.7: 150946982-150946982 on Build GRCh38
rs768903142 Chr.7: 150946844-150946844 on Build GRCh38
rs531761197 Chr.7: 150947074-150947074 on Build GRCh38
rs761811244 Chr.7: 150947022-150947022 on Build GRCh38
rs200181671 Chr.7: 150946846-150946846 on Build GRCh38
rs903510655 Chr.7: 150947237-150947237 on Build GRCh38
rs532489197 Chr.7: 150946828-150946828 on Build GRCh38
rs770485005 Chr.7: 150946861-150946861 on Build GRCh38
rs555269990 Chr.7: 150947035-150947035 on Build GRCh38
rs550785636 Chr.7: 150947134-150947135 on Build GRCh38
rs769425141 Chr.7: 150947000-150947000 on Build GRCh38
rs533103813 Chr.7: 150947059-150947059 on Build GRCh38
rs1043061010 Chr.7: 150947229-150947229 on Build GRCh38
rs369564588 Chr.7: 150946947-150946947 on Build GRCh38
rs202198564 Chr.7: 150946928-150946928 on Build GRCh38
rs1029745712 Chr.7: 150946989-150946989 on Build GRCh38
rs1005325992 Chr.7: 150946952-150946952 on Build GRCh38
rs757147615 Chr.7: 150946893-150946893 on Build GRCh38
rs887119541 Chr.7: 150946820-150946820 on Build GRCh38
rs753814212 Chr.7: 150946844-150946845 on Build GRCh38
rs779662956 Chr.7: 150947070-150947070 on Build GRCh38
rs201382073 Chr.7: 150946978-150946978 on Build GRCh38
rs760523800 Chr.7: 150947086-150947086 on Build GRCh38
rs569278564 Chr.7: 150946838-150946838 on Build GRCh38
rs199473031 Chr.7: 150946885-150946885 on Build GRCh38
rs372337645 Chr.7: 150947067-150947067 on Build GRCh38
rs200182770 Chr.7: 150946991-150946991 on Build GRCh38
rs41313752 Chr.7: 150947034-150947034 on Build GRCh38
rs932723749 Chr.7: 150947169-150947169 on Build GRCh38
rs199473030 Chr.7: 150946918-150946918 on Build GRCh38
rs761462107 Chr.7: 150946970-150946970 on Build GRCh38
rs781624566 Chr.7: 150946992-150946992 on Build GRCh38
rs189457419 Chr.7: 150946886-150946886 on Build GRCh38
rs41307270 Chr.7: 150947043-150947043 on Build GRCh38
rs41312087 Chr.7: 150946979-150946979 on Build GRCh38
rs1009804773 Chr.7: 150947215-150947215 on Build GRCh38
rs899592870 Chr.7: 150947096-150947096 on Build GRCh38
rs766532760 Chr.7: 150946828-150946828 on Build GRCh38
rs764749875 Chr.7: 150946883-150946883 on Build GRCh38
rs927776131 Chr.7: 150947066-150947066 on Build GRCh38
rs199473027 Chr.7: 150947011-150947011 on Build GRCh38
rs41313755 Chr.7: 150947069-150947069 on Build GRCh38
rs373034438 Chr.7: 150947144-150947144 on Build GRCh38
rs199473041 Chr.7: 150946905-150946905 on Build GRCh38
rs908463883 Chr.7: 150947261-150947261 on Build GRCh38
rs147510453 Chr.7: 150946932-150946932 on Build GRCh38
rs376889824 Chr.7: 150947125-150947125 on Build GRCh38
rs748606749 Chr.7: 150946919-150946919 on Build GRCh38
rs367544578 Chr.7: 150946856-150946856 on Build GRCh38
rs751302925 Chr.7: 150946889-150946889 on Build GRCh38
rs776385441 Chr.7: 150946865-150946865 on Build GRCh38
rs765524161 Chr.7: 150947037-150947037 on Build GRCh38
rs1051229135 Chr.7: 150947170-150947170 on Build GRCh38
rs957614475 Chr.7: 150947150-150947150 on Build GRCh38
rs767156792 Chr.7: 150947029-150947029 on Build GRCh38
rs755065620 Chr.7: 150946904-150946904 on Build GRCh38
rs368701315 Chr.7: 150947045-150947045 on Build GRCh38
rs181119727 Chr.7: 150947058-150947058 on Build GRCh38
rs541394173 Chr.7: 150947018-150947018 on Build GRCh38
rs755261687 Chr.7: 150946976-150946976 on Build GRCh38

View Full Product Details