Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30132394-30132902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEPROTL1
Entrez Gene ID
23484
Gene Name
leptin receptor overlapping transcript-like 1
Gene Aliases
HSPC112, Vps55, my047
UniGene
Hs.146585
Gene Chromosome Location
Chr.8: 30095406-30137706 on Build GRCh38
Transcripts
NM_001128208.1 NM_001100916.1 XM_017013726.1

Gene Symbol
MBOAT4
Entrez Gene ID
619373
Gene Name
membrane bound O-acyltransferase domain containing 4
Gene Aliases
FKSG89, GOAT, OACT4
UniGene
Hs.627194
Gene Chromosome Location
Chr.8: 30131454-30148768 on Build GRCh38
Transcripts
NM_001128208.1 NM_001100916.1 XM_017013726.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5409413 c.833G>A p.S278N Chr.8: 30132418-30132418 on Build GRCh38
4785642 c.370G>A p.A124T Chr.8: 30132563-30132563 on Build GRCh38
1489228 c.693A>G p.G231G Chr.8: 30132558-30132558 on Build GRCh38
1456575 c.786C>T p.D262D Chr.8: 30132465-30132465 on Build GRCh38
1456576 c.688G>A p.A230T Chr.8: 30132563-30132563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145757853 Chr.8: 30132627-30132627 on Build GRCh38
rs914049879 Chr.8: 30132468-30132468 on Build GRCh38
rs146230770 Chr.8: 30132457-30132457 on Build GRCh38
rs999591754 Chr.8: 30132825-30132825 on Build GRCh38
rs148229213 Chr.8: 30132711-30132711 on Build GRCh38
rs757655456 Chr.8: 30132571-30132571 on Build GRCh38
rs953346982 Chr.8: 30132700-30132700 on Build GRCh38
rs890040184 Chr.8: 30132768-30132768 on Build GRCh38
rs546935755 Chr.8: 30132781-30132781 on Build GRCh38
rs139513140 Chr.8: 30132850-30132850 on Build GRCh38
rs777359025 Chr.8: 30132562-30132562 on Build GRCh38
rs752092925 Chr.8: 30132553-30132553 on Build GRCh38
rs142593371 Chr.8: 30132428-30132428 on Build GRCh38
rs149781890 Chr.8: 30132629-30132629 on Build GRCh38
rs145609493 Chr.8: 30132799-30132799 on Build GRCh38
rs752614032 Chr.8: 30132782-30132785 on Build GRCh38
rs146052068 Chr.8: 30132656-30132656 on Build GRCh38
rs946870946 Chr.8: 30132481-30132481 on Build GRCh38
rs139580380 Chr.8: 30132804-30132804 on Build GRCh38
rs142363651 Chr.8: 30132772-30132772 on Build GRCh38
rs754928266 Chr.8: 30132535-30132535 on Build GRCh38
rs142604585 Chr.8: 30132564-30132564 on Build GRCh38
rs143535986 Chr.8: 30132837-30132837 on Build GRCh38
rs1027549366 Chr.8: 30132519-30132519 on Build GRCh38
rs16876563 Chr.8: 30132559-30132559 on Build GRCh38
rs957933474 Chr.8: 30132580-30132580 on Build GRCh38
rs760001524 Chr.8: 30132433-30132433 on Build GRCh38
rs1020238174 Chr.8: 30132793-30132793 on Build GRCh38
rs139450783 Chr.8: 30132796-30132796 on Build GRCh38
rs140014094 Chr.8: 30132640-30132640 on Build GRCh38
rs979647522 Chr.8: 30132843-30132843 on Build GRCh38
rs530929110 Chr.8: 30132620-30132620 on Build GRCh38
rs762897669 Chr.8: 30132526-30132526 on Build GRCh38
rs965421809 Chr.8: 30132818-30132818 on Build GRCh38
rs564667228 Chr.8: 30132464-30132464 on Build GRCh38
rs139376997 Chr.8: 30132537-30132537 on Build GRCh38
rs149271404 Chr.8: 30132458-30132458 on Build GRCh38
rs780304488 Chr.8: 30132859-30132859 on Build GRCh38
rs61745343 Chr.8: 30132685-30132685 on Build GRCh38
rs953326968 Chr.8: 30132721-30132749 on Build GRCh38
rs769971085 Chr.8: 30132734-30132734 on Build GRCh38
rs140544481 Chr.8: 30132765-30132765 on Build GRCh38
rs139358115 Chr.8: 30132687-30132687 on Build GRCh38
rs772168339 Chr.8: 30132444-30132444 on Build GRCh38
rs898854639 Chr.8: 30132763-30132763 on Build GRCh38
rs766256069 Chr.8: 30132534-30132534 on Build GRCh38
rs927697384 Chr.8: 30132676-30132676 on Build GRCh38
rs994927279 Chr.8: 30132766-30132766 on Build GRCh38
rs149726575 Chr.8: 30132802-30132802 on Build GRCh38
rs142642163 Chr.8: 30132743-30132743 on Build GRCh38
rs146026417 Chr.8: 30132425-30132425 on Build GRCh38
rs762096309 Chr.8: 30132779-30132779 on Build GRCh38
rs746394094 Chr.8: 30132601-30132601 on Build GRCh38
rs142899151 Chr.8: 30132619-30132619 on Build GRCh38
rs560382676 Chr.8: 30132522-30132522 on Build GRCh38
rs965721315 Chr.8: 30132724-30132724 on Build GRCh38
rs150467698 Chr.8: 30132762-30132762 on Build GRCh38
rs748914303 Chr.8: 30132568-30132568 on Build GRCh38
rs138538560 Chr.8: 30132753-30132753 on Build GRCh38
rs138447563 Chr.8: 30132461-30132461 on Build GRCh38
rs144199295 Chr.8: 30132790-30132790 on Build GRCh38
rs989895855 Chr.8: 30132858-30132858 on Build GRCh38
rs978057179 Chr.8: 30132561-30132561 on Build GRCh38
rs146547536 Chr.8: 30132784-30132784 on Build GRCh38
rs192400927 Chr.8: 30132594-30132594 on Build GRCh38
rs148967455 Chr.8: 30132622-30132622 on Build GRCh38
rs150098506 Chr.8: 30132465-30132465 on Build GRCh38
rs147722231 Chr.8: 30132576-30132576 on Build GRCh38
rs141252348 Chr.8: 30132698-30132698 on Build GRCh38
rs759562726 Chr.8: 30132827-30132827 on Build GRCh38
rs138038257 Chr.8: 30132450-30132450 on Build GRCh38
rs989392544 Chr.8: 30132595-30132595 on Build GRCh38
rs146255946 Chr.8: 30132697-30132697 on Build GRCh38
rs142693229 Chr.8: 30132876-30132876 on Build GRCh38
rs563758119 Chr.8: 30132521-30132521 on Build GRCh38
rs774240200 Chr.8: 30132805-30132805 on Build GRCh38
rs576685952 Chr.8: 30132618-30132618 on Build GRCh38
rs141275857 Chr.8: 30132774-30132774 on Build GRCh38
rs867794258 Chr.8: 30132435-30132435 on Build GRCh38
rs147364720 Chr.8: 30132871-30132871 on Build GRCh38
rs140690092 Chr.8: 30132513-30132513 on Build GRCh38
rs149476365 Chr.8: 30132714-30132714 on Build GRCh38
rs137925555 Chr.8: 30132718-30132718 on Build GRCh38
rs538616586 Chr.8: 30132674-30132674 on Build GRCh38
rs143939823 Chr.8: 30132712-30132712 on Build GRCh38
rs151282168 Chr.8: 30132767-30132767 on Build GRCh38
rs540570721 Chr.8: 30132417-30132417 on Build GRCh38
rs150553052 Chr.8: 30132836-30132836 on Build GRCh38
rs146920710 Chr.8: 30132719-30132719 on Build GRCh38
rs1014636960 Chr.8: 30132449-30132449 on Build GRCh38

View Full Product Details