Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31032786-31033317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PURG
Entrez Gene ID
29942
Gene Name
purine rich element binding protein G
Gene Aliases
PURG-A, PURG-B
UniGene
Hs.373778
Gene Chromosome Location
Chr.8: 30995802-31033715 on Build GRCh38
Transcripts
NM_001015508.2 NM_001323311.1 NM_001323312.1 NM_013357.2 NM_000553.4

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
UniGene
Hs.632050
Gene Chromosome Location
Chr.8: 31033262-31173761 on Build GRCh38
Transcripts
NM_001015508.2 NM_001323311.1 NM_001323312.1 NM_013357.2 NM_000553.4

SNP Detail

SNP ID Chromosome Location
rs569457300 Chr.8: 31032811-31032811 on Build GRCh38
rs2005383 Chr.8: 31032903-31032903 on Build GRCh38
rs943329669 Chr.8: 31033219-31033219 on Build GRCh38
rs1013306604 Chr.8: 31033127-31033127 on Build GRCh38
rs1033374670 Chr.8: 31033050-31033050 on Build GRCh38
rs984845943 Chr.8: 31032901-31032901 on Build GRCh38
rs767185289 Chr.8: 31032854-31032854 on Build GRCh38
rs1024640643 Chr.8: 31033128-31033133 on Build GRCh38
rs902032101 Chr.8: 31033013-31033013 on Build GRCh38
rs1004542544 Chr.8: 31032930-31032930 on Build GRCh38
rs976169255 Chr.8: 31033239-31033239 on Build GRCh38
rs770403998 Chr.8: 31032812-31032812 on Build GRCh38
rs1027229664 Chr.8: 31033191-31033191 on Build GRCh38
rs1043041638 Chr.8: 31033020-31033020 on Build GRCh38
rs972573664 Chr.8: 31033183-31033183 on Build GRCh38
rs943465158 Chr.8: 31032925-31032925 on Build GRCh38
rs866887549 Chr.8: 31032865-31032865 on Build GRCh38
rs945490119 Chr.8: 31033019-31033019 on Build GRCh38
rs1040454866 Chr.8: 31032982-31032982 on Build GRCh38
rs952681402 Chr.8: 31033193-31033193 on Build GRCh38
rs569223819 Chr.8: 31032839-31032839 on Build GRCh38
rs923317706 Chr.8: 31033249-31033249 on Build GRCh38
rs931213467 Chr.8: 31032858-31032858 on Build GRCh38
rs985765747 Chr.8: 31033195-31033195 on Build GRCh38
rs910567937 Chr.8: 31033217-31033217 on Build GRCh38
rs894774605 Chr.8: 31033067-31033067 on Build GRCh38
rs961201983 Chr.8: 31033173-31033173 on Build GRCh38
rs904140600 Chr.8: 31033032-31033032 on Build GRCh38
rs1056966578 Chr.8: 31032886-31032886 on Build GRCh38
rs146627351 Chr.8: 31032817-31032817 on Build GRCh38
rs11574154 Chr.8: 31033189-31033189 on Build GRCh38
rs1001522206 Chr.8: 31033040-31033040 on Build GRCh38
rs945436298 Chr.8: 31033251-31033251 on Build GRCh38

View Full Product Details