Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80022159-80022659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX22
Entrez Gene ID
50945
Gene Name
T-box 22
Gene Aliases
ABERS, CLPA, CPX, TBXX, dJ795G23.1
UniGene
Hs.374253
Gene Chromosome Location
Chr.X: 80014730-80035828 on Build GRCh38
Transcripts
NM_001109878.1 NM_001109879.1 NM_016954.2 XM_005262136.2 XM_006724657.3 XM_011530972.1

SNP Detail

SNP ID Chromosome Location
rs771304058 Chr.X: 80022223-80022223 on Build GRCh38
rs759649828 Chr.X: 80022393-80022393 on Build GRCh38
rs551664098 Chr.X: 80022286-80022286 on Build GRCh38
rs759639633 Chr.X: 80022518-80022518 on Build GRCh38
rs967085236 Chr.X: 80022214-80022214 on Build GRCh38
rs1033203876 Chr.X: 80022522-80022522 on Build GRCh38
rs772130317 Chr.X: 80022374-80022374 on Build GRCh38
rs945189214 Chr.X: 80022574-80022574 on Build GRCh38
rs200143305 Chr.X: 80022483-80022484 on Build GRCh38
rs750415957 Chr.X: 80022273-80022273 on Build GRCh38
rs778577936 Chr.X: 80022441-80022441 on Build GRCh38
rs1029348139 Chr.X: 80022199-80022199 on Build GRCh38
rs752053011 Chr.X: 80022428-80022428 on Build GRCh38
rs773703001 Chr.X: 80022359-80022359 on Build GRCh38
rs778207010 Chr.X: 80022328-80022328 on Build GRCh38
rs773969245 Chr.X: 80022262-80022262 on Build GRCh38
rs772915248 Chr.X: 80022471-80022471 on Build GRCh38
rs998446704 Chr.X: 80022493-80022493 on Build GRCh38
rs371055319 Chr.X: 80022378-80022378 on Build GRCh38
rs747809017 Chr.X: 80022444-80022444 on Build GRCh38
rs781388378 Chr.X: 80022438-80022438 on Build GRCh38
rs181230650 Chr.X: 80022626-80022626 on Build GRCh38
rs150098741 Chr.X: 80022300-80022300 on Build GRCh38
rs950279249 Chr.X: 80022402-80022402 on Build GRCh38
rs747337881 Chr.X: 80022222-80022222 on Build GRCh38
rs768403428 Chr.X: 80022358-80022358 on Build GRCh38
rs761364666 Chr.X: 80022360-80022360 on Build GRCh38
rs748427666 Chr.X: 80022282-80022282 on Build GRCh38
rs368489258 Chr.X: 80022242-80022242 on Build GRCh38
rs753743609 Chr.X: 80022383-80022388 on Build GRCh38
rs778255323 Chr.X: 80022470-80022470 on Build GRCh38
rs1039891745 Chr.X: 80022469-80022469 on Build GRCh38
rs148838409 Chr.X: 80022353-80022353 on Build GRCh38
rs771728632 Chr.X: 80022373-80022373 on Build GRCh38
rs902889360 Chr.X: 80022504-80022504 on Build GRCh38
rs185298778 Chr.X: 80022261-80022261 on Build GRCh38
rs766283070 Chr.X: 80022283-80022283 on Build GRCh38
rs138387312 Chr.X: 80022341-80022341 on Build GRCh38
rs761633810 Chr.X: 80022265-80022265 on Build GRCh38
rs370933828 Chr.X: 80022604-80022604 on Build GRCh38
rs73496513 Chr.X: 80022257-80022257 on Build GRCh38
rs104894945 Chr.X: 80022435-80022435 on Build GRCh38
rs891910998 Chr.X: 80022529-80022529 on Build GRCh38
rs757815223 Chr.X: 80022434-80022434 on Build GRCh38
rs5959656 Chr.X: 80022227-80022227 on Build GRCh38
rs770084170 Chr.X: 80022439-80022439 on Build GRCh38
rs367771247 Chr.X: 80022314-80022314 on Build GRCh38
rs753716446 Chr.X: 80022385-80022385 on Build GRCh38
rs760763118 Chr.X: 80022377-80022377 on Build GRCh38
rs766263474 Chr.X: 80022257-80022258 on Build GRCh38
rs901117376 Chr.X: 80022202-80022202 on Build GRCh38
rs755916335 Chr.X: 80022280-80022280 on Build GRCh38
rs1032841733 Chr.X: 80022266-80022266 on Build GRCh38
rs917472096 Chr.X: 80022375-80022375 on Build GRCh38
rs758506603 Chr.X: 80022325-80022325 on Build GRCh38
rs771215994 Chr.X: 80022342-80022342 on Build GRCh38
rs777832093 Chr.X: 80022425-80022425 on Build GRCh38
rs765461432 Chr.X: 80022407-80022407 on Build GRCh38
rs767601225 Chr.X: 80022269-80022269 on Build GRCh38
rs748304646 Chr.X: 80022319-80022319 on Build GRCh38
rs1050710663 Chr.X: 80022495-80022495 on Build GRCh38
rs777096494 Chr.X: 80022513-80022513 on Build GRCh38
rs771465183 Chr.X: 80022512-80022512 on Build GRCh38
rs997829130 Chr.X: 80022189-80022189 on Build GRCh38
rs1042244921 Chr.X: 80022587-80022587 on Build GRCh38
rs769828268 Chr.X: 80022620-80022620 on Build GRCh38
rs781665863 Chr.X: 80022345-80022345 on Build GRCh38
rs941550625 Chr.X: 80022384-80022384 on Build GRCh38
rs773538326 Chr.X: 80022219-80022219 on Build GRCh38
rs957207345 Chr.X: 80022603-80022603 on Build GRCh38
rs758412350 Chr.X: 80022415-80022415 on Build GRCh38
rs927263531 Chr.X: 80022454-80022454 on Build GRCh38
rs752888029 Chr.X: 80022414-80022414 on Build GRCh38
rs915850492 Chr.X: 80022292-80022292 on Build GRCh38

View Full Product Details