Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 85107037-85107541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SATL1
Entrez Gene ID
340562
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Gene Aliases
-
UniGene
Hs.640783
Gene Chromosome Location
Chr.X: 85092286-85116229 on Build GRCh38
Transcripts
NM_001012980.2 XM_011530940.2

SNP Detail

SNP ID Chromosome Location
rs146446228 Chr.X: 85107292-85107292 on Build GRCh38
rs946051962 Chr.X: 85107102-85107102 on Build GRCh38
rs866659733 Chr.X: 85107344-85107344 on Build GRCh38
rs375536211 Chr.X: 85107326-85107326 on Build GRCh38
rs141781891 Chr.X: 85107395-85107395 on Build GRCh38
rs759094694 Chr.X: 85107382-85107382 on Build GRCh38
rs780610049 Chr.X: 85107373-85107375 on Build GRCh38
rs745314749 Chr.X: 85107516-85107516 on Build GRCh38
rs750948898 Chr.X: 85107323-85107323 on Build GRCh38
rs1018436688 Chr.X: 85107371-85107371 on Build GRCh38
rs937299199 Chr.X: 85107161-85107161 on Build GRCh38
rs769086209 Chr.X: 85107357-85107357 on Build GRCh38
rs771661636 Chr.X: 85107466-85107467 on Build GRCh38
rs766745061 Chr.X: 85107489-85107489 on Build GRCh38
rs138651773 Chr.X: 85107370-85107370 on Build GRCh38
rs372564257 Chr.X: 85107487-85107487 on Build GRCh38
rs112006376 Chr.X: 85107403-85107403 on Build GRCh38
rs769581693 Chr.X: 85107389-85107389 on Build GRCh38
rs747189797 Chr.X: 85107499-85107499 on Build GRCh38
rs749574125 Chr.X: 85107356-85107356 on Build GRCh38
rs751875933 Chr.X: 85107295-85107295 on Build GRCh38
rs543639746 Chr.X: 85107210-85107210 on Build GRCh38
rs749741589 Chr.X: 85107379-85107380 on Build GRCh38
rs146269642 Chr.X: 85107518-85107518 on Build GRCh38
rs754411074 Chr.X: 85107493-85107493 on Build GRCh38
rs769398793 Chr.X: 85107517-85107517 on Build GRCh38
rs756579431 Chr.X: 85107301-85107301 on Build GRCh38
rs757213730 Chr.X: 85107410-85107410 on Build GRCh38
rs949811333 Chr.X: 85107254-85107254 on Build GRCh38
rs1036538720 Chr.X: 85107266-85107266 on Build GRCh38
rs756246895 Chr.X: 85107478-85107478 on Build GRCh38
rs185314077 Chr.X: 85107259-85107259 on Build GRCh38
rs1039499603 Chr.X: 85107198-85107198 on Build GRCh38
rs978667763 Chr.X: 85107112-85107112 on Build GRCh38
rs779144730 Chr.X: 85107509-85107509 on Build GRCh38
rs925876244 Chr.X: 85107152-85107152 on Build GRCh38
rs758459626 Chr.X: 85107369-85107369 on Build GRCh38
rs990209590 Chr.X: 85107124-85107124 on Build GRCh38
rs899786761 Chr.X: 85107228-85107228 on Build GRCh38
rs747703674 Chr.X: 85107438-85107439 on Build GRCh38
rs778285326 Chr.X: 85107432-85107432 on Build GRCh38
rs776064615 Chr.X: 85107378-85107378 on Build GRCh38
rs867698433 Chr.X: 85107297-85107297 on Build GRCh38
rs76610710 Chr.X: 85107173-85107173 on Build GRCh38
rs767865222 Chr.X: 85107470-85107470 on Build GRCh38
rs763912481 Chr.X: 85107084-85107084 on Build GRCh38
rs755569722 Chr.X: 85107421-85107422 on Build GRCh38
rs370694394 Chr.X: 85107303-85107303 on Build GRCh38
rs752372827 Chr.X: 85107280-85107280 on Build GRCh38
rs748526319 Chr.X: 85107504-85107504 on Build GRCh38
rs189944635 Chr.X: 85107333-85107333 on Build GRCh38
rs757379473 Chr.X: 85107296-85107296 on Build GRCh38
rs780858954 Chr.X: 85107511-85107511 on Build GRCh38
rs994916046 Chr.X: 85107302-85107302 on Build GRCh38
rs368314953 Chr.X: 85107364-85107364 on Build GRCh38
rs755295828 Chr.X: 85107494-85107494 on Build GRCh38
rs369540498 Chr.X: 85107179-85107179 on Build GRCh38
rs774706209 Chr.X: 85107424-85107424 on Build GRCh38
rs762180135 Chr.X: 85107453-85107453 on Build GRCh38
rs979169604 Chr.X: 85107519-85107519 on Build GRCh38
rs934778028 Chr.X: 85107118-85107120 on Build GRCh38
rs762841112 Chr.X: 85107284-85107284 on Build GRCh38
rs750460919 Chr.X: 85107477-85107477 on Build GRCh38

View Full Product Details