Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90962517-90963046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCCD1
Entrez Gene ID
91433
Gene Name
RCC1 domain containing 1
Gene Aliases
-
UniGene
Hs.655895
Gene Chromosome Location
Chr.15: 90954864-90964858 on Build GRCh38
Transcripts
NM_001017919.1 NM_033544.2

SNP Detail

SNP ID Chromosome Location
rs113557967 Chr.15: 90962653-90962653 on Build GRCh38
rs756473671 Chr.15: 90962774-90962774 on Build GRCh38
rs953865351 Chr.15: 90962917-90962917 on Build GRCh38
rs764903697 Chr.15: 90962789-90962789 on Build GRCh38
rs1048522224 Chr.15: 90962661-90962661 on Build GRCh38
rs753055750 Chr.15: 90962692-90962692 on Build GRCh38
rs974340368 Chr.15: 90962574-90962574 on Build GRCh38
rs553034199 Chr.15: 90962641-90962641 on Build GRCh38
rs868293629 Chr.15: 90962871-90962871 on Build GRCh38
rs567237213 Chr.15: 90962593-90962593 on Build GRCh38
rs76898760 Chr.15: 90962950-90962950 on Build GRCh38
rs916108924 Chr.15: 90962634-90962634 on Build GRCh38
rs1003734841 Chr.15: 90962993-90962993 on Build GRCh38
rs1034789369 Chr.15: 90963000-90963000 on Build GRCh38
rs902328455 Chr.15: 90962986-90962986 on Build GRCh38
rs1004643997 Chr.15: 90962877-90962877 on Build GRCh38
rs572649356 Chr.15: 90962702-90962702 on Build GRCh38
rs546518627 Chr.15: 90962782-90962782 on Build GRCh38
rs1042121951 Chr.15: 90962680-90962680 on Build GRCh38
rs973929272 Chr.15: 90962899-90962899 on Build GRCh38
rs757913265 Chr.15: 90962837-90962837 on Build GRCh38
rs908636562 Chr.15: 90962670-90962670 on Build GRCh38
rs541003926 Chr.15: 90963004-90963004 on Build GRCh38
rs79548680 Chr.15: 90962549-90962549 on Build GRCh38
rs1000362431 Chr.15: 90962686-90962686 on Build GRCh38
rs527595723 Chr.15: 90962689-90962690 on Build GRCh38
rs940138639 Chr.15: 90962935-90962935 on Build GRCh38
rs1049732312 Chr.15: 90962969-90962969 on Build GRCh38
rs185861994 Chr.15: 90962558-90962558 on Build GRCh38
rs74039175 Chr.15: 90962984-90962984 on Build GRCh38
rs912204806 Chr.15: 90962666-90962666 on Build GRCh38
rs936378952 Chr.15: 90963019-90963019 on Build GRCh38
rs903631778 Chr.15: 90962683-90962683 on Build GRCh38
rs1013088635 Chr.15: 90962778-90962778 on Build GRCh38
rs952927449 Chr.15: 90962893-90962893 on Build GRCh38
rs540550308 Chr.15: 90962565-90962565 on Build GRCh38
rs1027244723 Chr.15: 90963018-90963018 on Build GRCh38
rs750026482 Chr.15: 90962802-90962802 on Build GRCh38
rs932680185 Chr.15: 90962963-90962963 on Build GRCh38
rs1041116276 Chr.15: 90962936-90962936 on Build GRCh38
rs1003294040 Chr.15: 90962792-90962792 on Build GRCh38
rs894650568 Chr.15: 90962756-90962756 on Build GRCh38
rs1049657699 Chr.15: 90962701-90962701 on Build GRCh38
rs990418104 Chr.15: 90962865-90962865 on Build GRCh38
rs746524445 Chr.15: 90962977-90962977 on Build GRCh38
rs558030578 Chr.15: 90962846-90962846 on Build GRCh38
rs945044955 Chr.15: 90962679-90962679 on Build GRCh38
rs113150816 Chr.15: 90962648-90962648 on Build GRCh38
rs982801194 Chr.15: 90962544-90962544 on Build GRCh38
rs766762589 Chr.15: 90962863-90962863 on Build GRCh38
rs1055073905 Chr.15: 90962732-90962732 on Build GRCh38
rs959059577 Chr.15: 90962862-90962862 on Build GRCh38
rs79840930 Chr.15: 90962597-90962597 on Build GRCh38
rs781165680 Chr.15: 90963001-90963001 on Build GRCh38
rs576579167 Chr.15: 90962897-90962897 on Build GRCh38
rs1051010068 Chr.15: 90962644-90962644 on Build GRCh38
rs1020759628 Chr.15: 90962757-90962758 on Build GRCh38
rs146344457 Chr.15: 90962904-90962904 on Build GRCh38
rs1015665492 Chr.15: 90962890-90962890 on Build GRCh38
rs1010822628 Chr.15: 90962981-90962981 on Build GRCh38

View Full Product Details