Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35438782-35439357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLFN13
Entrez Gene ID
146857
Gene Name
schlafen family member 13
Gene Aliases
SLFN10
UniGene
Hs.462833
Gene Chromosome Location
Chr.17: 35435096-35448837 on Build GRCh38
Transcripts
NM_144682.5 XM_011524383.2 XM_017024232.1 XM_017024233.1

SNP Detail

SNP ID Chromosome Location
rs765646507 Chr.17: 35439056-35439056 on Build GRCh38
rs1033465486 Chr.17: 35438885-35438885 on Build GRCh38
rs1042120744 Chr.17: 35439054-35439054 on Build GRCh38
rs56188402 Chr.17: 35439064-35439065 on Build GRCh38
rs775788050 Chr.17: 35438906-35438906 on Build GRCh38
rs911379682 Chr.17: 35439057-35439058 on Build GRCh38
rs374984018 Chr.17: 35439057-35439058 on Build GRCh38
rs62078109 Chr.17: 35438894-35438894 on Build GRCh38
rs1024860203 Chr.17: 35438940-35438940 on Build GRCh38
rs556069943 Chr.17: 35439112-35439112 on Build GRCh38
rs985663293 Chr.17: 35439057-35439057 on Build GRCh38
rs201522840 Chr.17: 35439048-35439049 on Build GRCh38
rs1026646055 Chr.17: 35439276-35439276 on Build GRCh38
rs547130529 Chr.17: 35439058-35439058 on Build GRCh38
rs560678436 Chr.17: 35438922-35438922 on Build GRCh38
rs763156943 Chr.17: 35439157-35439157 on Build GRCh38
rs913502741 Chr.17: 35439334-35439334 on Build GRCh38
rs561232310 Chr.17: 35438858-35438858 on Build GRCh38
rs574846761 Chr.17: 35439313-35439313 on Build GRCh38
rs113520563 Chr.17: 35439303-35439303 on Build GRCh38
rs961301165 Chr.17: 35439219-35439219 on Build GRCh38
rs190321145 Chr.17: 35438862-35438862 on Build GRCh38
rs181504753 Chr.17: 35439068-35439068 on Build GRCh38
rs894980642 Chr.17: 35439082-35439082 on Build GRCh38
rs147257183 Chr.17: 35438959-35438959 on Build GRCh38
rs548187940 Chr.17: 35438997-35438997 on Build GRCh38
rs186934797 Chr.17: 35439318-35439318 on Build GRCh38
rs1025331415 Chr.17: 35439180-35439180 on Build GRCh38
rs931039137 Chr.17: 35439056-35439057 on Build GRCh38
rs577814980 Chr.17: 35438936-35438936 on Build GRCh38
rs773832419 Chr.17: 35439126-35439126 on Build GRCh38
rs1041704704 Chr.17: 35439063-35439063 on Build GRCh38
rs761000416 Chr.17: 35438992-35438992 on Build GRCh38
rs1043024936 Chr.17: 35439064-35439064 on Build GRCh38
rs993865145 Chr.17: 35439242-35439242 on Build GRCh38
rs868454693 Chr.17: 35439131-35439131 on Build GRCh38
rs985213486 Chr.17: 35439298-35439298 on Build GRCh38
rs562325700 Chr.17: 35439065-35439065 on Build GRCh38
rs781096456 Chr.17: 35439065-35439066 on Build GRCh38
rs201240520 Chr.17: 35439053-35439054 on Build GRCh38
rs952316305 Chr.17: 35439288-35439288 on Build GRCh38
rs368605250 Chr.17: 35439055-35439056 on Build GRCh38
rs57016744 Chr.17: 35439055-35439055 on Build GRCh38
rs55879909 Chr.17: 35439229-35439229 on Build GRCh38
rs766748246 Chr.17: 35439165-35439165 on Build GRCh38
rs116406202 Chr.17: 35439152-35439152 on Build GRCh38
rs558349546 Chr.17: 35438932-35438932 on Build GRCh38
rs117907611 Chr.17: 35438972-35438972 on Build GRCh38
rs79448911 Chr.17: 35439021-35439021 on Build GRCh38
rs373621022 Chr.17: 35439325-35439325 on Build GRCh38
rs114314599 Chr.17: 35438937-35438937 on Build GRCh38

View Full Product Details