Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 106424158-106424705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGPD3
Entrez Gene ID
653489
Gene Name
RANBP2-like and GRIP domain containing 3
Gene Aliases
RGP3
UniGene
Hs.726367
Gene Chromosome Location
Chr.2: 106404680-106470935 on Build GRCh38
Transcripts
NM_001144013.1 XM_017004738.1 XM_017004739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3564748 c.3262C>T p.L1088F Chr.2: 106424705-106424705 on Build GRCh38
3682204 c.3600T>G p.S1200R Chr.2: 106424367-106424367 on Build GRCh38
1326021 c.3782A>C p.D1261A Chr.2: 106424185-106424185 on Build GRCh38
7003287 c.3680G>A p.G1227D Chr.2: 106424287-106424287 on Build GRCh38
5780533 c.3599G>C p.S1200T Chr.2: 106424368-106424368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968504501 Chr.2: 106424467-106424467 on Build GRCh38
rs772947675 Chr.2: 106424297-106424297 on Build GRCh38
rs749158787 Chr.2: 106424285-106424285 on Build GRCh38
rs977620272 Chr.2: 106424250-106424250 on Build GRCh38
rs947122381 Chr.2: 106424249-106424249 on Build GRCh38
rs770745683 Chr.2: 106424513-106424513 on Build GRCh38
rs776902904 Chr.2: 106424651-106424651 on Build GRCh38
rs771232261 Chr.2: 106424611-106424611 on Build GRCh38
rs773882844 Chr.2: 106424222-106424222 on Build GRCh38
rs781240629 Chr.2: 106424279-106424279 on Build GRCh38
rs199659401 Chr.2: 106424318-106424318 on Build GRCh38
rs924792528 Chr.2: 106424499-106424499 on Build GRCh38
rs879214301 Chr.2: 106424474-106424474 on Build GRCh38
rs757130884 Chr.2: 106424625-106424625 on Build GRCh38
rs938928799 Chr.2: 106424284-106424284 on Build GRCh38
rs763231921 Chr.2: 106424352-106424353 on Build GRCh38
rs769237333 Chr.2: 106424444-106424444 on Build GRCh38
rs758402856 Chr.2: 106424501-106424501 on Build GRCh38
rs762760612 Chr.2: 106424213-106424213 on Build GRCh38
rs748763147 Chr.2: 106424594-106424594 on Build GRCh38
rs771703051 Chr.2: 106424370-106424370 on Build GRCh38
rs778919455 Chr.2: 106424479-106424479 on Build GRCh38
rs551764519 Chr.2: 106424600-106424600 on Build GRCh38
rs549192084 Chr.2: 106424628-106424628 on Build GRCh38
rs775367447 Chr.2: 106424204-106424204 on Build GRCh38
rs773000969 Chr.2: 106424387-106424387 on Build GRCh38
rs560165842 Chr.2: 106424526-106424526 on Build GRCh38
rs774558910 Chr.2: 106424670-106424670 on Build GRCh38
rs995388884 Chr.2: 106424194-106424194 on Build GRCh38
rs754996212 Chr.2: 106424334-106424334 on Build GRCh38
rs770692318 Chr.2: 106424428-106424428 on Build GRCh38
rs193124838 Chr.2: 106424507-106424507 on Build GRCh38
rs775483001 Chr.2: 106424618-106424618 on Build GRCh38
rs11885810 Chr.2: 106424529-106424529 on Build GRCh38
rs765297608 Chr.2: 106424556-106424556 on Build GRCh38
rs780911717 Chr.2: 106424576-106424576 on Build GRCh38
rs753194149 Chr.2: 106424246-106424246 on Build GRCh38
rs750784264 Chr.2: 106424461-106424461 on Build GRCh38
rs762296037 Chr.2: 106424530-106424530 on Build GRCh38
rs534325470 Chr.2: 106424623-106424623 on Build GRCh38
rs755759956 Chr.2: 106424640-106424640 on Build GRCh38
rs763129967 Chr.2: 106424660-106424660 on Build GRCh38
rs750586738 Chr.2: 106424322-106424322 on Build GRCh38
rs751847761 Chr.2: 106424657-106424658 on Build GRCh38
rs776274966 Chr.2: 106424301-106424301 on Build GRCh38
rs577262447 Chr.2: 106424280-106424280 on Build GRCh38
rs765858198 Chr.2: 106424244-106424244 on Build GRCh38
rs745406735 Chr.2: 106424518-106424518 on Build GRCh38
rs751855732 Chr.2: 106424256-106424256 on Build GRCh38
rs986649203 Chr.2: 106424207-106424207 on Build GRCh38
rs766568633 Chr.2: 106424468-106424468 on Build GRCh38
rs200799525 Chr.2: 106424531-106424531 on Build GRCh38
rs772910932 Chr.2: 106424234-106424234 on Build GRCh38
rs760272452 Chr.2: 106424298-106424298 on Build GRCh38
rs767066922 Chr.2: 106424225-106424225 on Build GRCh38
rs747876379 Chr.2: 106424295-106424295 on Build GRCh38
rs776328302 Chr.2: 106424432-106424432 on Build GRCh38
rs778270432 Chr.2: 106424648-106424648 on Build GRCh38
rs777044802 Chr.2: 106424614-106424614 on Build GRCh38
rs147653884 Chr.2: 106424339-106424339 on Build GRCh38
rs780951637 Chr.2: 106424515-106424515 on Build GRCh38
rs764211343 Chr.2: 106424669-106424669 on Build GRCh38
rs746722528 Chr.2: 106424392-106424392 on Build GRCh38
rs759747010 Chr.2: 106424616-106424616 on Build GRCh38
rs1037711653 Chr.2: 106424535-106424536 on Build GRCh38
rs559852822 Chr.2: 106424462-106424462 on Build GRCh38
rs764583955 Chr.2: 106424254-106424254 on Build GRCh38
rs758937710 Chr.2: 106424436-106424436 on Build GRCh38
rs773618367 Chr.2: 106424541-106424541 on Build GRCh38
rs761000929 Chr.2: 106424545-106424545 on Build GRCh38
rs959772064 Chr.2: 106424665-106424665 on Build GRCh38
rs757523584 Chr.2: 106424321-106424321 on Build GRCh38
rs760219195 Chr.2: 106424235-106424235 on Build GRCh38
rs766568934 Chr.2: 106424426-106424427 on Build GRCh38
rs565175357 Chr.2: 106424575-106424575 on Build GRCh38
rs769762999 Chr.2: 106424197-106424197 on Build GRCh38
rs750795403 Chr.2: 106424346-106424346 on Build GRCh38
rs753994280 Chr.2: 106424473-106424473 on Build GRCh38
rs779494469 Chr.2: 106424586-106424586 on Build GRCh38
rs544989584 Chr.2: 106424307-106424307 on Build GRCh38
rs747994044 Chr.2: 106424357-106424357 on Build GRCh38
rs754441240 Chr.2: 106424646-106424646 on Build GRCh38
rs769129506 Chr.2: 106424525-106424525 on Build GRCh38
rs769472604 Chr.2: 106424581-106424581 on Build GRCh38
rs757546980 Chr.2: 106424261-106424261 on Build GRCh38
rs567557657 Chr.2: 106424631-106424631 on Build GRCh38
rs758457767 Chr.2: 106424565-106424565 on Build GRCh38
rs566720639 Chr.2: 106424622-106424622 on Build GRCh38
rs779731415 Chr.2: 106424645-106424645 on Build GRCh38
rs756139387 Chr.2: 106424282-106424282 on Build GRCh38
rs185260298 Chr.2: 106424654-106424654 on Build GRCh38
rs765918840 Chr.2: 106424300-106424300 on Build GRCh38
rs532259308 Chr.2: 106424598-106424598 on Build GRCh38
rs267598824 Chr.2: 106424333-106424333 on Build GRCh38
rs775741965 Chr.2: 106424657-106424657 on Build GRCh38
rs768442396 Chr.2: 106424214-106424214 on Build GRCh38
rs764486101 Chr.2: 106424319-106424319 on Build GRCh38
rs1021668971 Chr.2: 106424466-106424466 on Build GRCh38
rs886916658 Chr.2: 106424431-106424431 on Build GRCh38
rs761596526 Chr.2: 106424223-106424223 on Build GRCh38
rs768475707 Chr.2: 106424287-106424287 on Build GRCh38
rs866504271 Chr.2: 106424629-106424629 on Build GRCh38
rs747401395 Chr.2: 106424601-106424601 on Build GRCh38
rs750231046 Chr.2: 106424681-106424681 on Build GRCh38
rs766623680 Chr.2: 106424546-106424546 on Build GRCh38
rs767456613 Chr.2: 106424673-106424673 on Build GRCh38
rs1025443337 Chr.2: 106424402-106424402 on Build GRCh38
rs779981715 Chr.2: 106424283-106424283 on Build GRCh38
rs754977024 Chr.2: 106424476-106424476 on Build GRCh38
rs774263963 Chr.2: 106424288-106424288 on Build GRCh38
rs765613607 Chr.2: 106424211-106424212 on Build GRCh38
rs751430277 Chr.2: 106424572-106424572 on Build GRCh38
rs774672837 Chr.2: 106424447-106424447 on Build GRCh38
rs550140559 Chr.2: 106424550-106424550 on Build GRCh38
rs951307012 Chr.2: 106424201-106424201 on Build GRCh38
rs781389779 Chr.2: 106424181-106424181 on Build GRCh38
rs771468333 Chr.2: 106424650-106424650 on Build GRCh38
rs1031453644 Chr.2: 106424662-106424662 on Build GRCh38
rs767804965 Chr.2: 106424460-106424460 on Build GRCh38
rs998504037 Chr.2: 106424639-106424639 on Build GRCh38
rs765469656 Chr.2: 106424617-106424617 on Build GRCh38
rs780177331 Chr.2: 106424327-106424327 on Build GRCh38
rs777821145 Chr.2: 106424568-106424568 on Build GRCh38
rs752507647 Chr.2: 106424482-106424482 on Build GRCh38
rs572121110 Chr.2: 106424505-106424505 on Build GRCh38
rs556428798 Chr.2: 106424212-106424212 on Build GRCh38
rs745452977 Chr.2: 106424579-106424579 on Build GRCh38
rs758691149 Chr.2: 106424251-106424251 on Build GRCh38
rs745926457 Chr.2: 106424189-106424189 on Build GRCh38
rs761907988 Chr.2: 106424672-106424672 on Build GRCh38
rs752806630 Chr.2: 106424561-106424561 on Build GRCh38

View Full Product Details