Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32153913-32154421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507547
Entrez Gene ID
100507547
Gene Name
uncharacterized LOC100507547
Gene Aliases
-
UniGene
Hs.549204
Gene Chromosome Location
Chr.6: 32152802-32154365 on Build GRCh38
Transcripts
NR_037169.1 NR_037170.1 NR_037171.1 NR_037172.1 NM_001204103.1 NM_005155.6 NM_138717.2 NR_037861.1

Gene Symbol
PPT2
Entrez Gene ID
9374
Gene Name
palmitoyl-protein thioesterase 2
Gene Aliases
C6orf8, G14, PPT-2
UniGene
Hs.332138
Gene Chromosome Location
Chr.6: 32153452-32163681 on Build GRCh38
Transcripts
NR_037169.1 NR_037170.1 NR_037171.1 NR_037172.1 NM_001204103.1 NM_005155.6 NM_138717.2 NR_037861.1

Gene Symbol
PPT2-EGFL8
Entrez Gene ID
100532746
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.731888
Gene Chromosome Location
Chr.6: 32153999-32168285 on Build GRCh38
Transcripts
NR_037169.1 NR_037170.1 NR_037171.1 NR_037172.1 NM_001204103.1 NM_005155.6 NM_138717.2 NR_037861.1

SNP Detail

SNP ID Chromosome Location
rs563097230 Chr.6: 32154111-32154111 on Build GRCh38
rs572279796 Chr.6: 32154194-32154194 on Build GRCh38
rs1009649951 Chr.6: 32154293-32154293 on Build GRCh38
rs766038131 Chr.6: 32154363-32154363 on Build GRCh38
rs973946094 Chr.6: 32154336-32154336 on Build GRCh38
rs890257143 Chr.6: 32154362-32154362 on Build GRCh38
rs1047046287 Chr.6: 32154118-32154118 on Build GRCh38
rs746503404 Chr.6: 32154092-32154092 on Build GRCh38
rs545934587 Chr.6: 32154232-32154232 on Build GRCh38
rs772690837 Chr.6: 32154066-32154066 on Build GRCh38
rs983061853 Chr.6: 32154239-32154239 on Build GRCh38
rs1009198057 Chr.6: 32153997-32153997 on Build GRCh38
rs1030863133 Chr.6: 32154157-32154157 on Build GRCh38
rs186990718 Chr.6: 32154084-32154084 on Build GRCh38
rs553025838 Chr.6: 32154032-32154032 on Build GRCh38
rs908707642 Chr.6: 32154091-32154091 on Build GRCh38
rs904150300 Chr.6: 32154043-32154043 on Build GRCh38
rs1053774897 Chr.6: 32154259-32154259 on Build GRCh38
rs574640895 Chr.6: 32154077-32154077 on Build GRCh38
rs938293093 Chr.6: 32154237-32154237 on Build GRCh38
rs561545255 Chr.6: 32154153-32154153 on Build GRCh38
rs896665278 Chr.6: 32153994-32153994 on Build GRCh38
rs534588584 Chr.6: 32154029-32154029 on Build GRCh38
rs758942191 Chr.6: 32153996-32153996 on Build GRCh38
rs983202708 Chr.6: 32154087-32154087 on Build GRCh38
rs893852599 Chr.6: 32154286-32154286 on Build GRCh38
rs505997 Chr.6: 32154155-32154155 on Build GRCh38
rs920287625 Chr.6: 32154054-32154054 on Build GRCh38
rs35111071 Chr.6: 32154121-32154122 on Build GRCh38
rs1042540619 Chr.6: 32154008-32154008 on Build GRCh38
rs146938399 Chr.6: 32154398-32154398 on Build GRCh38
rs1017526212 Chr.6: 32154001-32154001 on Build GRCh38
rs964705970 Chr.6: 32154027-32154027 on Build GRCh38
rs142636557 Chr.6: 32154330-32154330 on Build GRCh38

View Full Product Details