Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 66926923-66927493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA3
Entrez Gene ID
29119
Gene Name
catenin alpha 3
Gene Aliases
ARVD13, VR22
UniGene
Hs.21375
Gene Chromosome Location
Chr.10: 65912518-67696217 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 NR_111911.1 NM_178011.4

Gene Symbol
LOC101928961
Entrez Gene ID
101928961
Gene Name
uncharacterized LOC101928961
Gene Aliases
-
UniGene
Hs.687014
Gene Chromosome Location
Chr.10: 66894110-67012153 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 NR_111911.1 NM_178011.4

Gene Symbol
LRRTM3
Entrez Gene ID
347731
Gene Name
leucine rich repeat transmembrane neuronal 3
Gene Aliases
-
UniGene
Hs.652155
Gene Chromosome Location
Chr.10: 66926034-67101551 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 NR_111911.1 NM_178011.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4699331 c.69A>G p.L23L Chr.10: 66926985-66926985 on Build GRCh38
4551279 c.524G>A p.R175Q Chr.10: 66927440-66927440 on Build GRCh38
386281 c.518C>G p.P173R Chr.10: 66927434-66927434 on Build GRCh38
3439669 c.504C>T p.S168S Chr.10: 66927420-66927420 on Build GRCh38
3439664 c.394G>C p.V132L Chr.10: 66927310-66927310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186023766 Chr.10: 66926991-66926991 on Build GRCh38
rs760549306 Chr.10: 66926956-66926956 on Build GRCh38
rs746052416 Chr.10: 66927465-66927465 on Build GRCh38
rs749342450 Chr.10: 66927143-66927156 on Build GRCh38
rs868783072 Chr.10: 66927118-66927118 on Build GRCh38
rs764904513 Chr.10: 66927190-66927190 on Build GRCh38
rs749668716 Chr.10: 66927250-66927250 on Build GRCh38
rs145390422 Chr.10: 66927348-66927348 on Build GRCh38
rs761639638 Chr.10: 66927164-66927164 on Build GRCh38
rs774186675 Chr.10: 66927299-66927299 on Build GRCh38
rs778228696 Chr.10: 66927020-66927020 on Build GRCh38
rs953972593 Chr.10: 66927071-66927071 on Build GRCh38
rs931079500 Chr.10: 66927307-66927307 on Build GRCh38
rs267602549 Chr.10: 66927201-66927201 on Build GRCh38
rs757671784 Chr.10: 66927429-66927429 on Build GRCh38
rs770093765 Chr.10: 66926970-66926970 on Build GRCh38
rs750085717 Chr.10: 66926979-66926979 on Build GRCh38
rs1023288860 Chr.10: 66927381-66927381 on Build GRCh38
rs754583859 Chr.10: 66927215-66927215 on Build GRCh38
rs766106275 Chr.10: 66927284-66927284 on Build GRCh38
rs147373916 Chr.10: 66927334-66927334 on Build GRCh38
rs779079910 Chr.10: 66927447-66927447 on Build GRCh38
rs775674319 Chr.10: 66927266-66927266 on Build GRCh38
rs35733981 Chr.10: 66927421-66927421 on Build GRCh38
rs757467180 Chr.10: 66927251-66927251 on Build GRCh38
rs914562544 Chr.10: 66927006-66927006 on Build GRCh38
rs777943604 Chr.10: 66927244-66927244 on Build GRCh38
rs755532610 Chr.10: 66927007-66927007 on Build GRCh38
rs770877473 Chr.10: 66926952-66926952 on Build GRCh38
rs143239282 Chr.10: 66927242-66927242 on Build GRCh38
rs745930710 Chr.10: 66927255-66927255 on Build GRCh38
rs867922666 Chr.10: 66927058-66927058 on Build GRCh38
rs769772215 Chr.10: 66927272-66927272 on Build GRCh38
rs777020539 Chr.10: 66927207-66927207 on Build GRCh38
rs764729741 Chr.10: 66927378-66927378 on Build GRCh38
rs750242654 Chr.10: 66927198-66927198 on Build GRCh38
rs267602548 Chr.10: 66927011-66927011 on Build GRCh38
rs372182192 Chr.10: 66927010-66927010 on Build GRCh38
rs985751184 Chr.10: 66927179-66927179 on Build GRCh38
rs761574517 Chr.10: 66926976-66926976 on Build GRCh38
rs756668374 Chr.10: 66927232-66927232 on Build GRCh38
rs896448749 Chr.10: 66927264-66927264 on Build GRCh38
rs140888093 Chr.10: 66926980-66926980 on Build GRCh38
rs752265322 Chr.10: 66927356-66927356 on Build GRCh38
rs751097102 Chr.10: 66927212-66927212 on Build GRCh38
rs866105718 Chr.10: 66927154-66927154 on Build GRCh38
rs747974816 Chr.10: 66927269-66927269 on Build GRCh38
rs777864886 Chr.10: 66927147-66927147 on Build GRCh38
rs753104442 Chr.10: 66927225-66927225 on Build GRCh38
rs774437600 Chr.10: 66926953-66926953 on Build GRCh38
rs759680598 Chr.10: 66927090-66927090 on Build GRCh38
rs772875103 Chr.10: 66927273-66927273 on Build GRCh38
rs756513160 Chr.10: 66927375-66927375 on Build GRCh38
rs758644488 Chr.10: 66927469-66927469 on Build GRCh38
rs751115382 Chr.10: 66927003-66927003 on Build GRCh38
rs199858566 Chr.10: 66927384-66927384 on Build GRCh38
rs753630024 Chr.10: 66926971-66926971 on Build GRCh38
rs764024024 Chr.10: 66926965-66926965 on Build GRCh38
rs764923660 Chr.10: 66926977-66926977 on Build GRCh38
rs768106538 Chr.10: 66927220-66927220 on Build GRCh38
rs947475780 Chr.10: 66927136-66927136 on Build GRCh38
rs762861474 Chr.10: 66927278-66927278 on Build GRCh38
rs771156957 Chr.10: 66927041-66927041 on Build GRCh38
rs779168222 Chr.10: 66927253-66927253 on Build GRCh38
rs772182776 Chr.10: 66927258-66927258 on Build GRCh38
rs747469235 Chr.10: 66927156-66927156 on Build GRCh38
rs774347675 Chr.10: 66927057-66927057 on Build GRCh38
rs377040735 Chr.10: 66927025-66927025 on Build GRCh38

View Full Product Details