Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18103034-18103422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAAL1
Entrez Gene ID
113174
Gene Name
serum amyloid A like 1
Gene Aliases
SPACIA1
UniGene
Hs.591998
Gene Chromosome Location
Chr.11: 18080343-18106091 on Build GRCh38
Transcripts
NM_138421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3979466 c.249+5G>C p.? Chr.11: 18103228-18103228 on Build GRCh38
428772 c.136-1G>A p.? Chr.11: 18103347-18103347 on Build GRCh38
7169446 c.190G>A p.E64K Chr.11: 18103292-18103292 on Build GRCh38
1604393 c.167C>T p.S56L Chr.11: 18103315-18103315 on Build GRCh38
4750622 c.241A>C p.M81L Chr.11: 18103241-18103241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778043764 Chr.11: 18103263-18103263 on Build GRCh38
rs762939011 Chr.11: 18103137-18103137 on Build GRCh38
rs772230249 Chr.11: 18103294-18103294 on Build GRCh38
rs1012280107 Chr.11: 18103302-18103302 on Build GRCh38
rs1003021433 Chr.11: 18103283-18103283 on Build GRCh38
rs200368177 Chr.11: 18103285-18103285 on Build GRCh38
rs1041908373 Chr.11: 18103077-18103077 on Build GRCh38
rs773784815 Chr.11: 18103339-18103339 on Build GRCh38
rs754628356 Chr.11: 18103254-18103254 on Build GRCh38
rs573081476 Chr.11: 18103197-18103197 on Build GRCh38
rs751077471 Chr.11: 18103218-18103218 on Build GRCh38
rs374228988 Chr.11: 18103344-18103344 on Build GRCh38
rs763052080 Chr.11: 18103332-18103332 on Build GRCh38
rs907302432 Chr.11: 18103241-18103241 on Build GRCh38
rs1040635549 Chr.11: 18103142-18103142 on Build GRCh38
rs766213349 Chr.11: 18103303-18103304 on Build GRCh38
rs752349657 Chr.11: 18103346-18103346 on Build GRCh38
rs967768435 Chr.11: 18103106-18103106 on Build GRCh38
rs921859017 Chr.11: 18103148-18103148 on Build GRCh38
rs914924844 Chr.11: 18103072-18103072 on Build GRCh38
rs754194078 Chr.11: 18103383-18103383 on Build GRCh38
rs774994464 Chr.11: 18103198-18103198 on Build GRCh38
rs747165607 Chr.11: 18103327-18103327 on Build GRCh38
rs779450846 Chr.11: 18103391-18103391 on Build GRCh38
rs560793570 Chr.11: 18103183-18103183 on Build GRCh38
rs534566929 Chr.11: 18103177-18103177 on Build GRCh38
rs575344348 Chr.11: 18103119-18103119 on Build GRCh38
rs773131857 Chr.11: 18103329-18103329 on Build GRCh38
rs947420954 Chr.11: 18103385-18103385 on Build GRCh38
rs754670380 Chr.11: 18103165-18103165 on Build GRCh38
rs773291524 Chr.11: 18103196-18103196 on Build GRCh38
rs11555876 Chr.11: 18103315-18103315 on Build GRCh38
rs1012571918 Chr.11: 18103176-18103176 on Build GRCh38
rs370854148 Chr.11: 18103256-18103256 on Build GRCh38
rs959235674 Chr.11: 18103057-18103057 on Build GRCh38
rs773896131 Chr.11: 18103297-18103297 on Build GRCh38
rs931926246 Chr.11: 18103158-18103158 on Build GRCh38
rs772461882 Chr.11: 18103258-18103258 on Build GRCh38
rs944520065 Chr.11: 18103129-18103129 on Build GRCh38
rs761484261 Chr.11: 18103328-18103328 on Build GRCh38
rs760263176 Chr.11: 18103361-18103361 on Build GRCh38
rs750969935 Chr.11: 18103146-18103146 on Build GRCh38
rs780357718 Chr.11: 18103299-18103299 on Build GRCh38
rs767178556 Chr.11: 18103255-18103255 on Build GRCh38
rs923699608 Chr.11: 18103122-18103122 on Build GRCh38
rs977740356 Chr.11: 18103352-18103352 on Build GRCh38
rs757743630 Chr.11: 18103287-18103287 on Build GRCh38
rs373896935 Chr.11: 18103338-18103338 on Build GRCh38
rs764496839 Chr.11: 18103362-18103362 on Build GRCh38
rs201028896 Chr.11: 18103240-18103240 on Build GRCh38
rs1036421818 Chr.11: 18103238-18103238 on Build GRCh38
rs566868097 Chr.11: 18103058-18103058 on Build GRCh38
rs745927479 Chr.11: 18103293-18103293 on Build GRCh38
rs750283802 Chr.11: 18103218-18103218 on Build GRCh38
rs377550968 Chr.11: 18103205-18103205 on Build GRCh38
rs34778091 Chr.11: 18103290-18103290 on Build GRCh38
rs757298196 Chr.11: 18103165-18103166 on Build GRCh38
rs1049058505 Chr.11: 18103166-18103166 on Build GRCh38
rs756110139 Chr.11: 18103277-18103277 on Build GRCh38
rs962338291 Chr.11: 18103334-18103334 on Build GRCh38
rs757742318 Chr.11: 18103384-18103384 on Build GRCh38

View Full Product Details