Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47182253-47182851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PACSIN3
Entrez Gene ID
29763
Gene Name
protein kinase C and casein kinase substrate in neurons 3
Gene Aliases
SDPIII
UniGene
Hs.334639
Gene Chromosome Location
Chr.11: 47177522-47186459 on Build GRCh38
Transcripts
NM_001184974.1 NM_001184975.1 NM_016223.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6736810 c.137G>A p.R46H Chr.11: 47182477-47182477 on Build GRCh38
5451140 c.102G>A p.R34R Chr.11: 47182512-47182512 on Build GRCh38
3752382 c.182C>T p.A61V Chr.11: 47182432-47182432 on Build GRCh38
353744 c.17A>T p.D6V Chr.11: 47182711-47182711 on Build GRCh38
3447753 c.173C>T p.A58V Chr.11: 47182441-47182441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766449112 Chr.11: 47182551-47182551 on Build GRCh38
rs190309139 Chr.11: 47182620-47182620 on Build GRCh38
rs771522813 Chr.11: 47182735-47182735 on Build GRCh38
rs552022077 Chr.11: 47182762-47182762 on Build GRCh38
rs10838664 Chr.11: 47182353-47182353 on Build GRCh38
rs2279438 Chr.11: 47182624-47182624 on Build GRCh38
rs142191658 Chr.11: 47182544-47182544 on Build GRCh38
rs751462315 Chr.11: 47182388-47182388 on Build GRCh38
rs745589644 Chr.11: 47182506-47182506 on Build GRCh38
rs374790048 Chr.11: 47182703-47182703 on Build GRCh38
rs940236526 Chr.11: 47182616-47182616 on Build GRCh38
rs548697444 Chr.11: 47182736-47182736 on Build GRCh38
rs748580483 Chr.11: 47182513-47182513 on Build GRCh38
rs913574628 Chr.11: 47182333-47182333 on Build GRCh38
rs7106654 Chr.11: 47182432-47182432 on Build GRCh38
rs199642944 Chr.11: 47182571-47182571 on Build GRCh38
rs749992641 Chr.11: 47182380-47182380 on Build GRCh38
rs765389474 Chr.11: 47182472-47182490 on Build GRCh38
rs1016950346 Chr.11: 47182800-47182800 on Build GRCh38
rs767734028 Chr.11: 47182688-47182688 on Build GRCh38
rs758594991 Chr.11: 47182548-47182548 on Build GRCh38
rs574904901 Chr.11: 47182514-47182514 on Build GRCh38
rs1053574727 Chr.11: 47182502-47182502 on Build GRCh38
rs775420412 Chr.11: 47182539-47182539 on Build GRCh38
rs987784283 Chr.11: 47182328-47182328 on Build GRCh38
rs769326171 Chr.11: 47182505-47182505 on Build GRCh38
rs767680885 Chr.11: 47182756-47182756 on Build GRCh38
rs778197957 Chr.11: 47182710-47182710 on Build GRCh38
rs1021252373 Chr.11: 47182342-47182342 on Build GRCh38
rs113909496 Chr.11: 47182724-47182724 on Build GRCh38
rs748666144 Chr.11: 47182576-47182576 on Build GRCh38
rs147642333 Chr.11: 47182429-47182429 on Build GRCh38
rs778963549 Chr.11: 47182408-47182408 on Build GRCh38
rs968160177 Chr.11: 47182533-47182533 on Build GRCh38
rs776472739 Chr.11: 47182744-47182744 on Build GRCh38
rs760392311 Chr.11: 47182665-47182665 on Build GRCh38
rs763827628 Chr.11: 47182543-47182543 on Build GRCh38
rs376356554 Chr.11: 47182531-47182531 on Build GRCh38
rs766567091 Chr.11: 47182662-47182662 on Build GRCh38
rs374438423 Chr.11: 47182306-47182306 on Build GRCh38
rs201913511 Chr.11: 47182508-47182508 on Build GRCh38
rs901380163 Chr.11: 47182717-47182717 on Build GRCh38
rs757657415 Chr.11: 47182719-47182719 on Build GRCh38
rs146360819 Chr.11: 47182709-47182709 on Build GRCh38
rs568596069 Chr.11: 47182763-47182763 on Build GRCh38
rs1000913019 Chr.11: 47182796-47182796 on Build GRCh38
rs370359394 Chr.11: 47182399-47182399 on Build GRCh38
rs140875107 Chr.11: 47182413-47182413 on Build GRCh38
rs759857440 Chr.11: 47182755-47182755 on Build GRCh38
rs757461009 Chr.11: 47182477-47182477 on Build GRCh38
rs185936979 Chr.11: 47182532-47182532 on Build GRCh38
rs543895879 Chr.11: 47182524-47182524 on Build GRCh38
rs889946834 Chr.11: 47182761-47182761 on Build GRCh38
rs370056659 Chr.11: 47182671-47182671 on Build GRCh38
rs370811952 Chr.11: 47182754-47182754 on Build GRCh38
rs776888907 Chr.11: 47182369-47182370 on Build GRCh38
rs138966432 Chr.11: 47182503-47182503 on Build GRCh38
rs755528299 Chr.11: 47182567-47182567 on Build GRCh38
rs374357570 Chr.11: 47182772-47182772 on Build GRCh38
rs770454820 Chr.11: 47182521-47182521 on Build GRCh38
rs769631894 Chr.11: 47182750-47182750 on Build GRCh38
rs754733654 Chr.11: 47182397-47182397 on Build GRCh38
rs760396236 Chr.11: 47182540-47182540 on Build GRCh38
rs548709548 Chr.11: 47182436-47182436 on Build GRCh38
rs768541121 Chr.11: 47182743-47182743 on Build GRCh38
rs982314907 Chr.11: 47182595-47182595 on Build GRCh38
rs759468371 Chr.11: 47182530-47182530 on Build GRCh38
rs961455078 Chr.11: 47182801-47182801 on Build GRCh38
rs926397286 Chr.11: 47182545-47182545 on Build GRCh38
rs1017002602 Chr.11: 47182557-47182557 on Build GRCh38
rs763161975 Chr.11: 47182659-47182660 on Build GRCh38
rs367640067 Chr.11: 47182421-47182421 on Build GRCh38
rs377081778 Chr.11: 47182638-47182638 on Build GRCh38
rs779049324 Chr.11: 47182478-47182478 on Build GRCh38
rs138367699 Chr.11: 47182472-47182472 on Build GRCh38
rs770560316 Chr.11: 47182611-47182611 on Build GRCh38
rs200483884 Chr.11: 47182412-47182412 on Build GRCh38
rs529834327 Chr.11: 47182806-47182806 on Build GRCh38
rs367920822 Chr.11: 47182737-47182737 on Build GRCh38
rs752893660 Chr.11: 47182757-47182757 on Build GRCh38
rs775711188 Chr.11: 47182596-47182596 on Build GRCh38
rs1042081287 Chr.11: 47182713-47182713 on Build GRCh38
rs148855213 Chr.11: 47182476-47182476 on Build GRCh38
rs562829446 Chr.11: 47182659-47182659 on Build GRCh38
rs954161673 Chr.11: 47182384-47182384 on Build GRCh38
rs764451474 Chr.11: 47182766-47182766 on Build GRCh38
rs755038583 Chr.11: 47182686-47182686 on Build GRCh38
rs1009821104 Chr.11: 47182332-47182332 on Build GRCh38
rs765808197 Chr.11: 47182467-47182467 on Build GRCh38
rs772579039 Chr.11: 47182452-47182452 on Build GRCh38
rs558482473 Chr.11: 47182511-47182511 on Build GRCh38
rs762462847 Chr.11: 47182466-47182466 on Build GRCh38
rs780545729 Chr.11: 47182420-47182420 on Build GRCh38
rs532809427 Chr.11: 47182626-47182626 on Build GRCh38
rs759292322 Chr.11: 47182487-47182487 on Build GRCh38
rs568927214 Chr.11: 47182819-47182819 on Build GRCh38
rs889851202 Chr.11: 47182687-47182687 on Build GRCh38
rs538102995 Chr.11: 47182827-47182827 on Build GRCh38
rs145307845 Chr.11: 47182484-47182484 on Build GRCh38
rs768667036 Chr.11: 47182813-47182813 on Build GRCh38
rs766393865 Chr.11: 47182386-47182386 on Build GRCh38
rs376134100 Chr.11: 47182352-47182352 on Build GRCh38
rs749676389 Chr.11: 47182712-47182712 on Build GRCh38
rs567457200 Chr.11: 47182471-47182471 on Build GRCh38
rs780022026 Chr.11: 47182494-47182494 on Build GRCh38
rs919199703 Chr.11: 47182430-47182430 on Build GRCh38
rs745552400 Chr.11: 47182728-47182728 on Build GRCh38
rs756503952 Chr.11: 47182577-47182577 on Build GRCh38
rs759521291 Chr.11: 47182473-47182473 on Build GRCh38
rs375932112 Chr.11: 47182465-47182465 on Build GRCh38
rs371359220 Chr.11: 47182599-47182599 on Build GRCh38
rs965684269 Chr.11: 47182315-47182315 on Build GRCh38
rs374744200 Chr.11: 47182400-47182400 on Build GRCh38
rs772851730 Chr.11: 47182517-47182517 on Build GRCh38
rs530271460 Chr.11: 47182294-47182294 on Build GRCh38
rs199499148 Chr.11: 47182475-47182475 on Build GRCh38
rs776329510 Chr.11: 47182650-47182650 on Build GRCh38
rs762541531 Chr.11: 47182520-47182520 on Build GRCh38
rs145192231 Chr.11: 47182301-47182301 on Build GRCh38
rs778019388 Chr.11: 47182579-47182579 on Build GRCh38
rs760629311 Chr.11: 47182629-47182629 on Build GRCh38
rs377746852 Chr.11: 47182588-47182588 on Build GRCh38
rs372225317 Chr.11: 47182425-47182425 on Build GRCh38
rs769637513 Chr.11: 47182445-47182445 on Build GRCh38
rs761457847 Chr.11: 47182747-47182747 on Build GRCh38
rs762792007 Chr.11: 47182382-47182382 on Build GRCh38
rs373474939 Chr.11: 47182509-47182509 on Build GRCh38
rs759893619 Chr.11: 47182394-47182395 on Build GRCh38
rs751656794 Chr.11: 47182553-47182553 on Build GRCh38

View Full Product Details