Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 115175159-115175743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM1
Entrez Gene ID
23705
Gene Name
cell adhesion molecule 1
Gene Aliases
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
UniGene
Hs.370510
Gene Chromosome Location
Chr.11: 115173625-115504523 on Build GRCh38
Transcripts
NM_001098517.1 NM_001301043.1 NM_001301044.1 NM_001301045.1 NM_014333.3 XM_005271494.2 XM_017017457.1 XM_017017458.1 XM_017017459.1 XM_017017460.1 XM_017017461.1

SNP Detail

SNP ID Chromosome Location
rs753069422 Chr.11: 115175480-115175480 on Build GRCh38
rs778510591 Chr.11: 115175590-115175590 on Build GRCh38
rs889645275 Chr.11: 115175618-115175618 on Build GRCh38
rs940501632 Chr.11: 115175670-115175670 on Build GRCh38
rs765865944 Chr.11: 115175476-115175476 on Build GRCh38
rs937227901 Chr.11: 115175199-115175199 on Build GRCh38
rs968734581 Chr.11: 115175377-115175377 on Build GRCh38
rs889697669 Chr.11: 115175302-115175302 on Build GRCh38
rs1036101117 Chr.11: 115175681-115175681 on Build GRCh38
rs578129155 Chr.11: 115175466-115175466 on Build GRCh38
rs1032521333 Chr.11: 115175470-115175470 on Build GRCh38
rs896912155 Chr.11: 115175230-115175230 on Build GRCh38
rs201815214 Chr.11: 115175376-115175376 on Build GRCh38
rs528106440 Chr.11: 115175289-115175289 on Build GRCh38
rs796751592 Chr.11: 115175376-115175376 on Build GRCh38
rs1032098832 Chr.11: 115175662-115175662 on Build GRCh38
rs530293329 Chr.11: 115175397-115175397 on Build GRCh38
rs1004590557 Chr.11: 115175306-115175306 on Build GRCh38
rs186834100 Chr.11: 115175485-115175485 on Build GRCh38
rs997580686 Chr.11: 115175416-115175416 on Build GRCh38
rs563864463 Chr.11: 115175628-115175628 on Build GRCh38
rs183214086 Chr.11: 115175477-115175477 on Build GRCh38
rs758071943 Chr.11: 115175682-115175682 on Build GRCh38
rs761316780 Chr.11: 115175425-115175425 on Build GRCh38
rs56789298 Chr.11: 115175376-115175377 on Build GRCh38
rs1006799025 Chr.11: 115175633-115175633 on Build GRCh38
rs1037125314 Chr.11: 115175525-115175525 on Build GRCh38
rs542030545 Chr.11: 115175290-115175290 on Build GRCh38
rs1036048267 Chr.11: 115175392-115175392 on Build GRCh38
rs559831323 Chr.11: 115175529-115175529 on Build GRCh38
rs917440920 Chr.11: 115175457-115175457 on Build GRCh38
rs573138060 Chr.11: 115175490-115175490 on Build GRCh38
rs1017598335 Chr.11: 115175361-115175361 on Build GRCh38
rs754447706 Chr.11: 115175587-115175587 on Build GRCh38
rs59639020 Chr.11: 115175390-115175390 on Build GRCh38
rs575111056 Chr.11: 115175547-115175547 on Build GRCh38
rs1036813716 Chr.11: 115175227-115175227 on Build GRCh38
rs1000161176 Chr.11: 115175661-115175661 on Build GRCh38
rs753318832 Chr.11: 115175492-115175492 on Build GRCh38
rs567968562 Chr.11: 115175277-115175277 on Build GRCh38
rs941003920 Chr.11: 115175519-115175519 on Build GRCh38
rs538409766 Chr.11: 115175415-115175415 on Build GRCh38
rs74721216 Chr.11: 115175426-115175426 on Build GRCh38
rs374703534 Chr.11: 115175534-115175534 on Build GRCh38
rs397975978 Chr.11: 115175390-115175391 on Build GRCh38
rs535774069 Chr.11: 115175193-115175193 on Build GRCh38
rs543972819 Chr.11: 115175595-115175595 on Build GRCh38
rs750299940 Chr.11: 115175464-115175464 on Build GRCh38
rs879187990 Chr.11: 115175376-115175377 on Build GRCh38
rs909070381 Chr.11: 115175666-115175666 on Build GRCh38
rs532868294 Chr.11: 115175649-115175649 on Build GRCh38
rs569484067 Chr.11: 115175353-115175353 on Build GRCh38
rs997988369 Chr.11: 115175235-115175235 on Build GRCh38
rs756033089 Chr.11: 115175465-115175465 on Build GRCh38
rs1043634882 Chr.11: 115175648-115175648 on Build GRCh38
rs1006848211 Chr.11: 115175314-115175314 on Build GRCh38

View Full Product Details