Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50969718-50970316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2C
Entrez Gene ID
1031
Gene Name
cyclin dependent kinase inhibitor 2C
Gene Aliases
INK4C, p18, p18-INK4C
UniGene
Hs.525324
Gene Chromosome Location
Chr.1: 50968695-50974637 on Build GRCh38
Transcripts
NM_001262.2 NM_078626.2

SNP Detail

SNP ID Chromosome Location
rs3176454 Chr.1: 50970111-50970111 on Build GRCh38
rs1014524609 Chr.1: 50969964-50969964 on Build GRCh38
rs541270521 Chr.1: 50969873-50969873 on Build GRCh38
rs891900400 Chr.1: 50970173-50970173 on Build GRCh38
rs966295937 Chr.1: 50970027-50970027 on Build GRCh38
rs973827030 Chr.1: 50969806-50969806 on Build GRCh38
rs925631614 Chr.1: 50969955-50969955 on Build GRCh38
rs544690087 Chr.1: 50969957-50969957 on Build GRCh38
rs988727932 Chr.1: 50970110-50970110 on Build GRCh38
rs143935432 Chr.1: 50970150-50970150 on Build GRCh38
rs1001966623 Chr.1: 50969884-50969884 on Build GRCh38
rs1036170489 Chr.1: 50969965-50969965 on Build GRCh38
rs895698717 Chr.1: 50969962-50969962 on Build GRCh38
rs978544072 Chr.1: 50970122-50970122 on Build GRCh38
rs945786315 Chr.1: 50969951-50969951 on Build GRCh38
rs577334571 Chr.1: 50969939-50969939 on Build GRCh38
rs754915762 Chr.1: 50969948-50969948 on Build GRCh38
rs3176453 Chr.1: 50969978-50969978 on Build GRCh38
rs897642133 Chr.1: 50969974-50969974 on Build GRCh38
rs960405826 Chr.1: 50969952-50969952 on Build GRCh38
rs1049939442 Chr.1: 50969742-50969742 on Build GRCh38
rs985589920 Chr.1: 50970133-50970133 on Build GRCh38
rs1027693628 Chr.1: 50969980-50969981 on Build GRCh38
rs944109910 Chr.1: 50969745-50969745 on Build GRCh38
rs912138791 Chr.1: 50969930-50969930 on Build GRCh38
rs1022195523 Chr.1: 50969860-50969860 on Build GRCh38
rs1043466567 Chr.1: 50970214-50970214 on Build GRCh38
rs986385700 Chr.1: 50969925-50969925 on Build GRCh38
rs953362229 Chr.1: 50969919-50969920 on Build GRCh38
rs561027536 Chr.1: 50970083-50970083 on Build GRCh38
rs947388037 Chr.1: 50970205-50970205 on Build GRCh38
rs912735603 Chr.1: 50970156-50970156 on Build GRCh38
rs571449280 Chr.1: 50970166-50970166 on Build GRCh38
rs1055447765 Chr.1: 50970179-50970179 on Build GRCh38
rs991868037 Chr.1: 50969963-50969963 on Build GRCh38
rs76136361 Chr.1: 50969938-50969938 on Build GRCh38
rs555778434 Chr.1: 50969743-50969743 on Build GRCh38
rs1055753243 Chr.1: 50969960-50969960 on Build GRCh38
rs1042813043 Chr.1: 50969954-50969954 on Build GRCh38
rs968602428 Chr.1: 50970115-50970115 on Build GRCh38
rs1019158084 Chr.1: 50970022-50970022 on Build GRCh38
rs974168506 Chr.1: 50969997-50969997 on Build GRCh38
rs76280772 Chr.1: 50969979-50969979 on Build GRCh38
rs779523467 Chr.1: 50969768-50969768 on Build GRCh38
rs548922334 Chr.1: 50970051-50970051 on Build GRCh38
rs944224549 Chr.1: 50970158-50970158 on Build GRCh38
rs895688906 Chr.1: 50969776-50969776 on Build GRCh38
rs374516538 Chr.1: 50969749-50969749 on Build GRCh38
rs41285700 Chr.1: 50970274-50970274 on Build GRCh38
rs1012021174 Chr.1: 50969813-50969813 on Build GRCh38
rs778608051 Chr.1: 50970009-50970009 on Build GRCh38
rs944877188 Chr.1: 50969932-50969932 on Build GRCh38
rs891791279 Chr.1: 50969791-50969791 on Build GRCh38
rs1053068114 Chr.1: 50969746-50969746 on Build GRCh38
rs574237089 Chr.1: 50969787-50969787 on Build GRCh38
rs551898886 Chr.1: 50970280-50970280 on Build GRCh38

View Full Product Details