Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 26922501-26923076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS5
Entrez Gene ID
11096
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 5
Gene Aliases
ADAM-TS 11, ADAM-TS 5, ADAM-TS5, ADAMTS-11, ADAMTS-5, ADAMTS11, ADMP-2
UniGene
Hs.58324
Gene Chromosome Location
Chr.21: 26917912-26967120 on Build GRCh38
Transcripts
NM_007038.4 XM_011529448.2

SNP Detail

SNP ID Chromosome Location
rs1035779750 Chr.21: 26922935-26922935 on Build GRCh38
rs765059687 Chr.21: 26922782-26922782 on Build GRCh38
rs969282633 Chr.21: 26922702-26922702 on Build GRCh38
rs563261510 Chr.21: 26922834-26922834 on Build GRCh38
rs758365481 Chr.21: 26922971-26922971 on Build GRCh38
rs56273208 Chr.21: 26922862-26922862 on Build GRCh38
rs879469996 Chr.21: 26922965-26922965 on Build GRCh38
rs916204888 Chr.21: 26922683-26922683 on Build GRCh38
rs924793482 Chr.21: 26922746-26922746 on Build GRCh38
rs191048456 Chr.21: 26922997-26922997 on Build GRCh38
rs1004264036 Chr.21: 26922926-26922926 on Build GRCh38
rs544137512 Chr.21: 26922756-26922756 on Build GRCh38
rs759247935 Chr.21: 26922724-26922724 on Build GRCh38
rs899791091 Chr.21: 26922868-26922868 on Build GRCh38
rs886952287 Chr.21: 26922909-26922909 on Build GRCh38
rs920496842 Chr.21: 26922972-26922972 on Build GRCh38
rs182881823 Chr.21: 26923021-26923021 on Build GRCh38
rs995727069 Chr.21: 26922890-26922890 on Build GRCh38
rs951517631 Chr.21: 26922613-26922613 on Build GRCh38
rs1013981714 Chr.21: 26922752-26922752 on Build GRCh38
rs1052084013 Chr.21: 26922579-26922579 on Build GRCh38
rs780084480 Chr.21: 26922741-26922744 on Build GRCh38
rs769444246 Chr.21: 26922608-26922610 on Build GRCh38
rs1043863561 Chr.21: 26922546-26922546 on Build GRCh38
rs935067582 Chr.21: 26922797-26922797 on Build GRCh38
rs991700093 Chr.21: 26922676-26922676 on Build GRCh38
rs995387561 Chr.21: 26922600-26922600 on Build GRCh38
rs1020428399 Chr.21: 26922917-26922917 on Build GRCh38
rs536905255 Chr.21: 26923040-26923040 on Build GRCh38
rs115596608 Chr.21: 26922580-26922580 on Build GRCh38
rs905493645 Chr.21: 26922790-26922790 on Build GRCh38
rs181934706 Chr.21: 26922588-26922588 on Build GRCh38
rs903634825 Chr.21: 26922554-26922554 on Build GRCh38
rs148042542 Chr.21: 26922643-26922643 on Build GRCh38
rs912192854 Chr.21: 26922817-26922817 on Build GRCh38
rs911300996 Chr.21: 26922929-26922929 on Build GRCh38
rs550614873 Chr.21: 26923042-26923042 on Build GRCh38
rs229077 Chr.21: 26923020-26923020 on Build GRCh38
rs186300576 Chr.21: 26922669-26922669 on Build GRCh38
rs1012954922 Chr.21: 26923002-26923002 on Build GRCh38
rs375030163 Chr.21: 26922915-26922915 on Build GRCh38
rs559216019 Chr.21: 26922894-26922894 on Build GRCh38
rs1039600906 Chr.21: 26922590-26922590 on Build GRCh38
rs229076 Chr.21: 26922540-26922540 on Build GRCh38
rs571261048 Chr.21: 26922583-26922584 on Build GRCh38
rs549039031 Chr.21: 26922865-26922865 on Build GRCh38
rs9979410 Chr.21: 26922633-26922633 on Build GRCh38
rs1052621937 Chr.21: 26922805-26922805 on Build GRCh38
rs1027470827 Chr.21: 26922605-26922605 on Build GRCh38
rs899675994 Chr.21: 26922591-26922591 on Build GRCh38
rs1024488073 Chr.21: 26922788-26922788 on Build GRCh38
rs1048123195 Chr.21: 26923050-26923050 on Build GRCh38
rs576366546 Chr.21: 26922576-26922576 on Build GRCh38
rs551303171 Chr.21: 26922679-26922682 on Build GRCh38
rs528612611 Chr.21: 26922730-26922730 on Build GRCh38
rs1004722248 Chr.21: 26922614-26922614 on Build GRCh38
rs942702266 Chr.21: 26922931-26922931 on Build GRCh38
rs968975330 Chr.21: 26923046-26923046 on Build GRCh38

View Full Product Details