Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25739430-25739823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGLY1
Entrez Gene ID
55768
Gene Name
N-glycanase 1
Gene Aliases
CDDG, CDG1V, PNG1, PNGase
UniGene
Hs.368960
Gene Chromosome Location
Chr.3: 25718944-25790039 on Build GRCh38
Transcripts
NM_001145293.1 NM_001145294.1 NM_001145295.1 NM_018297.3 XM_005265316.1 XM_005265317.1 XM_011533944.1 XM_011533945.2 XM_017006839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1043393 c.871C>T p.R291* Chr.3: 25739587-25739587 on Build GRCh38
4323328 c.811C>T p.L271L Chr.3: 25739647-25739647 on Build GRCh38
3592116 c.774G>T p.R258S Chr.3: 25739684-25739684 on Build GRCh38
4706829 c.709T>A p.F237I Chr.3: 25739749-25739749 on Build GRCh38
1043394 c.685C>A p.L229I Chr.3: 25739773-25739773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775976410 Chr.3: 25739718-25739718 on Build GRCh38
rs375575749 Chr.3: 25739584-25739584 on Build GRCh38
rs754797226 Chr.3: 25739770-25739770 on Build GRCh38
rs758181219 Chr.3: 25739565-25739565 on Build GRCh38
rs754806231 Chr.3: 25739670-25739670 on Build GRCh38
rs766273064 Chr.3: 25739537-25739537 on Build GRCh38
rs766628355 Chr.3: 25739594-25739594 on Build GRCh38
rs917135418 Chr.3: 25739547-25739547 on Build GRCh38
rs554875318 Chr.3: 25739520-25739520 on Build GRCh38
rs529208533 Chr.3: 25739787-25739787 on Build GRCh38
rs147368318 Chr.3: 25739620-25739620 on Build GRCh38
rs374184658 Chr.3: 25739663-25739663 on Build GRCh38
rs769396030 Chr.3: 25739693-25739693 on Build GRCh38
rs201904265 Chr.3: 25739791-25739791 on Build GRCh38
rs768233891 Chr.3: 25739579-25739579 on Build GRCh38
rs143998271 Chr.3: 25739569-25739569 on Build GRCh38
rs745717960 Chr.3: 25739560-25739560 on Build GRCh38
rs751255754 Chr.3: 25739766-25739766 on Build GRCh38
rs747727275 Chr.3: 25739687-25739687 on Build GRCh38
rs760098202 Chr.3: 25739715-25739715 on Build GRCh38
rs748897361 Chr.3: 25739697-25739697 on Build GRCh38
rs771354261 Chr.3: 25739700-25739700 on Build GRCh38
rs866736138 Chr.3: 25739758-25739758 on Build GRCh38
rs200446388 Chr.3: 25739617-25739617 on Build GRCh38
rs920352938 Chr.3: 25739621-25739621 on Build GRCh38
rs371573253 Chr.3: 25739729-25739729 on Build GRCh38
rs766201614 Chr.3: 25739754-25739754 on Build GRCh38
rs772994617 Chr.3: 25739587-25739587 on Build GRCh38
rs369388667 Chr.3: 25739797-25739797 on Build GRCh38
rs761901772 Chr.3: 25739744-25739744 on Build GRCh38
rs769627493 Chr.3: 25739586-25739586 on Build GRCh38
rs370397253 Chr.3: 25739548-25739548 on Build GRCh38
rs780077758 Chr.3: 25739567-25739567 on Build GRCh38
rs147024365 Chr.3: 25739675-25739675 on Build GRCh38
rs779666541 Chr.3: 25739664-25739664 on Build GRCh38
rs777616612 Chr.3: 25739694-25739694 on Build GRCh38
rs755705306 Chr.3: 25739793-25739793 on Build GRCh38
rs752711308 Chr.3: 25739635-25739635 on Build GRCh38
rs774509442 Chr.3: 25739708-25739708 on Build GRCh38
rs370175393 Chr.3: 25739589-25739589 on Build GRCh38
rs867899358 Chr.3: 25739681-25739681 on Build GRCh38
rs961056421 Chr.3: 25739508-25739508 on Build GRCh38
rs544014972 Chr.3: 25739500-25739500 on Build GRCh38
rs865828868 Chr.3: 25739692-25739692 on Build GRCh38
rs916085942 Chr.3: 25739628-25739628 on Build GRCh38
rs752785850 Chr.3: 25739617-25739617 on Build GRCh38
rs143353538 Chr.3: 25739762-25739762 on Build GRCh38
rs960973368 Chr.3: 25739524-25739524 on Build GRCh38
rs1005486419 Chr.3: 25739460-25739460 on Build GRCh38
rs202027326 Chr.3: 25739640-25739640 on Build GRCh38
rs754083716 Chr.3: 25739741-25739741 on Build GRCh38
rs368580768 Chr.3: 25739717-25739717 on Build GRCh38
rs199578758 Chr.3: 25739657-25739657 on Build GRCh38
rs147901411 Chr.3: 25739658-25739658 on Build GRCh38
rs200925557 Chr.3: 25739686-25739686 on Build GRCh38
rs926188075 Chr.3: 25739645-25739645 on Build GRCh38
rs775963261 Chr.3: 25739581-25739581 on Build GRCh38
rs900718702 Chr.3: 25739789-25739789 on Build GRCh38
rs764370236 Chr.3: 25739651-25739651 on Build GRCh38
rs373934325 Chr.3: 25739795-25739795 on Build GRCh38
rs756248327 Chr.3: 25739629-25739629 on Build GRCh38
rs879084076 Chr.3: 25739493-25739493 on Build GRCh38
rs1016839490 Chr.3: 25739468-25739468 on Build GRCh38
rs948858807 Chr.3: 25739629-25739629 on Build GRCh38
rs760786749 Chr.3: 25739722-25739722 on Build GRCh38
rs577284168 Chr.3: 25739526-25739529 on Build GRCh38
rs769279743 Chr.3: 25739521-25739521 on Build GRCh38

View Full Product Details