Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 76452364-76452813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP4
Entrez Gene ID
85445
Gene Name
contactin associated protein like 4
Gene Aliases
CASPR4
UniGene
Hs.461389
Gene Chromosome Location
Chr.16: 76277217-76563610 on Build GRCh38
Transcripts
NM_001322178.1 NM_001322179.1 NM_001322180.1 NM_001322181.1 NM_001322187.1 NM_001322188.1 NM_001322189.1 NM_001322190.1 NM_001322191.1 NM_033401.4 NM_138994.4 NR_136210.1 NR_136213.1 NR_136216.1 XM_011523403.2 XM_017023805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6900685 c.1033G>A p.G345S Chr.16: 76452688-76452688 on Build GRCh38
6079962 c.936C>A p.D312E Chr.16: 76452591-76452591 on Build GRCh38
3721304 c.1219C>A p.L407I Chr.16: 76452730-76452730 on Build GRCh38
4427902 c.1050G>A p.L350L Chr.16: 76452561-76452561 on Build GRCh38
4965765 c.1089T>G p.C363W Chr.16: 76452528-76452528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571073193 Chr.16: 76452568-76452568 on Build GRCh38
rs752884883 Chr.16: 76452758-76452758 on Build GRCh38
rs776963288 Chr.16: 76452547-76452547 on Build GRCh38
rs372936480 Chr.16: 76452569-76452569 on Build GRCh38
rs374318989 Chr.16: 76452764-76452764 on Build GRCh38
rs150449060 Chr.16: 76452734-76452734 on Build GRCh38
rs142063319 Chr.16: 76452688-76452688 on Build GRCh38
rs144033719 Chr.16: 76452609-76452609 on Build GRCh38
rs371586768 Chr.16: 76452536-76452536 on Build GRCh38
rs773018451 Chr.16: 76452751-76452751 on Build GRCh38
rs778902268 Chr.16: 76452619-76452619 on Build GRCh38
rs765918338 Chr.16: 76452490-76452490 on Build GRCh38
rs922168719 Chr.16: 76452676-76452676 on Build GRCh38
rs999578080 Chr.16: 76452616-76452616 on Build GRCh38
rs751486424 Chr.16: 76452658-76452658 on Build GRCh38
rs764826662 Chr.16: 76452582-76452582 on Build GRCh38
rs185992629 Chr.16: 76452744-76452744 on Build GRCh38
rs765307012 Chr.16: 76452500-76452500 on Build GRCh38
rs763011352 Chr.16: 76452648-76452648 on Build GRCh38
rs570306162 Chr.16: 76452632-76452632 on Build GRCh38
rs1022923969 Chr.16: 76452755-76452755 on Build GRCh38
rs139625830 Chr.16: 76452550-76452550 on Build GRCh38
rs918380656 Chr.16: 76452708-76452708 on Build GRCh38
rs761749846 Chr.16: 76452747-76452747 on Build GRCh38
rs35181488 Chr.16: 76452558-76452559 on Build GRCh38
rs536046377 Chr.16: 76452653-76452653 on Build GRCh38
rs747380170 Chr.16: 76452635-76452635 on Build GRCh38
rs939151498 Chr.16: 76452720-76452720 on Build GRCh38
rs771323494 Chr.16: 76452636-76452636 on Build GRCh38
rs374126037 Chr.16: 76452723-76452723 on Build GRCh38
rs541566591 Chr.16: 76452451-76452451 on Build GRCh38
rs759170338 Chr.16: 76452640-76452640 on Build GRCh38
rs759037703 Chr.16: 76452576-76452576 on Build GRCh38
rs758449417 Chr.16: 76452694-76452694 on Build GRCh38
rs765491925 Chr.16: 76452753-76452753 on Build GRCh38
rs780366753 Chr.16: 76452689-76452689 on Build GRCh38
rs767860625 Chr.16: 76452470-76452470 on Build GRCh38
rs534954437 Chr.16: 76452705-76452705 on Build GRCh38
rs144559073 Chr.16: 76452565-76452565 on Build GRCh38
rs149288291 Chr.16: 76452553-76452553 on Build GRCh38
rs555904247 Chr.16: 76452675-76452675 on Build GRCh38
rs779072224 Chr.16: 76452422-76452422 on Build GRCh38
rs759774756 Chr.16: 76452752-76452752 on Build GRCh38
rs888121820 Chr.16: 76452415-76452415 on Build GRCh38
rs758532902 Chr.16: 76452760-76452760 on Build GRCh38
rs899609739 Chr.16: 76452454-76452454 on Build GRCh38
rs772904636 Chr.16: 76452482-76452482 on Build GRCh38
rs369744076 Chr.16: 76452465-76452465 on Build GRCh38
rs776206945 Chr.16: 76452638-76452638 on Build GRCh38
rs144662940 Chr.16: 76452549-76452549 on Build GRCh38
rs765026284 Chr.16: 76452601-76452603 on Build GRCh38
rs762257554 Chr.16: 76452561-76452561 on Build GRCh38
rs767217993 Chr.16: 76452748-76452748 on Build GRCh38
rs370415071 Chr.16: 76452642-76452642 on Build GRCh38
rs777653864 Chr.16: 76452531-76452531 on Build GRCh38
rs762203484 Chr.16: 76452459-76452459 on Build GRCh38
rs932316092 Chr.16: 76452412-76452412 on Build GRCh38
rs780176495 Chr.16: 76452598-76452598 on Build GRCh38
rs146466106 Chr.16: 76452627-76452627 on Build GRCh38
rs754963944 Chr.16: 76452495-76452495 on Build GRCh38
rs142342411 Chr.16: 76452413-76452413 on Build GRCh38
rs761502729 Chr.16: 76452497-76452497 on Build GRCh38
rs751734837 Chr.16: 76452772-76452775 on Build GRCh38
rs757743283 Chr.16: 76452778-76452778 on Build GRCh38
rs1005257702 Chr.16: 76452434-76452434 on Build GRCh38
rs998332573 Chr.16: 76452455-76452455 on Build GRCh38
rs751001764 Chr.16: 76452725-76452725 on Build GRCh38
rs991667764 Chr.16: 76452738-76452738 on Build GRCh38
rs760187020 Chr.16: 76452484-76452484 on Build GRCh38
rs775104563 Chr.16: 76452643-76452643 on Build GRCh38
rs746286778 Chr.16: 76452788-76452788 on Build GRCh38
rs751266058 Chr.16: 76452589-76452589 on Build GRCh38
rs761576739 Chr.16: 76452669-76452669 on Build GRCh38
rs977707362 Chr.16: 76452540-76452540 on Build GRCh38
rs941089642 Chr.16: 76452428-76452428 on Build GRCh38
rs374394050 Chr.16: 76452520-76452520 on Build GRCh38
rs772527435 Chr.16: 76452630-76452630 on Build GRCh38
rs754084194 Chr.16: 76452603-76452603 on Build GRCh38
rs531400109 Chr.16: 76452414-76452414 on Build GRCh38
rs772435913 Chr.16: 76452567-76452567 on Build GRCh38
rs372308727 Chr.16: 76452779-76452779 on Build GRCh38
rs778182909 Chr.16: 76452700-76452700 on Build GRCh38
rs151315964 Chr.16: 76452693-76452693 on Build GRCh38
rs899462414 Chr.16: 76452663-76452663 on Build GRCh38
rs752695501 Chr.16: 76452583-76452583 on Build GRCh38
rs368744120 Chr.16: 76452777-76452777 on Build GRCh38
rs757055752 Chr.16: 76452592-76452592 on Build GRCh38
rs147873764 Chr.16: 76452575-76452575 on Build GRCh38
rs1036776208 Chr.16: 76452707-76452707 on Build GRCh38
rs28584228 Chr.16: 76452684-76452684 on Build GRCh38
rs8055272 Chr.16: 76452726-76452726 on Build GRCh38
rs1029846534 Chr.16: 76452477-76452477 on Build GRCh38
rs752679184 Chr.16: 76452514-76452514 on Build GRCh38

View Full Product Details