Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126109765-126110280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1L1
Entrez Gene ID
10840
Gene Name
aldehyde dehydrogenase 1 family member L1
Gene Aliases
10-FTHFDH, 10-fTHF, FDH, FTHFD
UniGene
Hs.434435
Gene Chromosome Location
Chr.3: 126103561-126181526 on Build GRCh38
Transcripts
NM_001270364.1 NM_001270365.1 NM_012190.3 NR_072979.1 XM_006713481.2 XM_011512355.1 XM_017005613.1 XM_017005614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1693398 c.2251C>T p.H751Y Chr.3: 126110040-126110040 on Build GRCh38
6499254 c.2232C>T p.T744T Chr.3: 126110059-126110059 on Build GRCh38
6511486 c.2208G>T p.V736V Chr.3: 126110083-126110083 on Build GRCh38
7187908 c.2324G>C p.G775A Chr.3: 126109967-126109967 on Build GRCh38
445497 c.2266G>T p.V756L Chr.3: 126110025-126110025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757355845 Chr.3: 126109924-126109924 on Build GRCh38
rs76009701 Chr.3: 126109957-126109957 on Build GRCh38
rs182288944 Chr.3: 126109823-126109823 on Build GRCh38
rs548625315 Chr.3: 126110195-126110195 on Build GRCh38
rs776035780 Chr.3: 126109966-126109966 on Build GRCh38
rs747561724 Chr.3: 126110036-126110036 on Build GRCh38
rs763483275 Chr.3: 126110058-126110058 on Build GRCh38
rs768207805 Chr.3: 126109933-126109934 on Build GRCh38
rs920941953 Chr.3: 126110226-126110226 on Build GRCh38
rs201592037 Chr.3: 126109961-126109961 on Build GRCh38
rs201903896 Chr.3: 126109980-126109980 on Build GRCh38
rs7628056 Chr.3: 126109844-126109844 on Build GRCh38
rs1035031009 Chr.3: 126110153-126110153 on Build GRCh38
rs34779203 Chr.3: 126110196-126110196 on Build GRCh38
rs559416906 Chr.3: 126110231-126110231 on Build GRCh38
rs200819795 Chr.3: 126110075-126110075 on Build GRCh38
rs141918591 Chr.3: 126110059-126110059 on Build GRCh38
rs983388038 Chr.3: 126110172-126110172 on Build GRCh38
rs563865613 Chr.3: 126109788-126109788 on Build GRCh38
rs989348574 Chr.3: 126109917-126109917 on Build GRCh38
rs200292180 Chr.3: 126109995-126109995 on Build GRCh38
rs747332355 Chr.3: 126109924-126109925 on Build GRCh38
rs199644243 Chr.3: 126110079-126110079 on Build GRCh38
rs532835714 Chr.3: 126109921-126109921 on Build GRCh38
rs369739118 Chr.3: 126109920-126109920 on Build GRCh38
rs764165796 Chr.3: 126109969-126109969 on Build GRCh38
rs771587048 Chr.3: 126110047-126110047 on Build GRCh38
rs767684111 Chr.3: 126109985-126109985 on Build GRCh38
rs542892028 Chr.3: 126110203-126110203 on Build GRCh38
rs770533321 Chr.3: 126110138-126110138 on Build GRCh38
rs940958546 Chr.3: 126110184-126110184 on Build GRCh38
rs780316276 Chr.3: 126109899-126109899 on Build GRCh38
rs774210844 Chr.3: 126109899-126109899 on Build GRCh38
rs746651860 Chr.3: 126110052-126110052 on Build GRCh38
rs974326698 Chr.3: 126110217-126110217 on Build GRCh38
rs960777195 Chr.3: 126110167-126110167 on Build GRCh38
rs376051813 Chr.3: 126109909-126109909 on Build GRCh38
rs41265741 Chr.3: 126109939-126109939 on Build GRCh38
rs775861728 Chr.3: 126109990-126109990 on Build GRCh38
rs773355120 Chr.3: 126110160-126110160 on Build GRCh38
rs969492689 Chr.3: 126109842-126109842 on Build GRCh38
rs941612354 Chr.3: 126110016-126110016 on Build GRCh38
rs751822141 Chr.3: 126109923-126109923 on Build GRCh38
rs370912743 Chr.3: 126109949-126109949 on Build GRCh38
rs767634341 Chr.3: 126110077-126110077 on Build GRCh38
rs893071728 Chr.3: 126110119-126110119 on Build GRCh38
rs963311761 Chr.3: 126110213-126110213 on Build GRCh38
rs780729377 Chr.3: 126109812-126109812 on Build GRCh38
rs754894527 Chr.3: 126110025-126110025 on Build GRCh38
rs145725373 Chr.3: 126110085-126110085 on Build GRCh38
rs550614591 Chr.3: 126109941-126109941 on Build GRCh38
rs1047276955 Chr.3: 126109860-126109860 on Build GRCh38
rs772779224 Chr.3: 126110050-126110050 on Build GRCh38
rs774901568 Chr.3: 126110060-126110060 on Build GRCh38
rs749247148 Chr.3: 126110020-126110020 on Build GRCh38
rs375361827 Chr.3: 126109965-126109965 on Build GRCh38
rs145429013 Chr.3: 126109954-126109954 on Build GRCh38
rs898933022 Chr.3: 126109784-126109784 on Build GRCh38
rs757912175 Chr.3: 126110124-126110124 on Build GRCh38
rs940986848 Chr.3: 126109885-126109885 on Build GRCh38
rs892397923 Chr.3: 126110067-126110067 on Build GRCh38
rs1022282085 Chr.3: 126109970-126109970 on Build GRCh38
rs745604085 Chr.3: 126109933-126109933 on Build GRCh38
rs768335580 Chr.3: 126109944-126109944 on Build GRCh38
rs928118895 Chr.3: 126109849-126109849 on Build GRCh38
rs145136430 Chr.3: 126109968-126109968 on Build GRCh38
rs559453611 Chr.3: 126109836-126109836 on Build GRCh38
rs201259609 Chr.3: 126109996-126109996 on Build GRCh38
rs780958924 Chr.3: 126110069-126110069 on Build GRCh38
rs761353929 Chr.3: 126109905-126109905 on Build GRCh38
rs769694739 Chr.3: 126109936-126109936 on Build GRCh38
rs202186993 Chr.3: 126110142-126110142 on Build GRCh38
rs373455389 Chr.3: 126109929-126109929 on Build GRCh38
rs778512889 Chr.3: 126110028-126110028 on Build GRCh38
rs760118673 Chr.3: 126109912-126109912 on Build GRCh38
rs768253751 Chr.3: 126109894-126109894 on Build GRCh38
rs756342517 Chr.3: 126110083-126110083 on Build GRCh38
rs774791551 Chr.3: 126110151-126110151 on Build GRCh38
rs987506785 Chr.3: 126110163-126110163 on Build GRCh38
rs374600464 Chr.3: 126110113-126110113 on Build GRCh38
rs746670036 Chr.3: 126110133-126110133 on Build GRCh38
rs747838364 Chr.3: 126109951-126109951 on Build GRCh38
rs775766256 Chr.3: 126110055-126110055 on Build GRCh38
rs920919650 Chr.3: 126110048-126110048 on Build GRCh38
rs770157307 Chr.3: 126110053-126110053 on Build GRCh38
rs1001635618 Chr.3: 126110150-126110150 on Build GRCh38
rs549433806 Chr.3: 126109834-126109834 on Build GRCh38
rs146931112 Chr.3: 126109910-126109910 on Build GRCh38
rs939625318 Chr.3: 126109857-126109857 on Build GRCh38
rs753643469 Chr.3: 126110096-126110096 on Build GRCh38
rs947786765 Chr.3: 126109993-126109993 on Build GRCh38
rs377289959 Chr.3: 126109913-126109913 on Build GRCh38
rs367736275 Chr.3: 126110013-126110013 on Build GRCh38
rs559987615 Chr.3: 126110056-126110056 on Build GRCh38
rs138816561 Chr.3: 126110022-126110022 on Build GRCh38
rs148110119 Chr.3: 126110097-126110097 on Build GRCh38
rs1022736590 Chr.3: 126110134-126110134 on Build GRCh38
rs747738867 Chr.3: 126109821-126109821 on Build GRCh38
rs955716381 Chr.3: 126110198-126110209 on Build GRCh38
rs563935343 Chr.3: 126109872-126109872 on Build GRCh38
rs199672127 Chr.3: 126109978-126109978 on Build GRCh38
rs769037003 Chr.3: 126110144-126110144 on Build GRCh38
rs137860341 Chr.3: 126110127-126110127 on Build GRCh38
rs777817670 Chr.3: 126109921-126109921 on Build GRCh38

View Full Product Details